Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49327126-49327397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TROAP
Entrez Gene ID
10024
Gene Name
trophinin associated protein
Gene Aliases
TASTIN
UniGene
Hs.524399
Gene Chromosome Location
Chr.12: 49322135-49331733 on Build GRCh38
Transcripts
NM_005480.3 XM_006719181.1 XM_011537723.1 XM_011537724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
147500 c.891+9C>T p.? Chr.12: 49327339-49327339 on Build GRCh38
3461654 c.770C>T p.A257V Chr.12: 49327209-49327209 on Build GRCh38
1582948 c.785A>G p.K262R Chr.12: 49327224-49327224 on Build GRCh38
7291239 c.839G>A p.G280D Chr.12: 49327278-49327278 on Build GRCh38
3461655 c.888C>T p.T296T Chr.12: 49327327-49327327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004566359 Chr.12: 49327285-49327285 on Build GRCh38
rs756331546 Chr.12: 49327215-49327215 on Build GRCh38
rs887510099 Chr.12: 49327284-49327284 on Build GRCh38
rs547105215 Chr.12: 49327262-49327262 on Build GRCh38
rs1015060081 Chr.12: 49327317-49327317 on Build GRCh38
rs754043819 Chr.12: 49327327-49327327 on Build GRCh38
rs745528769 Chr.12: 49327351-49327351 on Build GRCh38
rs756241721 Chr.12: 49327320-49327320 on Build GRCh38
rs201119814 Chr.12: 49327235-49327235 on Build GRCh38
rs779171030 Chr.12: 49327338-49327338 on Build GRCh38
rs78961476 Chr.12: 49327232-49327232 on Build GRCh38
rs370846897 Chr.12: 49327192-49327192 on Build GRCh38
rs772661052 Chr.12: 49327242-49327242 on Build GRCh38
rs764989091 Chr.12: 49327280-49327280 on Build GRCh38
rs976880397 Chr.12: 49327349-49327349 on Build GRCh38
rs750245684 Chr.12: 49327311-49327311 on Build GRCh38
rs769445972 Chr.12: 49327358-49327358 on Build GRCh38
rs759194516 Chr.12: 49327171-49327171 on Build GRCh38
rs759223770 Chr.12: 49327277-49327277 on Build GRCh38
rs747505295 Chr.12: 49327343-49327343 on Build GRCh38
rs73309977 Chr.12: 49327339-49327339 on Build GRCh38
rs775151278 Chr.12: 49327363-49327363 on Build GRCh38
rs1007718133 Chr.12: 49327265-49327265 on Build GRCh38
rs1048826856 Chr.12: 49327282-49327282 on Build GRCh38
rs758333624 Chr.12: 49327202-49327202 on Build GRCh38
rs142606809 Chr.12: 49327250-49327250 on Build GRCh38
rs750460331 Chr.12: 49327206-49327206 on Build GRCh38
rs138091021 Chr.12: 49327233-49327233 on Build GRCh38
rs748657209 Chr.12: 49327238-49327238 on Build GRCh38
rs762827543 Chr.12: 49327290-49327290 on Build GRCh38
rs377706259 Chr.12: 49327158-49327158 on Build GRCh38
rs900205328 Chr.12: 49327251-49327251 on Build GRCh38
rs763950798 Chr.12: 49327297-49327297 on Build GRCh38
rs766437381 Chr.12: 49327325-49327325 on Build GRCh38
rs778368270 Chr.12: 49327340-49327340 on Build GRCh38
rs1024325866 Chr.12: 49327151-49327151 on Build GRCh38
rs755361126 Chr.12: 49327234-49327234 on Build GRCh38
rs770633076 Chr.12: 49327346-49327346 on Build GRCh38
rs201645820 Chr.12: 49327205-49327205 on Build GRCh38
rs369672579 Chr.12: 49327337-49327337 on Build GRCh38
rs183043172 Chr.12: 49327258-49327258 on Build GRCh38
rs200842033 Chr.12: 49327191-49327191 on Build GRCh38
rs199775733 Chr.12: 49327203-49327203 on Build GRCh38
rs946552976 Chr.12: 49327196-49327196 on Build GRCh38
rs780189629 Chr.12: 49327224-49327224 on Build GRCh38
rs760429751 Chr.12: 49327161-49327161 on Build GRCh38
rs766091660 Chr.12: 49327165-49327165 on Build GRCh38
rs765067340 Chr.12: 49327181-49327181 on Build GRCh38
rs776572615 Chr.12: 49327169-49327169 on Build GRCh38
rs746454558 Chr.12: 49327253-49327253 on Build GRCh38
rs767556501 Chr.12: 49327348-49327348 on Build GRCh38
rs34098910 Chr.12: 49327149-49327149 on Build GRCh38
rs144038186 Chr.12: 49327289-49327289 on Build GRCh38

View Full Product Details