Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138596328-138596589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SVOPL
Entrez Gene ID
136306
Gene Name
SVOP like
Gene Aliases
-
UniGene
Hs.99414
Gene Chromosome Location
Chr.7: 138591689-138701547 on Build GRCh38
Transcripts
NM_001139456.1 NM_174959.3 XM_005250143.3 XM_011515797.2 XM_017011746.1 XM_017011747.1 XM_017011748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1086003 c.1445A>G p.E482G Chr.7: 138596439-138596439 on Build GRCh38
1699419 c.1412_1413TA>AC p.V471D Chr.7: 138596471-138596472 on Build GRCh38
1086005 c.1444G>A p.E482K Chr.7: 138596440-138596440 on Build GRCh38
1448384 c.911C>T p.A304V Chr.7: 138596517-138596517 on Build GRCh38
3995375 c.1397C>T p.S466L Chr.7: 138596487-138596487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774596808 Chr.7: 138596440-138596441 on Build GRCh38
rs767613967 Chr.7: 138596499-138596500 on Build GRCh38
rs111821973 Chr.7: 138596428-138596428 on Build GRCh38
rs979184809 Chr.7: 138596460-138596460 on Build GRCh38
rs748356494 Chr.7: 138596436-138596436 on Build GRCh38
rs367625070 Chr.7: 138596408-138596408 on Build GRCh38
rs141398137 Chr.7: 138596443-138596443 on Build GRCh38
rs746456994 Chr.7: 138596420-138596420 on Build GRCh38
rs979230692 Chr.7: 138596449-138596449 on Build GRCh38
rs371547841 Chr.7: 138596380-138596380 on Build GRCh38
rs766316586 Chr.7: 138596376-138596376 on Build GRCh38
rs1023463012 Chr.7: 138596406-138596406 on Build GRCh38
rs150828068 Chr.7: 138596468-138596468 on Build GRCh38
rs781683701 Chr.7: 138596418-138596418 on Build GRCh38
rs767968195 Chr.7: 138596478-138596478 on Build GRCh38
rs535241944 Chr.7: 138596472-138596472 on Build GRCh38
rs769148274 Chr.7: 138596444-138596444 on Build GRCh38
rs756672620 Chr.7: 138596489-138596489 on Build GRCh38
rs762451629 Chr.7: 138596509-138596509 on Build GRCh38
rs767560886 Chr.7: 138596542-138596543 on Build GRCh38
rs145071968 Chr.7: 138596427-138596427 on Build GRCh38
rs760392112 Chr.7: 138596371-138596371 on Build GRCh38
rs768908973 Chr.7: 138596386-138596386 on Build GRCh38
rs968240047 Chr.7: 138596455-138596455 on Build GRCh38
rs753961271 Chr.7: 138596501-138596501 on Build GRCh38
rs762486712 Chr.7: 138596457-138596457 on Build GRCh38
rs142898787 Chr.7: 138596523-138596523 on Build GRCh38
rs898629796 Chr.7: 138596375-138596375 on Build GRCh38
rs750701386 Chr.7: 138596483-138596483 on Build GRCh38
rs200673028 Chr.7: 138596502-138596502 on Build GRCh38
rs748407679 Chr.7: 138596512-138596512 on Build GRCh38
rs758094362 Chr.7: 138596403-138596403 on Build GRCh38
rs199985220 Chr.7: 138596516-138596516 on Build GRCh38
rs148292708 Chr.7: 138596441-138596441 on Build GRCh38
rs138625059 Chr.7: 138596471-138596471 on Build GRCh38
rs150951971 Chr.7: 138596405-138596405 on Build GRCh38
rs775097536 Chr.7: 138596451-138596451 on Build GRCh38
rs764841727 Chr.7: 138596388-138596388 on Build GRCh38
rs766021619 Chr.7: 138596545-138596545 on Build GRCh38
rs752127640 Chr.7: 138596396-138596396 on Build GRCh38
rs563467876 Chr.7: 138596385-138596385 on Build GRCh38
rs757641771 Chr.7: 138596417-138596417 on Build GRCh38
rs774498176 Chr.7: 138596431-138596431 on Build GRCh38
rs759385758 Chr.7: 138596383-138596383 on Build GRCh38
rs761966983 Chr.7: 138596483-138596487 on Build GRCh38
rs774211487 Chr.7: 138596368-138596368 on Build GRCh38
rs868659436 Chr.7: 138596387-138596387 on Build GRCh38
rs111250132 Chr.7: 138596464-138596464 on Build GRCh38
rs780353489 Chr.7: 138596499-138596499 on Build GRCh38
rs770776844 Chr.7: 138596440-138596440 on Build GRCh38
rs922861847 Chr.7: 138596518-138596518 on Build GRCh38
rs776814871 Chr.7: 138596467-138596467 on Build GRCh38
rs940246621 Chr.7: 138596368-138596368 on Build GRCh38

View Full Product Details