Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 92745397-92745662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHG7
Entrez Gene ID
440107
Gene Name
pleckstrin homology and RhoGEF domain containing G7
Gene Aliases
-
UniGene
Hs.591013
Gene Chromosome Location
Chr.12: 92736489-92772092 on Build GRCh38
Transcripts
NM_001004330.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6000804 c.208C>T p.R70* Chr.12: 92745484-92745484 on Build GRCh38
7384549 c.278G>C p.S93T Chr.12: 92745554-92745554 on Build GRCh38
3466455 c.207C>T p.F69F Chr.12: 92745483-92745483 on Build GRCh38
6482933 c.279C>G p.S93R Chr.12: 92745555-92745555 on Build GRCh38
5717698 c.285G>A p.R95R Chr.12: 92745561-92745561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567571587 Chr.12: 92745627-92745627 on Build GRCh38
rs766460424 Chr.12: 92745594-92745595 on Build GRCh38
rs747335271 Chr.12: 92745619-92745619 on Build GRCh38
rs765207809 Chr.12: 92745540-92745540 on Build GRCh38
rs569273789 Chr.12: 92745451-92745451 on Build GRCh38
rs116611401 Chr.12: 92745554-92745554 on Build GRCh38
rs1011107062 Chr.12: 92745422-92745422 on Build GRCh38
rs752432986 Chr.12: 92745561-92745561 on Build GRCh38
rs772644922 Chr.12: 92745599-92745599 on Build GRCh38
rs920690455 Chr.12: 92745623-92745623 on Build GRCh38
rs754956040 Chr.12: 92745442-92745442 on Build GRCh38
rs777100309 Chr.12: 92745629-92745629 on Build GRCh38
rs761131404 Chr.12: 92745512-92745512 on Build GRCh38
rs763248231 Chr.12: 92745563-92745564 on Build GRCh38
rs755750887 Chr.12: 92745465-92745465 on Build GRCh38
rs755867975 Chr.12: 92745571-92745571 on Build GRCh38
rs201467498 Chr.12: 92745585-92745585 on Build GRCh38
rs776747193 Chr.12: 92745529-92745529 on Build GRCh38
rs17790310 Chr.12: 92745572-92745572 on Build GRCh38
rs149205825 Chr.12: 92745477-92745477 on Build GRCh38
rs141629476 Chr.12: 92745575-92745575 on Build GRCh38
rs775980136 Chr.12: 92745501-92745501 on Build GRCh38
rs772750947 Chr.12: 92745497-92745497 on Build GRCh38
rs560682168 Chr.12: 92745560-92745560 on Build GRCh38
rs751472505 Chr.12: 92745441-92745441 on Build GRCh38
rs766297653 Chr.12: 92745440-92745440 on Build GRCh38
rs10859378 Chr.12: 92745458-92745458 on Build GRCh38
rs756985851 Chr.12: 92745583-92745583 on Build GRCh38
rs927771974 Chr.12: 92745518-92745518 on Build GRCh38
rs761901275 Chr.12: 92745535-92745535 on Build GRCh38
rs145096041 Chr.12: 92745485-92745485 on Build GRCh38
rs368591056 Chr.12: 92745489-92745489 on Build GRCh38
rs761801199 Chr.12: 92745427-92745427 on Build GRCh38
rs149128225 Chr.12: 92745527-92745527 on Build GRCh38
rs866307473 Chr.12: 92745488-92745488 on Build GRCh38
rs966822623 Chr.12: 92745466-92745466 on Build GRCh38
rs745442244 Chr.12: 92745589-92745589 on Build GRCh38
rs12305577 Chr.12: 92745616-92745616 on Build GRCh38
rs750541518 Chr.12: 92745545-92745545 on Build GRCh38
rs551879225 Chr.12: 92745484-92745484 on Build GRCh38

View Full Product Details