Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51681655-51681914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCG3
Entrez Gene ID
29106
Gene Name
secretogranin III
Gene Aliases
SGIII
UniGene
Hs.232618
Gene Chromosome Location
Chr.15: 51681353-51721026 on Build GRCh38
Transcripts
NM_001165257.1 NM_013243.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4540183 c.27G>A p.W9* Chr.15: 51681782-51681782 on Build GRCh38
4723931 c.13G>A p.G5R Chr.15: 51681768-51681768 on Build GRCh38
81685 c.36G>C p.V12V Chr.15: 51681791-51681791 on Build GRCh38
6772663 c.43C>T p.L15F Chr.15: 51681798-51681798 on Build GRCh38
701052 c.10C>T p.L4F Chr.15: 51681765-51681765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749361693 Chr.15: 51681737-51681737 on Build GRCh38
rs200963072 Chr.15: 51681733-51681733 on Build GRCh38
rs772807810 Chr.15: 51681830-51681830 on Build GRCh38
rs761318559 Chr.15: 51681717-51681717 on Build GRCh38
rs772382555 Chr.15: 51681756-51681756 on Build GRCh38
rs370895368 Chr.15: 51681815-51681815 on Build GRCh38
rs753408385 Chr.15: 51681713-51681713 on Build GRCh38
rs775982011 Chr.15: 51681759-51681759 on Build GRCh38
rs901045713 Chr.15: 51681692-51681692 on Build GRCh38
rs368414593 Chr.15: 51681868-51681868 on Build GRCh38
rs903703713 Chr.15: 51681768-51681768 on Build GRCh38
rs779042125 Chr.15: 51681884-51681884 on Build GRCh38
rs896442048 Chr.15: 51681679-51681679 on Build GRCh38
rs767467701 Chr.15: 51681876-51681876 on Build GRCh38
rs764826974 Chr.15: 51681720-51681720 on Build GRCh38
rs986497990 Chr.15: 51681843-51681843 on Build GRCh38
rs750042489 Chr.15: 51681722-51681722 on Build GRCh38
rs764781091 Chr.15: 51681825-51681825 on Build GRCh38
rs568430902 Chr.15: 51681744-51681744 on Build GRCh38
rs758175839 Chr.15: 51681723-51681723 on Build GRCh38
rs763752731 Chr.15: 51681708-51681708 on Build GRCh38
rs754923583 Chr.15: 51681729-51681729 on Build GRCh38
rs762654563 Chr.15: 51681836-51681836 on Build GRCh38
rs74013500 Chr.15: 51681878-51681878 on Build GRCh38
rs779057789 Chr.15: 51681738-51681738 on Build GRCh38
rs528456856 Chr.15: 51681688-51681688 on Build GRCh38
rs960992607 Chr.15: 51681810-51681810 on Build GRCh38
rs190544989 Chr.15: 51681829-51681829 on Build GRCh38
rs747316994 Chr.15: 51681769-51681769 on Build GRCh38
rs950276906 Chr.15: 51681887-51681887 on Build GRCh38
rs751361230 Chr.15: 51681848-51681848 on Build GRCh38
rs779820466 Chr.15: 51681724-51681724 on Build GRCh38
rs757324931 Chr.15: 51681881-51681881 on Build GRCh38
rs11553135 Chr.15: 51681767-51681767 on Build GRCh38
rs761251542 Chr.15: 51681824-51681824 on Build GRCh38
rs765960357 Chr.15: 51681844-51681844 on Build GRCh38
rs761255924 Chr.15: 51681705-51681705 on Build GRCh38
rs746129733 Chr.15: 51681888-51681888 on Build GRCh38
rs1028074974 Chr.15: 51681794-51681794 on Build GRCh38
rs769289615 Chr.15: 51681786-51681786 on Build GRCh38
rs953926938 Chr.15: 51681818-51681818 on Build GRCh38
rs1001073222 Chr.15: 51681785-51681785 on Build GRCh38
rs972457210 Chr.15: 51681849-51681849 on Build GRCh38
rs751557164 Chr.15: 51681727-51681727 on Build GRCh38

View Full Product Details