Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 38391575-38391804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
UniGene
Hs.592182
Gene Chromosome Location
Chr.7: 38383694-38631567 on Build GRCh38
Transcripts
NM_001635.3 NM_139316.2 XM_006715689.3 XM_006715690.3 XM_011515271.2 XM_017011995.1 XM_017011996.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3663175 c.1749C>A p.Y583* Chr.7: 38391751-38391751 on Build GRCh38
354585 c.1840G>A p.E614K Chr.7: 38391786-38391786 on Build GRCh38
6412074 c.1736C>G p.P579R Chr.7: 38391764-38391764 on Build GRCh38
6412073 c.1862C>G p.P621R Chr.7: 38391764-38391764 on Build GRCh38
7295510 c.1852G>A p.E618K Chr.7: 38391774-38391774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191249431 Chr.7: 38391725-38391725 on Build GRCh38
rs889591523 Chr.7: 38391603-38391603 on Build GRCh38
rs183053116 Chr.7: 38391768-38391768 on Build GRCh38
rs781427682 Chr.7: 38391687-38391687 on Build GRCh38
rs772072210 Chr.7: 38391710-38391711 on Build GRCh38
rs188472800 Chr.7: 38391654-38391654 on Build GRCh38
rs762519553 Chr.7: 38391751-38391751 on Build GRCh38
rs769557623 Chr.7: 38391733-38391733 on Build GRCh38
rs993833024 Chr.7: 38391691-38391691 on Build GRCh38
rs757057276 Chr.7: 38391655-38391656 on Build GRCh38
rs1801448 Chr.7: 38391772-38391772 on Build GRCh38
rs754193962 Chr.7: 38391779-38391779 on Build GRCh38
rs531672658 Chr.7: 38391728-38391728 on Build GRCh38
rs745448139 Chr.7: 38391732-38391732 on Build GRCh38
rs777544075 Chr.7: 38391709-38391709 on Build GRCh38
rs758413817 Chr.7: 38391697-38391697 on Build GRCh38
rs942528039 Chr.7: 38391606-38391606 on Build GRCh38
rs772663774 Chr.7: 38391764-38391764 on Build GRCh38
rs138082297 Chr.7: 38391784-38391784 on Build GRCh38
rs1044463278 Chr.7: 38391625-38391625 on Build GRCh38
rs775188616 Chr.7: 38391738-38391738 on Build GRCh38
rs927376923 Chr.7: 38391656-38391661 on Build GRCh38
rs137888041 Chr.7: 38391720-38391720 on Build GRCh38
rs34322947 Chr.7: 38391655-38391655 on Build GRCh38
rs367707025 Chr.7: 38391754-38391754 on Build GRCh38
rs780392713 Chr.7: 38391771-38391771 on Build GRCh38
rs1038118067 Chr.7: 38391614-38391614 on Build GRCh38
rs766581511 Chr.7: 38391776-38391776 on Build GRCh38

View Full Product Details