Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43050980-43051229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCA1
Entrez Gene ID
672
Gene Name
BRCA1, DNA repair associated
Gene Aliases
BRCAI, BRCC1, BROVCA1, FANCS, IRIS, PNCA4, PPP1R53, PSCP, RNF53
UniGene
Hs.194143
Gene Chromosome Location
Chr.17: 43044295-43125483 on Build GRCh38
Transcripts
NM_007294.3 NM_007297.3 NM_007298.3 NM_007299.3 NM_007300.3 NR_027676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6915174 c.5348G>T p.R1783M Chr.17: 43051110-43051110 on Build GRCh38
5009436 c.5347A>G p.R1783G Chr.17: 43051111-43051111 on Build GRCh38
6915175 c.5285G>T p.R1762M Chr.17: 43051110-43051110 on Build GRCh38
5752767 c.5315_5316ins23 p.F1772fs*29 Chr.17: 43051079-43051080 on Build GRCh38
3795649 c.5353C>A p.L1785I Chr.17: 43051105-43051105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs876660010 Chr.17: 43051067-43051067 on Build GRCh38
rs397509263 Chr.17: 43051062-43051062 on Build GRCh38
rs185646848 Chr.17: 43051139-43051139 on Build GRCh38
rs273901761 Chr.17: 43051085-43051085 on Build GRCh38
rs193149108 Chr.17: 43051137-43051137 on Build GRCh38
rs80357581 Chr.17: 43051087-43051087 on Build GRCh38
rs372391060 Chr.17: 43051050-43051050 on Build GRCh38
rs80357818 Chr.17: 43051074-43051075 on Build GRCh38
rs886040917 Chr.17: 43051057-43051064 on Build GRCh38
rs397509255 Chr.17: 43051106-43051106 on Build GRCh38
rs80358099 Chr.17: 43051118-43051118 on Build GRCh38
rs80357025 Chr.17: 43051083-43051083 on Build GRCh38
rs863224765 Chr.17: 43051086-43051086 on Build GRCh38
rs772616764 Chr.17: 43051055-43051055 on Build GRCh38
rs80357007 Chr.17: 43051107-43051107 on Build GRCh38
rs80358105 Chr.17: 43051131-43051131 on Build GRCh38
rs80357281 Chr.17: 43051104-43051104 on Build GRCh38
rs138493864 Chr.17: 43051091-43051091 on Build GRCh38
rs273901760 Chr.17: 43051119-43051121 on Build GRCh38
rs878853286 Chr.17: 43051062-43051064 on Build GRCh38
rs80357324 Chr.17: 43051078-43051078 on Build GRCh38
rs606231394 Chr.17: 43051108-43051109 on Build GRCh38
rs774813458 Chr.17: 43051044-43051044 on Build GRCh38
rs397509257 Chr.17: 43051089-43051089 on Build GRCh38
rs879255296 Chr.17: 43051066-43051066 on Build GRCh38
rs80358041 Chr.17: 43051062-43051062 on Build GRCh38
rs397509259 Chr.17: 43051085-43051085 on Build GRCh38
rs80357463 Chr.17: 43051098-43051098 on Build GRCh38
rs80356905 Chr.17: 43051113-43051113 on Build GRCh38
rs199858368 Chr.17: 43051148-43051148 on Build GRCh38
rs397509254 Chr.17: 43051121-43051123 on Build GRCh38
rs766614917 Chr.17: 43051060-43051060 on Build GRCh38
rs773655919 Chr.17: 43051056-43051056 on Build GRCh38
rs397509262 Chr.17: 43051071-43051072 on Build GRCh38
rs957547480 Chr.17: 43051168-43051168 on Build GRCh38
rs780795747 Chr.17: 43051031-43051031 on Build GRCh38
rs397509256 Chr.17: 43051102-43051102 on Build GRCh38
rs398122695 Chr.17: 43051068-43051068 on Build GRCh38
rs1800757 Chr.17: 43051069-43051069 on Build GRCh38
rs397509261 Chr.17: 43051080-43051080 on Build GRCh38
rs80358190 Chr.17: 43051032-43051032 on Build GRCh38
rs886040286 Chr.17: 43051096-43051096 on Build GRCh38
rs776854280 Chr.17: 43051162-43051162 on Build GRCh38
rs200407477 Chr.17: 43051041-43051041 on Build GRCh38
rs80357112 Chr.17: 43051063-43051063 on Build GRCh38
rs80357886 Chr.17: 43051109-43051110 on Build GRCh38
rs886040285 Chr.17: 43051113-43051113 on Build GRCh38
rs397509260 Chr.17: 43051084-43051085 on Build GRCh38
rs886040916 Chr.17: 43051062-43051084 on Build GRCh38
rs772224921 Chr.17: 43051011-43051011 on Build GRCh38
rs878853285 Chr.17: 43051119-43051119 on Build GRCh38
rs750040616 Chr.17: 43051115-43051115 on Build GRCh38
rs876660907 Chr.17: 43051108-43051108 on Build GRCh38
rs886039314 Chr.17: 43051099-43051099 on Build GRCh38
rs759867616 Chr.17: 43051067-43051067 on Build GRCh38
rs80357428 Chr.17: 43051077-43051077 on Build GRCh38
rs398122696 Chr.17: 43051065-43051065 on Build GRCh38
rs397507248 Chr.17: 43051105-43051106 on Build GRCh38
rs924705615 Chr.17: 43051186-43051186 on Build GRCh38
rs431825416 Chr.17: 43051093-43051093 on Build GRCh38
rs587781825 Chr.17: 43051084-43051085 on Build GRCh38
rs41293463 Chr.17: 43051071-43051071 on Build GRCh38
rs886040287 Chr.17: 43051094-43051095 on Build GRCh38
rs397509253 Chr.17: 43051119-43051119 on Build GRCh38
rs80357684 Chr.17: 43051111-43051111 on Build GRCh38
rs1022076404 Chr.17: 43051106-43051106 on Build GRCh38
rs730881497 Chr.17: 43051092-43051092 on Build GRCh38
rs397509258 Chr.17: 43051088-43051088 on Build GRCh38
rs886040289 Chr.17: 43051093-43051093 on Build GRCh38
rs80357959 Chr.17: 43051091-43051091 on Build GRCh38
rs80358166 Chr.17: 43051059-43051059 on Build GRCh38
rs80358148 Chr.17: 43051048-43051048 on Build GRCh38
rs1039237241 Chr.17: 43051014-43051014 on Build GRCh38
rs398122694 Chr.17: 43051110-43051110 on Build GRCh38
rs80357823 Chr.17: 43051075-43051076 on Build GRCh38
rs745573228 Chr.17: 43051035-43051035 on Build GRCh38
rs771092891 Chr.17: 43051149-43051149 on Build GRCh38
rs576727185 Chr.17: 43051084-43051084 on Build GRCh38
rs587781770 Chr.17: 43051074-43051074 on Build GRCh38
rs786203963 Chr.17: 43051120-43051120 on Build GRCh38
rs80358182 Chr.17: 43051061-43051061 on Build GRCh38
rs80357751 Chr.17: 43051066-43051067 on Build GRCh38
rs886040288 Chr.17: 43051094-43051094 on Build GRCh38

View Full Product Details