Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44071069-44071340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC3
Entrez Gene ID
92579
Gene Name
glucose-6-phosphatase catalytic subunit 3
Gene Aliases
SCN4, UGRP
UniGene
Hs.294005
Gene Chromosome Location
Chr.17: 44070700-44076344 on Build GRCh38
Transcripts
NM_001319945.1 NM_138387.3 NR_028581.1 NR_028582.1 XM_011525473.2 XM_011525474.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5554216 c.129C>A p.F43L Chr.17: 44071094-44071094 on Build GRCh38
1493966 c.195G>A p.E65E Chr.17: 44071160-44071160 on Build GRCh38
5450734 c.194A>G p.E65G Chr.17: 44071159-44071159 on Build GRCh38
3721873 c.164T>C p.I55T Chr.17: 44071129-44071129 on Build GRCh38
7127228 c.189C>T p.I63I Chr.17: 44071154-44071154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750408232 Chr.17: 44071234-44071234 on Build GRCh38
rs767499059 Chr.17: 44071223-44071223 on Build GRCh38
rs144594971 Chr.17: 44071154-44071154 on Build GRCh38
rs747508165 Chr.17: 44071185-44071185 on Build GRCh38
rs758088058 Chr.17: 44071142-44071142 on Build GRCh38
rs150477295 Chr.17: 44071134-44071134 on Build GRCh38
rs765390045 Chr.17: 44071205-44071205 on Build GRCh38
rs746408227 Chr.17: 44071252-44071252 on Build GRCh38
rs777080450 Chr.17: 44071241-44071241 on Build GRCh38
rs745318917 Chr.17: 44071159-44071159 on Build GRCh38
rs775525754 Chr.17: 44071210-44071210 on Build GRCh38
rs780837377 Chr.17: 44071157-44071157 on Build GRCh38
rs987290187 Chr.17: 44071304-44071304 on Build GRCh38
rs190571148 Chr.17: 44071247-44071247 on Build GRCh38
rs890590656 Chr.17: 44071170-44071170 on Build GRCh38
rs779667990 Chr.17: 44071238-44071238 on Build GRCh38
rs759787635 Chr.17: 44071136-44071136 on Build GRCh38
rs765317970 Chr.17: 44071139-44071139 on Build GRCh38
rs118203970 Chr.17: 44071106-44071106 on Build GRCh38
rs763978045 Chr.17: 44071217-44071217 on Build GRCh38
rs777135332 Chr.17: 44071197-44071197 on Build GRCh38
rs773663467 Chr.17: 44071108-44071108 on Build GRCh38
rs775224457 Chr.17: 44071095-44071095 on Build GRCh38
rs1020804767 Chr.17: 44071273-44071273 on Build GRCh38
rs1001863607 Chr.17: 44071231-44071231 on Build GRCh38
rs375273894 Chr.17: 44071166-44071166 on Build GRCh38
rs1004716621 Chr.17: 44071260-44071260 on Build GRCh38
rs752966267 Chr.17: 44071140-44071140 on Build GRCh38
rs772298089 Chr.17: 44071184-44071184 on Build GRCh38
rs898790675 Chr.17: 44071112-44071112 on Build GRCh38
rs755535333 Chr.17: 44071236-44071236 on Build GRCh38
rs763276794 Chr.17: 44071214-44071214 on Build GRCh38
rs200427598 Chr.17: 44071097-44071097 on Build GRCh38
rs954672505 Chr.17: 44071281-44071281 on Build GRCh38
rs755657110 Chr.17: 44071164-44071164 on Build GRCh38
rs757132842 Chr.17: 44071221-44071221 on Build GRCh38
rs371295401 Chr.17: 44071192-44071192 on Build GRCh38
rs548313333 Chr.17: 44071239-44071239 on Build GRCh38
rs751511819 Chr.17: 44071292-44071292 on Build GRCh38
rs761215881 Chr.17: 44071118-44071118 on Build GRCh38
rs971177305 Chr.17: 44071307-44071307 on Build GRCh38
rs769739687 Chr.17: 44071272-44071272 on Build GRCh38
rs762019955 Chr.17: 44071096-44071096 on Build GRCh38
rs1024194130 Chr.17: 44071290-44071290 on Build GRCh38
rs771287844 Chr.17: 44071240-44071240 on Build GRCh38
rs769441127 Chr.17: 44071175-44071175 on Build GRCh38
rs759465632 Chr.17: 44071198-44071198 on Build GRCh38
rs748995827 Chr.17: 44071172-44071172 on Build GRCh38
rs751460586 Chr.17: 44071220-44071220 on Build GRCh38
rs140585667 Chr.17: 44071131-44071131 on Build GRCh38
rs34878178 Chr.17: 44071152-44071152 on Build GRCh38
rs1001762610 Chr.17: 44071319-44071319 on Build GRCh38

View Full Product Details