Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42577041-42577268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC30
Entrez Gene ID
728621
Gene Name
coiled-coil domain containing 30
Gene Aliases
PFD6L, PFDN6L
UniGene
Hs.729640
Gene Chromosome Location
Chr.1: 42521240-42654664 on Build GRCh38
Transcripts
NM_001080850.2 XM_011542067.2 XM_011542068.2 XM_011542069.2 XM_011542070.2 XM_011542071.2 XM_017002225.1 XM_017002226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419236 c.998A>C p.K333T Chr.1: 42577162-42577162 on Build GRCh38
3738215 c.1059G>T p.E353D Chr.1: 42577223-42577223 on Build GRCh38
4577351 c.368C>T p.S123L Chr.1: 42577165-42577165 on Build GRCh38
3489731 c.1033G>A p.E345K Chr.1: 42577197-42577197 on Build GRCh38
4896475 c.923C>T p.T308M Chr.1: 42577087-42577087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756176570 Chr.1: 42577229-42577229 on Build GRCh38
rs372685385 Chr.1: 42577130-42577130 on Build GRCh38
rs61729282 Chr.1: 42577096-42577096 on Build GRCh38
rs772843460 Chr.1: 42577074-42577074 on Build GRCh38
rs901099155 Chr.1: 42577146-42577146 on Build GRCh38
rs748832067 Chr.1: 42577237-42577237 on Build GRCh38
rs551724357 Chr.1: 42577102-42577102 on Build GRCh38
rs774916473 Chr.1: 42577078-42577078 on Build GRCh38
rs1046091945 Chr.1: 42577115-42577115 on Build GRCh38
rs762884591 Chr.1: 42577151-42577151 on Build GRCh38
rs747874408 Chr.1: 42577112-42577113 on Build GRCh38
rs1028125578 Chr.1: 42577176-42577176 on Build GRCh38
rs376297024 Chr.1: 42577220-42577220 on Build GRCh38
rs781185046 Chr.1: 42577206-42577206 on Build GRCh38
rs750574938 Chr.1: 42577143-42577143 on Build GRCh38
rs909078897 Chr.1: 42577071-42577071 on Build GRCh38
rs368345722 Chr.1: 42577119-42577119 on Build GRCh38
rs571405961 Chr.1: 42577118-42577118 on Build GRCh38
rs766632929 Chr.1: 42577164-42577164 on Build GRCh38
rs999153043 Chr.1: 42577124-42577124 on Build GRCh38
rs894870972 Chr.1: 42577094-42577094 on Build GRCh38
rs770306943 Chr.1: 42577238-42577238 on Build GRCh38
rs751796287 Chr.1: 42577177-42577177 on Build GRCh38
rs1048640732 Chr.1: 42577103-42577103 on Build GRCh38
rs539831054 Chr.1: 42577216-42577216 on Build GRCh38
rs147422175 Chr.1: 42577088-42577088 on Build GRCh38
rs187080929 Chr.1: 42577120-42577120 on Build GRCh38
rs773805083 Chr.1: 42577081-42577081 on Build GRCh38
rs562709953 Chr.1: 42577087-42577087 on Build GRCh38
rs754845684 Chr.1: 42577195-42577195 on Build GRCh38
rs770626434 Chr.1: 42577079-42577079 on Build GRCh38

View Full Product Details