Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 172952020-172952293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNTL6
Entrez Gene ID
442117
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 6
Gene Aliases
GALNACT20, GALNT17, GalNAc-T6L
UniGene
Hs.386236
Gene Chromosome Location
Chr.4: 171812690-173040408 on Build GRCh38
Transcripts
NM_001034845.2 XM_011531993.2 XM_011531994.1 XM_011531995.1 XM_011531996.1 XM_011531997.1 XM_017008244.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1695261 c.1258G>A p.E420K Chr.4: 172952145-172952145 on Build GRCh38
1053238 c.1317G>T p.W439C Chr.4: 172952204-172952204 on Build GRCh38
6545922 c.1197C>T p.Y399Y Chr.4: 172952084-172952084 on Build GRCh38
1053237 c.1245C>A p.I415I Chr.4: 172952132-172952132 on Build GRCh38
5963718 c.1251C>T p.A417A Chr.4: 172952138-172952138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745725849 Chr.4: 172952252-172952252 on Build GRCh38
rs903500948 Chr.4: 172952051-172952051 on Build GRCh38
rs753980713 Chr.4: 172952091-172952091 on Build GRCh38
rs780663046 Chr.4: 172952123-172952123 on Build GRCh38
rs762114714 Chr.4: 172952093-172952093 on Build GRCh38
rs138303336 Chr.4: 172952179-172952179 on Build GRCh38
rs755485297 Chr.4: 172952122-172952122 on Build GRCh38
rs996572118 Chr.4: 172952099-172952099 on Build GRCh38
rs757975655 Chr.4: 172952094-172952094 on Build GRCh38
rs766704424 Chr.4: 172952208-172952208 on Build GRCh38
rs747244591 Chr.4: 172952085-172952086 on Build GRCh38
rs770722633 Chr.4: 172952162-172952162 on Build GRCh38
rs774299083 Chr.4: 172952077-172952077 on Build GRCh38
rs774025904 Chr.4: 172952163-172952163 on Build GRCh38
rs144094710 Chr.4: 172952207-172952207 on Build GRCh38
rs768114416 Chr.4: 172952040-172952040 on Build GRCh38
rs75539306 Chr.4: 172952104-172952104 on Build GRCh38
rs748907934 Chr.4: 172952235-172952235 on Build GRCh38
rs867780203 Chr.4: 172952099-172952109 on Build GRCh38
rs748918926 Chr.4: 172952153-172952153 on Build GRCh38
rs61737890 Chr.4: 172952105-172952105 on Build GRCh38
rs755470500 Chr.4: 172952220-172952220 on Build GRCh38
rs765252858 Chr.4: 172952194-172952194 on Build GRCh38
rs772194736 Chr.4: 172952174-172952174 on Build GRCh38
rs375135669 Chr.4: 172952228-172952228 on Build GRCh38
rs773442229 Chr.4: 172952195-172952195 on Build GRCh38
rs777182943 Chr.4: 172952230-172952230 on Build GRCh38
rs762849391 Chr.4: 172952067-172952067 on Build GRCh38
rs184699355 Chr.4: 172952211-172952211 on Build GRCh38
rs752183772 Chr.4: 172952224-172952224 on Build GRCh38
rs778756030 Chr.4: 172952240-172952240 on Build GRCh38
rs759581339 Chr.4: 172952081-172952081 on Build GRCh38
rs746973659 Chr.4: 172952268-172952268 on Build GRCh38
rs769845425 Chr.4: 172952272-172952272 on Build GRCh38
rs758739433 Chr.4: 172952102-172952102 on Build GRCh38
rs751178438 Chr.4: 172952261-172952261 on Build GRCh38
rs777532711 Chr.4: 172952152-172952152 on Build GRCh38
rs769402595 Chr.4: 172952130-172952130 on Build GRCh38
rs750729931 Chr.4: 172952100-172952100 on Build GRCh38
rs770754753 Chr.4: 172952074-172952074 on Build GRCh38
rs747738443 Chr.4: 172952047-172952047 on Build GRCh38
rs772106515 Chr.4: 172952253-172952253 on Build GRCh38
rs775517964 Chr.4: 172952256-172952256 on Build GRCh38
rs780681283 Chr.4: 172952044-172952044 on Build GRCh38
rs377199176 Chr.4: 172952239-172952239 on Build GRCh38
rs201080641 Chr.4: 172952085-172952085 on Build GRCh38
rs370310928 Chr.4: 172952164-172952164 on Build GRCh38
rs575271829 Chr.4: 172952060-172952060 on Build GRCh38
rs927386294 Chr.4: 172952258-172952258 on Build GRCh38
rs750015890 Chr.4: 172952049-172952049 on Build GRCh38
rs768050701 Chr.4: 172952223-172952223 on Build GRCh38

View Full Product Details