Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119527668-119527941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD80
Entrez Gene ID
941
Gene Name
CD80 molecule
Gene Aliases
B7, B7-1, B7.1, BB1, CD28LG, CD28LG1, LAB7
UniGene
Hs.838
Gene Chromosome Location
Chr.3: 119523909-119559709 on Build GRCh38
Transcripts
NM_005191.3 XM_011513327.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
397743 c.832G>C p.E278Q Chr.3: 119527806-119527806 on Build GRCh38
4559866 c.821G>A p.R274K Chr.3: 119527817-119527817 on Build GRCh38
6285017 c.827G>A p.R276K Chr.3: 119527811-119527811 on Build GRCh38
1693230 c.824G>A p.R275K Chr.3: 119527814-119527814 on Build GRCh38
1036754 c.840G>A p.L280L Chr.3: 119527798-119527798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748062893 Chr.3: 119527872-119527875 on Build GRCh38
rs780526034 Chr.3: 119527828-119527828 on Build GRCh38
rs781288699 Chr.3: 119527813-119527813 on Build GRCh38
rs766411633 Chr.3: 119527786-119527786 on Build GRCh38
rs888148986 Chr.3: 119527723-119527723 on Build GRCh38
rs757454283 Chr.3: 119527808-119527808 on Build GRCh38
rs772805829 Chr.3: 119527860-119527860 on Build GRCh38
rs771689032 Chr.3: 119527857-119527857 on Build GRCh38
rs752468937 Chr.3: 119527804-119527804 on Build GRCh38
rs1048506983 Chr.3: 119527705-119527705 on Build GRCh38
rs977629823 Chr.3: 119527799-119527799 on Build GRCh38
rs554371785 Chr.3: 119527820-119527820 on Build GRCh38
rs748473435 Chr.3: 119527853-119527853 on Build GRCh38
rs753880114 Chr.3: 119527787-119527787 on Build GRCh38
rs1037433039 Chr.3: 119527899-119527899 on Build GRCh38
rs373840808 Chr.3: 119527784-119527784 on Build GRCh38
rs754372048 Chr.3: 119527763-119527763 on Build GRCh38
rs576885399 Chr.3: 119527893-119527893 on Build GRCh38
rs774727630 Chr.3: 119527850-119527850 on Build GRCh38
rs761485937 Chr.3: 119527781-119527781 on Build GRCh38
rs776676186 Chr.3: 119527887-119527887 on Build GRCh38
rs773854424 Chr.3: 119527775-119527775 on Build GRCh38
rs74934063 Chr.3: 119527823-119527823 on Build GRCh38
rs145279608 Chr.3: 119527782-119527782 on Build GRCh38
rs371086780 Chr.3: 119527773-119527773 on Build GRCh38
rs540321705 Chr.3: 119527832-119527832 on Build GRCh38
rs377503363 Chr.3: 119527746-119527746 on Build GRCh38
rs926327867 Chr.3: 119527697-119527697 on Build GRCh38
rs760245789 Chr.3: 119527863-119527863 on Build GRCh38
rs781295426 Chr.3: 119527724-119527724 on Build GRCh38
rs760192600 Chr.3: 119527790-119527790 on Build GRCh38
rs749648891 Chr.3: 119527769-119527769 on Build GRCh38
rs924864193 Chr.3: 119527816-119527819 on Build GRCh38
rs768979372 Chr.3: 119527841-119527841 on Build GRCh38
rs765161589 Chr.3: 119527788-119527788 on Build GRCh38
rs771800284 Chr.3: 119527698-119527698 on Build GRCh38
rs35376118 Chr.3: 119527885-119527886 on Build GRCh38
rs180804720 Chr.3: 119527736-119527736 on Build GRCh38
rs953556555 Chr.3: 119527885-119527885 on Build GRCh38
rs199938816 Chr.3: 119527865-119527865 on Build GRCh38
rs1028175071 Chr.3: 119527855-119527855 on Build GRCh38

View Full Product Details