Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HFE
Entrez Gene ID
3077
Gene Name
hemochromatosis
Gene Aliases
HFE1, HH, HLA-H, MVCD7, TFQTL2
Chromosome Location
Chr.6: 26087281 - 26096216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139006.2 NP_620575.1 3 - 4 783 111
NM_139008.2 NP_620577.1 2 - 3 519 111
GenBank mRNA AF079407.1 - 3 - 4 659 111
AF079409.1 - 2 - 3 395 111

View Full Product Details