Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LTF
Entrez Gene ID
4057
Gene Name
lactotransferrin
Gene Aliases
GIG12, HEL110, HLF2, LF
Chromosome Location
Chr.3: 46436005 - 46485234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199149.1 NP_001186078.1 15 - 16 2097 71
NM_001321121.1 NP_001308050.1 15 - 16 2202 71
NM_001321122.1 NP_001308051.1 18 - 19 2441 71
NM_002343.5 NP_002334.2 15 - 16 2208 71
GenBank mRNA AF332168.1 - 15 - 16 1915 71
AK290859.1 - 15 - 16 1953 71
AK292813.1 - 15 - 16 1953 71
AK298035.1 - 15 - 16 2097 71
AK303995.1 - 7 - 8 816 71
AK316330.1 - 15 - 16 2017 71
AY137470.1 - 15 - 16 1915 71
AY165046.1 - 15 - 16 1927 71
AY178998.2 - 15 - 16 1915 71
AY360320.1 - 15 - 16 1910 71
AY493417.1 - 15 - 16 2025 71
AY875691.1 - 15 - 16 1915 71
BC015822.1 - 15 - 16 1957 71
BC015823.1 - 15 - 16 1959 71
BC022347.1 - 15 - 16 1953 71
EU779935.1 - 15 - 16 1915 71
EU794669.1 - 15 - 16 1950 71
KF992208.1 - 15 - 16 1927 71
KT006756.1 - 15 - 16 1915 71
M18642.1 - 5 - 6 610 71
M83202.1 - 15 - 16 1922 71
M83205.1 - 15 - 16 1901 71
M93150.1 - 15 - 16 1932 71
U07643.1 - 15 - 16 2012 71
X52941.1 - 15 - 16 1909 71
X53961.1 - 15 - 16 2209 71

View Full Product Details