Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRAS
Entrez Gene ID
3845
Gene Name
KRAS proto-oncogene, GTPase
Gene Aliases
C-K-RAS, CFC2, K-RAS2A, K-RAS2B, K-RAS4A, K-RAS4B, KI-RAS, KRAS1, KRAS2, NS, NS3, RALD, RASK2
Chromosome Location
Chr.12: 25204789 - 25252093 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033360.3 NP_203524.1 5 - 6 768 116
XM_006719069.3 XP_006719132.1 5 - 6 774 116

View Full Product Details