Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MX2
Entrez Gene ID
4600
Gene Name
MX dynamin like GTPase 2
Gene Aliases
MXB
Chromosome Location
Chr.21: 41361986 - 41408943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002463.1 NP_002454.1 3 - 4 548 114
XM_005260983.4 XP_005261040.1 3 - 4 646 114
XM_005260984.1 XP_005261041.1 3 - 4 826 114
XM_011529571.1 XP_011527873.1 3 - 4 581 114
XM_011529572.1 XP_011527874.1 3 - 4 684 114
GenBank mRNA AK308060.1 - 3 - 4 545 114
BC035293.1 - 2 - 3 510 114
M30818.1 - 3 - 4 548 114
M33883.1 - 2 - 3 361 114

View Full Product Details