Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
GAPDH
Entrez Gene ID
2597
Gene Name
glyceraldehyde-3-phosphate dehydrogenase
Gene Aliases
G3PD, GAPD, HEL-S-162eP
Chromosome Location
Chr.12: 6534405 - 6538375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256799.2 NP_001243728.1 7 - 7 765 95
NM_001289745.1 NP_001276674.1 8 - 8 823 95
NM_001289746.1 NP_001276675.1 7 - 7 717 95
NM_002046.5 NP_002037.2 8 - 8 731 95
GenBank mRNA AB062273.1 - 8 - 8 595 95
AF261085.1 - 8 - 8 646 95
AK026525.1 - 8 - 8 668 95
AK308198.1 - 8 - 8 619 95
AY007133.1 - 8 - 8 595 95
AY633612.1 - 7 - 7 567 95
BC001601.1 - 8 - 8 597 95
BC004109.2 - 8 - 8 595 95
BC009081.1 - 8 - 8 604 95
BC013310.2 - 8 - 8 603 95
BC020308.1 - 7 - 7 581 95
BC023632.2 - 7 - 7 567 95
BC025925.1 - 8 - 8 600 95
BC026907.1 - 7 - 7 581 95
BC029340.1 - 8 - 8 589 95
BC029618.1 - 8 - 8 597 95
BC083511.1 - 8 - 8 595 95
BI463134.1 - 7 - 7 748 96
BT006893.1 - 7 - 7 543 95
CR407671.1 - 7 - 7 543 95
DQ403057.1 - 6 - 6 468 95
EF036498.1 - 4 - 4 281 95
EU668321.1 - 8 - 8 645 95
J02642.1 - 7 - 7 544 95
M17851.1 - 7 - 7 576 95
M33197.1 - 8 - 8 603 95
X01677.1 - 7 - 7 603 95
X53778.1 - 8 - 8 599 94

View Full Product Details