Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Braf
Entrez Gene ID
109880
Gene Name
Braf transforming gene
Gene Aliases
9930012E13Rik, AA120551, AA387315, AA473386, B-raf, Braf-2, Braf2, C230098H17, C87398, D6Ertd631e
Chromosome Location
Chr.6: 39603231 - 39725656 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139294.5 NP_647455.3 2 - 3 452 73
GenBank mRNA AK140431.1 - 2 - 3 453 73
BF466677.1 - 2 - 3 315 73

View Full Product Details