Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTPS1
Entrez Gene ID
695498
Gene Name
CTP synthase 1
Gene Aliases
CTPS, EGK_00626
Chromosome Location
Chr.1: 183557621 - 183590705 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015136681.2 XP_014992167.1 - 1654 66
XM_015136688.2 XP_014992174.1 - 1619 66
XM_028835304.1 XP_028691137.1 - 1240 66
GenBank mRNA JU322260.1 - - 1188 66
JU474436.1 - - 1210 66
JV046270.1 - - 1239 66
JV643321.1 - - 1011 66

View Full Product Details