Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppid
Entrez Gene ID
361967
Gene Name
peptidylprolyl isomerase D
Gene Aliases
Cyp-40, CypD
Chromosome Location
Chr.2: 178354830 - 178366843 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004279.1 NP_001004279.1 2 - 3 282 79
GenBank mRNA BC076386.1 - 2 - 3 282 79
FQ214826.1 - 2 - 3 285 79

Gene Symbol
Ppidl1
Entrez Gene ID
501204
Gene Name
peptidylprolyl isomerase D-like 1
Gene Aliases
RGD1560149
Chromosome Location
Chr.9: 121456769 - 121458035 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750757.3 XP_003750805.1 1 - 1 230 79

View Full Product Details