Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VHL
Entrez Gene ID
494000
Gene Name
von Hippel-Lindau tumor suppressor
Gene Aliases
-
Chromosome Location
Chr.20: 8206616 - 8211347 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008552.1 NP_001008552.1 - 287 98
XM_014121534.2 XP_013977009.1 - 311 98
GenBank mRNA AY764285.1 - - 287 98

View Full Product Details