Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npy
Entrez Gene ID
100758436
Gene Name
neuropeptide Y
Gene Aliases
I79_004427
Chromosome Location
Chr.Un|NW_003613695: 2299175 - 2305021 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498558.1 XP_003498606.1 1 - 2 182 68

View Full Product Details