Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73648642-73649144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHB1
Entrez Gene ID
58473
Gene Name
pleckstrin homology domain containing B1
Gene Aliases
KPL1, PHR1, PHRET1
UniGene
Hs.445489
Gene Chromosome Location
Chr.11: 73646178-73662819 on Build GRCh38
Transcripts
NM_001130033.1 NM_001130034.1 NM_001130035.1 NM_001130036.1 NM_021200.2 XM_011545192.2 XM_011545193.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6252801 c.24G>T p.P8P Chr.11: 73649017-73649017 on Build GRCh38
2017048 c.76G>A p.G26S Chr.11: 73649069-73649069 on Build GRCh38
5636018 c.93G>C p.Q31H Chr.11: 73649086-73649086 on Build GRCh38
4715615 c.69G>T p.V23V Chr.11: 73649062-73649062 on Build GRCh38
4983244 c.34G>A p.A12T Chr.11: 73649027-73649027 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368334880 Chr.11: 73649026-73649026 on Build GRCh38
rs113579209 Chr.11: 73648726-73648726 on Build GRCh38
rs150366990 Chr.11: 73648824-73648824 on Build GRCh38
rs1037975536 Chr.11: 73649123-73649123 on Build GRCh38
rs968075621 Chr.11: 73648733-73648733 on Build GRCh38
rs778642589 Chr.11: 73649059-73649059 on Build GRCh38
rs774178155 Chr.11: 73649016-73649016 on Build GRCh38
rs35809621 Chr.11: 73648866-73648867 on Build GRCh38
rs118101918 Chr.11: 73648740-73648740 on Build GRCh38
rs576641014 Chr.11: 73648949-73648949 on Build GRCh38
rs529821522 Chr.11: 73649061-73649061 on Build GRCh38
rs992751554 Chr.11: 73648841-73648841 on Build GRCh38
rs537111663 Chr.11: 73648964-73648964 on Build GRCh38
rs577064718 Chr.11: 73648837-73648837 on Build GRCh38
rs769851852 Chr.11: 73649079-73649079 on Build GRCh38
rs771500638 Chr.11: 73648871-73648871 on Build GRCh38
rs761597650 Chr.11: 73649017-73649017 on Build GRCh38
rs574882465 Chr.11: 73648925-73648925 on Build GRCh38
rs2509065 Chr.11: 73648757-73648757 on Build GRCh38
rs141371201 Chr.11: 73648697-73648697 on Build GRCh38
rs891725373 Chr.11: 73648708-73648708 on Build GRCh38
rs374531350 Chr.11: 73648977-73648977 on Build GRCh38
rs745319822 Chr.11: 73648800-73648800 on Build GRCh38
rs2512634 Chr.11: 73648682-73648682 on Build GRCh38
rs754747578 Chr.11: 73649113-73649113 on Build GRCh38
rs953847444 Chr.11: 73648879-73648879 on Build GRCh38
rs138039493 Chr.11: 73648996-73648996 on Build GRCh38
rs891653694 Chr.11: 73649013-73649013 on Build GRCh38
rs1049348097 Chr.11: 73648955-73648955 on Build GRCh38
rs780447046 Chr.11: 73649096-73649096 on Build GRCh38
rs767251915 Chr.11: 73649024-73649024 on Build GRCh38
rs546756133 Chr.11: 73649068-73649068 on Build GRCh38
rs1010176515 Chr.11: 73648710-73648710 on Build GRCh38
rs367691921 Chr.11: 73648984-73648984 on Build GRCh38
rs1028504094 Chr.11: 73648868-73648868 on Build GRCh38
rs910983827 Chr.11: 73648966-73648966 on Build GRCh38
rs766229210 Chr.11: 73649028-73649028 on Build GRCh38
rs774255736 Chr.11: 73649097-73649097 on Build GRCh38
rs745939806 Chr.11: 73648974-73648974 on Build GRCh38
rs777120153 Chr.11: 73649063-73649063 on Build GRCh38
rs973042104 Chr.11: 73648939-73648939 on Build GRCh38
rs776021852 Chr.11: 73648942-73648942 on Build GRCh38
rs759533621 Chr.11: 73649114-73649114 on Build GRCh38
rs372832415 Chr.11: 73648995-73648995 on Build GRCh38
rs749762591 Chr.11: 73649098-73649098 on Build GRCh38
rs1019444617 Chr.11: 73648887-73648887 on Build GRCh38
rs1008021352 Chr.11: 73648685-73648685 on Build GRCh38
rs776062869 Chr.11: 73648992-73648992 on Build GRCh38
rs770799304 Chr.11: 73649085-73649086 on Build GRCh38
rs762333274 Chr.11: 73649111-73649111 on Build GRCh38
rs760225040 Chr.11: 73649027-73649027 on Build GRCh38
rs918070748 Chr.11: 73648873-73648873 on Build GRCh38
rs747439300 Chr.11: 73648943-73648943 on Build GRCh38
rs563072852 Chr.11: 73648940-73648940 on Build GRCh38
rs1033766800 Chr.11: 73648794-73648794 on Build GRCh38
rs754755105 Chr.11: 73649046-73649046 on Build GRCh38
rs113878239 Chr.11: 73648885-73648885 on Build GRCh38
rs545963775 Chr.11: 73648917-73648917 on Build GRCh38
rs769254783 Chr.11: 73649102-73649102 on Build GRCh38
rs560284419 Chr.11: 73649122-73649122 on Build GRCh38
rs943778969 Chr.11: 73648986-73648986 on Build GRCh38
rs1021949182 Chr.11: 73648732-73648732 on Build GRCh38
rs753753316 Chr.11: 73649044-73649044 on Build GRCh38
rs372276436 Chr.11: 73649069-73649069 on Build GRCh38

View Full Product Details