Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73876666-73877173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COA4
Entrez Gene ID
51287
Gene Name
cytochrome c oxidase assembly factor 4 homolog
Gene Aliases
CHCHD8, CMC3, E2IG2
UniGene
Hs.475387
Gene Chromosome Location
Chr.11: 73870258-73876901 on Build GRCh38
Transcripts
NM_016565.2 XM_017017884.1 NM_001267803.1 NM_001267804.1 NM_001267805.1 NM_001267806.1 NM_025155.2 XM_017018357.1 XM_017018358.1

Gene Symbol
PAAF1
Entrez Gene ID
80227
Gene Name
proteasomal ATPase associated factor 1
Gene Aliases
PAAF, Rpn14, WDR71
UniGene
Hs.525017
Gene Chromosome Location
Chr.11: 73876699-73927736 on Build GRCh38
Transcripts
NM_016565.2 XM_017017884.1 NM_001267803.1 NM_001267804.1 NM_001267805.1 NM_001267806.1 NM_025155.2 XM_017018357.1 XM_017018358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6401705 c.35C>T p.A12V Chr.11: 73877056-73877056 on Build GRCh38
5970376 c.40G>T p.A14S Chr.11: 73877061-73877061 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1052350904 Chr.11: 73876946-73876946 on Build GRCh38
rs1050060487 Chr.11: 73876702-73876702 on Build GRCh38
rs1008778563 Chr.11: 73876716-73876716 on Build GRCh38
rs551884659 Chr.11: 73876707-73876707 on Build GRCh38
rs995912731 Chr.11: 73876993-73876994 on Build GRCh38
rs561262144 Chr.11: 73876891-73876892 on Build GRCh38
rs963158412 Chr.11: 73876803-73876803 on Build GRCh38
rs920722367 Chr.11: 73877083-73877083 on Build GRCh38
rs950168724 Chr.11: 73877132-73877132 on Build GRCh38
rs1034313077 Chr.11: 73876800-73876800 on Build GRCh38
rs534910737 Chr.11: 73876822-73876822 on Build GRCh38
rs757409908 Chr.11: 73877043-73877043 on Build GRCh38
rs979094493 Chr.11: 73877061-73877061 on Build GRCh38
rs985893626 Chr.11: 73877087-73877087 on Build GRCh38
rs932179063 Chr.11: 73877084-73877084 on Build GRCh38
rs192274848 Chr.11: 73877077-73877077 on Build GRCh38
rs967317303 Chr.11: 73876835-73876835 on Build GRCh38
rs527945936 Chr.11: 73876742-73876742 on Build GRCh38
rs993260286 Chr.11: 73877048-73877048 on Build GRCh38
rs920774755 Chr.11: 73876826-73876826 on Build GRCh38
rs1022049179 Chr.11: 73876969-73876969 on Build GRCh38
rs76332820 Chr.11: 73876808-73876808 on Build GRCh38
rs758460343 Chr.11: 73876891-73876891 on Build GRCh38
rs2010583 Chr.11: 73876952-73876952 on Build GRCh38
rs1005659600 Chr.11: 73876796-73876796 on Build GRCh38
rs11217 Chr.11: 73876788-73876788 on Build GRCh38
rs887349860 Chr.11: 73876906-73876906 on Build GRCh38
rs954468723 Chr.11: 73877022-73877022 on Build GRCh38
rs987394762 Chr.11: 73876689-73876689 on Build GRCh38
rs925780916 Chr.11: 73876842-73876842 on Build GRCh38
rs201724282 Chr.11: 73876997-73876997 on Build GRCh38
rs753718658 Chr.11: 73876971-73876971 on Build GRCh38
rs1014429701 Chr.11: 73876786-73876786 on Build GRCh38
rs913042706 Chr.11: 73876693-73876693 on Build GRCh38
rs944406780 Chr.11: 73876925-73876925 on Build GRCh38
rs556219943 Chr.11: 73877074-73877074 on Build GRCh38
rs950035351 Chr.11: 73876873-73876873 on Build GRCh38
rs777603917 Chr.11: 73876696-73876696 on Build GRCh38
rs895812109 Chr.11: 73876766-73876766 on Build GRCh38
rs917224250 Chr.11: 73876864-73876864 on Build GRCh38
rs1017551316 Chr.11: 73876797-73876797 on Build GRCh38
rs141282994 Chr.11: 73876985-73876985 on Build GRCh38
rs757562367 Chr.11: 73876794-73876794 on Build GRCh38
rs890126643 Chr.11: 73876711-73876711 on Build GRCh38
rs548045017 Chr.11: 73876970-73876970 on Build GRCh38
rs756965775 Chr.11: 73876746-73876746 on Build GRCh38
rs978610664 Chr.11: 73876838-73876838 on Build GRCh38
rs779057893 Chr.11: 73876799-73876799 on Build GRCh38
rs371421703 Chr.11: 73877139-73877139 on Build GRCh38
rs1034881257 Chr.11: 73877040-73877040 on Build GRCh38
rs968743805 Chr.11: 73876764-73876764 on Build GRCh38
rs756482117 Chr.11: 73877086-73877086 on Build GRCh38
rs960054375 Chr.11: 73877044-73877044 on Build GRCh38
rs749135118 Chr.11: 73876732-73876732 on Build GRCh38
rs905034600 Chr.11: 73876977-73876977 on Build GRCh38
rs960355293 Chr.11: 73876806-73876806 on Build GRCh38
rs892293345 Chr.11: 73876738-73876738 on Build GRCh38
rs568364042 Chr.11: 73876984-73876984 on Build GRCh38
rs1029179872 Chr.11: 73876996-73876996 on Build GRCh38
rs1050175193 Chr.11: 73876861-73876861 on Build GRCh38
rs539591671 Chr.11: 73877069-73877069 on Build GRCh38
rs149131885 Chr.11: 73876819-73876820 on Build GRCh38
rs776979896 Chr.11: 73877100-73877100 on Build GRCh38
rs148117364 Chr.11: 73876846-73876846 on Build GRCh38
rs78960781 Chr.11: 73876890-73876890 on Build GRCh38
rs1008555732 Chr.11: 73877024-73877024 on Build GRCh38
rs781250049 Chr.11: 73877067-73877067 on Build GRCh38
rs769087936 Chr.11: 73877127-73877127 on Build GRCh38
rs371802088 Chr.11: 73876815-73876815 on Build GRCh38
rs201021789 Chr.11: 73877075-73877075 on Build GRCh38
rs554147808 Chr.11: 73876987-73876987 on Build GRCh38
rs1028624910 Chr.11: 73876813-73876813 on Build GRCh38
rs868834286 Chr.11: 73876748-73876748 on Build GRCh38
rs187071361 Chr.11: 73876719-73876719 on Build GRCh38
rs945882418 Chr.11: 73876710-73876710 on Build GRCh38
rs747497342 Chr.11: 73877018-73877018 on Build GRCh38
rs993087053 Chr.11: 73876807-73876807 on Build GRCh38
rs1010790271 Chr.11: 73876966-73876966 on Build GRCh38
rs144695799 Chr.11: 73877113-73877114 on Build GRCh38
rs1019996232 Chr.11: 73877031-73877031 on Build GRCh38

View Full Product Details