Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74006831-74007329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCP3
Entrez Gene ID
7352
Gene Name
uncoupling protein 3
Gene Aliases
SLC25A9
UniGene
Hs.101337
Gene Chromosome Location
Chr.11: 74000281-74009237 on Build GRCh38
Transcripts
NM_003356.3 NM_022803.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6269913 c.33C>A p.P11P Chr.11: 74007010-74007010 on Build GRCh38
1579252 c.88G>A p.V30I Chr.11: 74006955-74006955 on Build GRCh38
1298594 c.12G>A p.L4L Chr.11: 74007031-74007031 on Build GRCh38
1475890 c.40G>A p.A14T Chr.11: 74007003-74007003 on Build GRCh38
4964817 c.58G>C p.A20P Chr.11: 74006985-74006985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958262713 Chr.11: 74007172-74007172 on Build GRCh38
rs763087706 Chr.11: 74006915-74006915 on Build GRCh38
rs569451034 Chr.11: 74007186-74007186 on Build GRCh38
rs552472433 Chr.11: 74007078-74007078 on Build GRCh38
rs564239289 Chr.11: 74007269-74007269 on Build GRCh38
rs753543579 Chr.11: 74006951-74006952 on Build GRCh38
rs143098507 Chr.11: 74007287-74007287 on Build GRCh38
rs764423149 Chr.11: 74007042-74007042 on Build GRCh38
rs756938226 Chr.11: 74007066-74007066 on Build GRCh38
rs534397046 Chr.11: 74007229-74007229 on Build GRCh38
rs771739253 Chr.11: 74006878-74006878 on Build GRCh38
rs1020113364 Chr.11: 74007099-74007099 on Build GRCh38
rs147975989 Chr.11: 74007262-74007262 on Build GRCh38
rs753185649 Chr.11: 74007028-74007028 on Build GRCh38
rs773606533 Chr.11: 74007007-74007007 on Build GRCh38
rs1032581191 Chr.11: 74007145-74007145 on Build GRCh38
rs201532497 Chr.11: 74006922-74006922 on Build GRCh38
rs779123450 Chr.11: 74006872-74006872 on Build GRCh38
rs1008269903 Chr.11: 74007090-74007090 on Build GRCh38
rs765735895 Chr.11: 74006933-74006933 on Build GRCh38
rs1004085360 Chr.11: 74007010-74007010 on Build GRCh38
rs769726795 Chr.11: 74006912-74006912 on Build GRCh38
rs1043811762 Chr.11: 74006927-74006927 on Build GRCh38
rs1019187474 Chr.11: 74007135-74007135 on Build GRCh38
rs988737997 Chr.11: 74007189-74007189 on Build GRCh38
rs990963959 Chr.11: 74007182-74007182 on Build GRCh38
rs777336013 Chr.11: 74006884-74006884 on Build GRCh38
rs370757772 Chr.11: 74006898-74006898 on Build GRCh38
rs937626824 Chr.11: 74007268-74007268 on Build GRCh38
rs898428016 Chr.11: 74007253-74007253 on Build GRCh38
rs753662787 Chr.11: 74007238-74007238 on Build GRCh38
rs70965455 Chr.11: 74006971-74006971 on Build GRCh38
rs199679366 Chr.11: 74006985-74006985 on Build GRCh38
rs754417229 Chr.11: 74006926-74006926 on Build GRCh38
rs745773835 Chr.11: 74006910-74006910 on Build GRCh38
rs755409260 Chr.11: 74006867-74006867 on Build GRCh38
rs748496766 Chr.11: 74006876-74006876 on Build GRCh38
rs371361898 Chr.11: 74006956-74006956 on Build GRCh38
rs978052260 Chr.11: 74007131-74007131 on Build GRCh38
rs979181746 Chr.11: 74007248-74007248 on Build GRCh38
rs756995001 Chr.11: 74006985-74006985 on Build GRCh38
rs1019723226 Chr.11: 74007307-74007307 on Build GRCh38
rs953339169 Chr.11: 74007080-74007080 on Build GRCh38
rs939902583 Chr.11: 74007301-74007301 on Build GRCh38
rs775441399 Chr.11: 74006984-74006984 on Build GRCh38
rs753004564 Chr.11: 74006943-74006943 on Build GRCh38
rs199727434 Chr.11: 74006925-74006925 on Build GRCh38
rs746657933 Chr.11: 74006970-74006970 on Build GRCh38
rs755461584 Chr.11: 74006929-74006929 on Build GRCh38
rs770412541 Chr.11: 74006889-74006889 on Build GRCh38
rs765673001 Chr.11: 74007019-74007019 on Build GRCh38
rs916721185 Chr.11: 74007184-74007184 on Build GRCh38
rs760955370 Chr.11: 74007009-74007009 on Build GRCh38
rs1036674387 Chr.11: 74007250-74007250 on Build GRCh38
rs573218240 Chr.11: 74006924-74006924 on Build GRCh38
rs199939445 Chr.11: 74006955-74006955 on Build GRCh38
rs575619046 Chr.11: 74006973-74006973 on Build GRCh38
rs375349129 Chr.11: 74006892-74006892 on Build GRCh38
rs751874952 Chr.11: 74007059-74007059 on Build GRCh38
rs1007771164 Chr.11: 74007130-74007130 on Build GRCh38
rs141482270 Chr.11: 74007032-74007032 on Build GRCh38
rs746533067 Chr.11: 74006886-74006886 on Build GRCh38
rs143470338 Chr.11: 74007210-74007210 on Build GRCh38
rs755640693 Chr.11: 74007085-74007085 on Build GRCh38
rs1028086908 Chr.11: 74007053-74007053 on Build GRCh38
rs181883754 Chr.11: 74006980-74006980 on Build GRCh38
rs762770256 Chr.11: 74006916-74006916 on Build GRCh38
rs766573609 Chr.11: 74007015-74007015 on Build GRCh38
rs8179180 Chr.11: 74007018-74007018 on Build GRCh38
rs148847746 Chr.11: 74007016-74007016 on Build GRCh38
rs917881261 Chr.11: 74007288-74007288 on Build GRCh38
rs200938041 Chr.11: 74007077-74007077 on Build GRCh38
rs894092312 Chr.11: 74007171-74007171 on Build GRCh38
rs70965454 Chr.11: 74007006-74007006 on Build GRCh38
rs928366680 Chr.11: 74007214-74007214 on Build GRCh38
rs754295074 Chr.11: 74006866-74006866 on Build GRCh38
rs536504852 Chr.11: 74006860-74006860 on Build GRCh38

View Full Product Details