Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78172372-78172892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD21
Entrez Gene ID
283219
Gene Name
potassium channel tetramerization domain containing 21
Gene Aliases
KCASH2
UniGene
Hs.528439
Gene Chromosome Location
Chr.11: 78171243-78188854 on Build GRCh38
Transcripts
NM_001029859.2 XM_005273925.3 XM_005273928.1 XM_006718516.3 XM_006718517.2 XM_006718518.3 XM_011544955.2 NR_102280.1 NR_102281.1

Gene Symbol
KCTD21-AS1
Entrez Gene ID
100289388
Gene Name
KCTD21 antisense RNA 1
Gene Aliases
-
UniGene
Hs.628183
Gene Chromosome Location
Chr.11: 78139793-78173977 on Build GRCh38
Transcripts
NM_001029859.2 XM_005273925.3 XM_005273928.1 XM_006718516.3 XM_006718517.2 XM_006718518.3 XM_011544955.2 NR_102280.1 NR_102281.1

SNP Detail

SNP ID Chromosome Location
rs1005533486 Chr.11: 78172773-78172773 on Build GRCh38
rs536252690 Chr.11: 78172445-78172445 on Build GRCh38
rs866639888 Chr.11: 78172473-78172473 on Build GRCh38
rs576989615 Chr.11: 78172864-78172864 on Build GRCh38
rs184793098 Chr.11: 78172670-78172670 on Build GRCh38
rs769935885 Chr.11: 78172406-78172406 on Build GRCh38
rs778067804 Chr.11: 78172513-78172515 on Build GRCh38
rs913575656 Chr.11: 78172848-78172848 on Build GRCh38
rs1053429104 Chr.11: 78172652-78172652 on Build GRCh38
rs926424957 Chr.11: 78172803-78172803 on Build GRCh38
rs369908243 Chr.11: 78172782-78172782 on Build GRCh38
rs537585897 Chr.11: 78172414-78172414 on Build GRCh38
rs767977930 Chr.11: 78172802-78172802 on Build GRCh38
rs961919236 Chr.11: 78172513-78172513 on Build GRCh38
rs1004146076 Chr.11: 78172822-78172822 on Build GRCh38
rs945622189 Chr.11: 78172662-78172662 on Build GRCh38
rs575762029 Chr.11: 78172542-78172542 on Build GRCh38
rs573243583 Chr.11: 78172501-78172501 on Build GRCh38
rs996946294 Chr.11: 78172474-78172474 on Build GRCh38
rs1024816182 Chr.11: 78172482-78172482 on Build GRCh38
rs977966071 Chr.11: 78172675-78172675 on Build GRCh38
rs971931809 Chr.11: 78172518-78172518 on Build GRCh38
rs230662 Chr.11: 78172809-78172809 on Build GRCh38
rs560823251 Chr.11: 78172646-78172646 on Build GRCh38
rs574433787 Chr.11: 78172661-78172661 on Build GRCh38
rs527462 Chr.11: 78172432-78172432 on Build GRCh38
rs917772539 Chr.11: 78172522-78172522 on Build GRCh38
rs145317421 Chr.11: 78172531-78172531 on Build GRCh38
rs901343602 Chr.11: 78172684-78172684 on Build GRCh38
rs538922904 Chr.11: 78172523-78172523 on Build GRCh38
rs936386275 Chr.11: 78172617-78172617 on Build GRCh38
rs560187042 Chr.11: 78172714-78172714 on Build GRCh38
rs989198977 Chr.11: 78172814-78172814 on Build GRCh38
rs931353932 Chr.11: 78172799-78172799 on Build GRCh38
rs779222802 Chr.11: 78172859-78172859 on Build GRCh38
rs892112046 Chr.11: 78172654-78172655 on Build GRCh38
rs536338610 Chr.11: 78172817-78172817 on Build GRCh38
rs952782011 Chr.11: 78172491-78172491 on Build GRCh38
rs562768611 Chr.11: 78172800-78172800 on Build GRCh38
rs892716255 Chr.11: 78172400-78172400 on Build GRCh38
rs552485936 Chr.11: 78172795-78172795 on Build GRCh38
rs1028441393 Chr.11: 78172734-78172734 on Build GRCh38
rs532478162 Chr.11: 78172744-78172744 on Build GRCh38
rs187574842 Chr.11: 78172487-78172487 on Build GRCh38
rs944105282 Chr.11: 78172777-78172777 on Build GRCh38
rs753941225 Chr.11: 78172792-78172792 on Build GRCh38
rs914865011 Chr.11: 78172545-78172545 on Build GRCh38
rs970629321 Chr.11: 78172794-78172794 on Build GRCh38
rs191994039 Chr.11: 78172601-78172601 on Build GRCh38
rs1003961826 Chr.11: 78172477-78172477 on Build GRCh38
rs117114945 Chr.11: 78172444-78172444 on Build GRCh38
rs1025204778 Chr.11: 78172860-78172860 on Build GRCh38
rs531428355 Chr.11: 78172801-78172801 on Build GRCh38
rs980564628 Chr.11: 78172488-78172488 on Build GRCh38
rs1009760474 Chr.11: 78172401-78172401 on Build GRCh38
rs997420348 Chr.11: 78172712-78172712 on Build GRCh38
rs1039819465 Chr.11: 78172793-78172793 on Build GRCh38
rs984077334 Chr.11: 78172496-78172496 on Build GRCh38
rs548320918 Chr.11: 78172806-78172806 on Build GRCh38

View Full Product Details