Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85631573-85632121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM126B
Entrez Gene ID
55863
Gene Name
transmembrane protein 126B
Gene Aliases
HT007
UniGene
Hs.525063
Gene Chromosome Location
Chr.11: 85628573-85636539 on Build GRCh38
Transcripts
NM_001193537.2 NM_001193538.2 NM_001256546.1 NM_001256547.1 NM_018480.4 XM_011545165.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4201358 c.199G>T p.E67* Chr.11: 85631804-85631804 on Build GRCh38
7394066 c.152C>A p.P51Q Chr.11: 85631757-85631757 on Build GRCh38
7394067 c.62C>A p.P21Q Chr.11: 85631757-85631757 on Build GRCh38
4201357 c.89A>T p.N30I Chr.11: 85631784-85631784 on Build GRCh38
3398175 c.80T>C p.I27T Chr.11: 85631775-85631775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892962335 Chr.11: 85631919-85631919 on Build GRCh38
rs762169263 Chr.11: 85631815-85631815 on Build GRCh38
rs139974357 Chr.11: 85631754-85631754 on Build GRCh38
rs2508343 Chr.11: 85631973-85631973 on Build GRCh38
rs764121091 Chr.11: 85631712-85631712 on Build GRCh38
rs745953172 Chr.11: 85631811-85631812 on Build GRCh38
rs761155840 Chr.11: 85631692-85631702 on Build GRCh38
rs547778179 Chr.11: 85631766-85631766 on Build GRCh38
rs115581134 Chr.11: 85631688-85631688 on Build GRCh38
rs751104967 Chr.11: 85631786-85631786 on Build GRCh38
rs771251099 Chr.11: 85631686-85631686 on Build GRCh38
rs781414986 Chr.11: 85631774-85631774 on Build GRCh38
rs10501595 Chr.11: 85631915-85631915 on Build GRCh38
rs751995536 Chr.11: 85631821-85631821 on Build GRCh38
rs749345722 Chr.11: 85631804-85631804 on Build GRCh38
rs749539943 Chr.11: 85631854-85631854 on Build GRCh38
rs766117185 Chr.11: 85631755-85631755 on Build GRCh38
rs754437170 Chr.11: 85631820-85631820 on Build GRCh38
rs774465244 Chr.11: 85631727-85631727 on Build GRCh38
rs772873069 Chr.11: 85631813-85631813 on Build GRCh38
rs144210716 Chr.11: 85631695-85631695 on Build GRCh38
rs953962857 Chr.11: 85631941-85631941 on Build GRCh38
rs962834652 Chr.11: 85631602-85631602 on Build GRCh38
rs757716030 Chr.11: 85631823-85631823 on Build GRCh38
rs763180872 Chr.11: 85631708-85631708 on Build GRCh38
rs778321372 Chr.11: 85631652-85631652 on Build GRCh38
rs779437126 Chr.11: 85631637-85631637 on Build GRCh38
rs558351504 Chr.11: 85631784-85631785 on Build GRCh38
rs777108257 Chr.11: 85631814-85631815 on Build GRCh38
rs757959963 Chr.11: 85631877-85631877 on Build GRCh38
rs528300437 Chr.11: 85631991-85631991 on Build GRCh38
rs775918690 Chr.11: 85631795-85631795 on Build GRCh38
rs868347401 Chr.11: 85631685-85631685 on Build GRCh38
rs943172756 Chr.11: 85631841-85631841 on Build GRCh38
rs754525612 Chr.11: 85631759-85631759 on Build GRCh38
rs1019225189 Chr.11: 85631987-85631987 on Build GRCh38
rs635338 Chr.11: 85631859-85631859 on Build GRCh38
rs371123037 Chr.11: 85631663-85631663 on Build GRCh38
rs775830779 Chr.11: 85631812-85631813 on Build GRCh38
rs764565613 Chr.11: 85631742-85631742 on Build GRCh38
rs923935826 Chr.11: 85632082-85632082 on Build GRCh38
rs370857212 Chr.11: 85631932-85631932 on Build GRCh38
rs757074062 Chr.11: 85631723-85631723 on Build GRCh38
rs568620806 Chr.11: 85631773-85631773 on Build GRCh38
rs962886765 Chr.11: 85631957-85631957 on Build GRCh38
rs1008330597 Chr.11: 85631944-85631944 on Build GRCh38
rs757845368 Chr.11: 85631778-85631778 on Build GRCh38
rs767289129 Chr.11: 85631811-85631811 on Build GRCh38
rs2508344 Chr.11: 85632049-85632049 on Build GRCh38
rs1013064327 Chr.11: 85631968-85631968 on Build GRCh38
rs767250668 Chr.11: 85631734-85631734 on Build GRCh38
rs796313882 Chr.11: 85631810-85631811 on Build GRCh38
rs754264854 Chr.11: 85631774-85631775 on Build GRCh38
rs377378732 Chr.11: 85631704-85631704 on Build GRCh38
rs754558258 Chr.11: 85631810-85631811 on Build GRCh38
rs866614389 Chr.11: 85631922-85631922 on Build GRCh38
rs772555902 Chr.11: 85631649-85631649 on Build GRCh38
rs753047056 Chr.11: 85631673-85631674 on Build GRCh38
rs990981055 Chr.11: 85632014-85632014 on Build GRCh38
rs972902224 Chr.11: 85631709-85631709 on Build GRCh38
rs990033908 Chr.11: 85632079-85632079 on Build GRCh38
rs536791394 Chr.11: 85632019-85632019 on Build GRCh38
rs901575900 Chr.11: 85631866-85631866 on Build GRCh38
rs972452255 Chr.11: 85632001-85632001 on Build GRCh38
rs991581356 Chr.11: 85631647-85631647 on Build GRCh38
rs767869463 Chr.11: 85631674-85631674 on Build GRCh38
rs981445594 Chr.11: 85631701-85631701 on Build GRCh38
rs868768625 Chr.11: 85632026-85632026 on Build GRCh38
rs554995625 Chr.11: 85632042-85632042 on Build GRCh38
rs746210909 Chr.11: 85631831-85631831 on Build GRCh38
rs919644846 Chr.11: 85632010-85632010 on Build GRCh38
rs774378991 Chr.11: 85631810-85631810 on Build GRCh38
rs1017166274 Chr.11: 85631951-85631951 on Build GRCh38
rs375814704 Chr.11: 85631840-85631840 on Build GRCh38
rs753343420 Chr.11: 85631757-85631757 on Build GRCh38
rs769893074 Chr.11: 85631790-85631790 on Build GRCh38
rs548046319 Chr.11: 85631956-85631956 on Build GRCh38
rs74405350 Chr.11: 85631672-85631672 on Build GRCh38
rs779413531 Chr.11: 85631808-85631809 on Build GRCh38
rs779151075 Chr.11: 85631860-85631860 on Build GRCh38
rs768792953 Chr.11: 85631858-85631858 on Build GRCh38
rs949649084 Chr.11: 85632033-85632033 on Build GRCh38
rs981882582 Chr.11: 85632015-85632015 on Build GRCh38
rs143403924 Chr.11: 85631730-85631730 on Build GRCh38
rs1046468592 Chr.11: 85632040-85632040 on Build GRCh38
rs956194657 Chr.11: 85632068-85632068 on Build GRCh38
rs751082995 Chr.11: 85631809-85631810 on Build GRCh38
rs35174772 Chr.11: 85631745-85631745 on Build GRCh38
rs778231539 Chr.11: 85631762-85631762 on Build GRCh38
rs750029897 Chr.11: 85631738-85631738 on Build GRCh38
rs768901452 Chr.11: 85631806-85631806 on Build GRCh38
rs953007796 Chr.11: 85632011-85632011 on Build GRCh38
rs751430602 Chr.11: 85631929-85631929 on Build GRCh38
rs916842673 Chr.11: 85632028-85632028 on Build GRCh38
rs765918115 Chr.11: 85631815-85631815 on Build GRCh38
rs769843609 Chr.11: 85631703-85631703 on Build GRCh38
rs748599740 Chr.11: 85631641-85631641 on Build GRCh38
rs912329024 Chr.11: 85631643-85631643 on Build GRCh38
rs759823337 Chr.11: 85631672-85631672 on Build GRCh38
rs77232861 Chr.11: 85631878-85631878 on Build GRCh38
rs151235178 Chr.11: 85631829-85631829 on Build GRCh38
rs530536615 Chr.11: 85631833-85631833 on Build GRCh38
rs780397560 Chr.11: 85631843-85631843 on Build GRCh38

View Full Product Details