Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85663637-85664130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBZF
Entrez Gene ID
58487
Gene Name
CREB/ATF bZIP transcription factor
Gene Aliases
SMILE, ZF
UniGene
Hs.535319
Gene Chromosome Location
Chr.11: 85657564-85665148 on Build GRCh38
Transcripts
NM_001039618.2 NR_028024.1 NR_028025.1 NR_028026.1 NR_028027.1 XM_006718642.3 XM_011545195.2 XM_017018086.1 XM_017018087.1 XM_017018088.1 XM_017018089.1 XM_017018090.1 XM_017018091.1 XM_017018092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5045950 c.966C>G p.D322E Chr.11: 85663910-85663910 on Build GRCh38
2221214 c.1063T>C p.*355Q Chr.11: 85663813-85663813 on Build GRCh38
2221215 c.1051_1053delTCT p.S351delS Chr.11: 85663823-85663825 on Build GRCh38
5360445 c.772delC p.Q258fs*14 Chr.11: 85664104-85664104 on Build GRCh38
1676348 c.989A>C p.H330P Chr.11: 85663887-85663887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752637526 Chr.11: 85664066-85664066 on Build GRCh38
rs772395373 Chr.11: 85663760-85663760 on Build GRCh38
rs918084703 Chr.11: 85663928-85663928 on Build GRCh38
rs764579967 Chr.11: 85663900-85663900 on Build GRCh38
rs374604639 Chr.11: 85663797-85663797 on Build GRCh38
rs770552159 Chr.11: 85663976-85663976 on Build GRCh38
rs978564274 Chr.11: 85664013-85664013 on Build GRCh38
rs996602799 Chr.11: 85663767-85663767 on Build GRCh38
rs933546988 Chr.11: 85663751-85663751 on Build GRCh38
rs201745821 Chr.11: 85663836-85663836 on Build GRCh38
rs992451178 Chr.11: 85663927-85663927 on Build GRCh38
rs756281035 Chr.11: 85663804-85663804 on Build GRCh38
rs947504802 Chr.11: 85663691-85663691 on Build GRCh38
rs760138722 Chr.11: 85663882-85663882 on Build GRCh38
rs3180641 Chr.11: 85663832-85663832 on Build GRCh38
rs759005133 Chr.11: 85663982-85663982 on Build GRCh38
rs750613960 Chr.11: 85664034-85664034 on Build GRCh38
rs900708725 Chr.11: 85663697-85663697 on Build GRCh38
rs746680433 Chr.11: 85663973-85663973 on Build GRCh38
rs71482595 Chr.11: 85664089-85664089 on Build GRCh38
rs749905822 Chr.11: 85663761-85663761 on Build GRCh38
rs751918146 Chr.11: 85663901-85663901 on Build GRCh38
rs528150107 Chr.11: 85663778-85663778 on Build GRCh38
rs756195340 Chr.11: 85663924-85663924 on Build GRCh38
rs780196227 Chr.11: 85663931-85663931 on Build GRCh38
rs762225182 Chr.11: 85664028-85664028 on Build GRCh38
rs528927037 Chr.11: 85663958-85663958 on Build GRCh38
rs368743578 Chr.11: 85663794-85663794 on Build GRCh38
rs748937747 Chr.11: 85663823-85663825 on Build GRCh38
rs1052041951 Chr.11: 85663757-85663757 on Build GRCh38
rs765922496 Chr.11: 85663887-85663887 on Build GRCh38
rs1039019571 Chr.11: 85664107-85664107 on Build GRCh38
rs780845882 Chr.11: 85663822-85663822 on Build GRCh38
rs375094844 Chr.11: 85663766-85663766 on Build GRCh38
rs947488879 Chr.11: 85664029-85664029 on Build GRCh38
rs758627659 Chr.11: 85663777-85663777 on Build GRCh38
rs1015372692 Chr.11: 85663856-85663856 on Build GRCh38
rs1042513206 Chr.11: 85663695-85663695 on Build GRCh38
rs767795438 Chr.11: 85663910-85663910 on Build GRCh38
rs373154709 Chr.11: 85663879-85663879 on Build GRCh38
rs758134105 Chr.11: 85664071-85664071 on Build GRCh38
rs1192 Chr.11: 85663949-85663949 on Build GRCh38
rs778872228 Chr.11: 85664054-85664054 on Build GRCh38
rs377275449 Chr.11: 85663880-85663880 on Build GRCh38
rs754170897 Chr.11: 85663916-85663916 on Build GRCh38
rs749384145 Chr.11: 85663939-85663939 on Build GRCh38
rs1054277338 Chr.11: 85663748-85663748 on Build GRCh38
rs903841142 Chr.11: 85663699-85663699 on Build GRCh38
rs775807726 Chr.11: 85663889-85663889 on Build GRCh38
rs370630820 Chr.11: 85663895-85663895 on Build GRCh38
rs770589645 Chr.11: 85664098-85664098 on Build GRCh38
rs945452170 Chr.11: 85663674-85663674 on Build GRCh38
rs898240708 Chr.11: 85663821-85663821 on Build GRCh38
rs779815478 Chr.11: 85663820-85663820 on Build GRCh38
rs763938070 Chr.11: 85663827-85663829 on Build GRCh38
rs548587594 Chr.11: 85663999-85663999 on Build GRCh38
rs746876771 Chr.11: 85664077-85664077 on Build GRCh38
rs765684202 Chr.11: 85663770-85663770 on Build GRCh38
rs747978455 Chr.11: 85663850-85663850 on Build GRCh38
rs943325137 Chr.11: 85664002-85664002 on Build GRCh38
rs1000897710 Chr.11: 85663733-85663733 on Build GRCh38
rs780753635 Chr.11: 85664108-85664108 on Build GRCh38
rs749251989 Chr.11: 85663824-85663824 on Build GRCh38
rs755036034 Chr.11: 85664045-85664045 on Build GRCh38
rs757742260 Chr.11: 85663904-85663904 on Build GRCh38
rs914598144 Chr.11: 85664000-85664000 on Build GRCh38
rs777533538 Chr.11: 85664072-85664072 on Build GRCh38
rs899774664 Chr.11: 85663752-85663752 on Build GRCh38
rs753932211 Chr.11: 85664041-85664041 on Build GRCh38
rs962670859 Chr.11: 85663871-85663871 on Build GRCh38
rs772788789 Chr.11: 85663959-85663959 on Build GRCh38
rs772923265 Chr.11: 85663807-85663808 on Build GRCh38
rs936640060 Chr.11: 85664096-85664096 on Build GRCh38
rs969885931 Chr.11: 85663851-85663851 on Build GRCh38
rs754913786 Chr.11: 85663941-85663941 on Build GRCh38
rs750702014 Chr.11: 85663923-85663923 on Build GRCh38
rs760965505 Chr.11: 85664038-85664038 on Build GRCh38
rs914668192 Chr.11: 85663689-85663689 on Build GRCh38
rs188846430 Chr.11: 85663798-85663798 on Build GRCh38
rs773743197 Chr.11: 85663827-85663827 on Build GRCh38
rs775018789 Chr.11: 85664011-85664011 on Build GRCh38
rs371179178 Chr.11: 85663873-85663873 on Build GRCh38
rs753282844 Chr.11: 85663776-85663776 on Build GRCh38
rs776204067 Chr.11: 85663977-85663977 on Build GRCh38
rs766575388 Chr.11: 85664040-85664040 on Build GRCh38
rs747709751 Chr.11: 85663671-85663671 on Build GRCh38
rs774181076 Chr.11: 85663841-85663841 on Build GRCh38

View Full Product Details