Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86256269-86256776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EED
Entrez Gene ID
8726
Gene Name
embryonic ectoderm development
Gene Aliases
HEED, WAIT1
UniGene
Hs.503510
Gene Chromosome Location
Chr.11: 86244384-86285420 on Build GRCh38
Transcripts
NM_001308007.1 NM_003797.4 XM_005274373.2 XM_011545330.1 XM_011545331.1 XM_017018512.1 XM_017018513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6983563 c.500G>T p.G167V Chr.11: 86256460-86256460 on Build GRCh38
6370800 c.473C>T p.T158M Chr.11: 86256433-86256433 on Build GRCh38
6960278 c.445A>T p.T149S Chr.11: 86256405-86256405 on Build GRCh38
3870351 c.482C>T p.P161L Chr.11: 86256442-86256442 on Build GRCh38
6856913 c.552+6_552+7insT p.? Chr.11: 86256518-86256519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753803438 Chr.11: 86256556-86256556 on Build GRCh38
rs193258630 Chr.11: 86256446-86256446 on Build GRCh38
rs977393685 Chr.11: 86256563-86256563 on Build GRCh38
rs375789080 Chr.11: 86256378-86256378 on Build GRCh38
rs761264705 Chr.11: 86256552-86256552 on Build GRCh38
rs755960470 Chr.11: 86256413-86256413 on Build GRCh38
rs763345233 Chr.11: 86256353-86256353 on Build GRCh38
rs777795883 Chr.11: 86256401-86256401 on Build GRCh38
rs141323002 Chr.11: 86256433-86256433 on Build GRCh38
rs1046329368 Chr.11: 86256749-86256749 on Build GRCh38
rs552803691 Chr.11: 86256610-86256610 on Build GRCh38
rs776070197 Chr.11: 86256537-86256537 on Build GRCh38
rs377184333 Chr.11: 86256541-86256541 on Build GRCh38
rs751766336 Chr.11: 86256380-86256380 on Build GRCh38
rs749625617 Chr.11: 86256555-86256555 on Build GRCh38
rs930119710 Chr.11: 86256583-86256583 on Build GRCh38
rs769135069 Chr.11: 86256621-86256621 on Build GRCh38
rs747800796 Chr.11: 86256461-86256461 on Build GRCh38
rs201693325 Chr.11: 86256545-86256545 on Build GRCh38
rs578180965 Chr.11: 86256743-86256743 on Build GRCh38
rs778911575 Chr.11: 86256437-86256437 on Build GRCh38
rs957753412 Chr.11: 86256332-86256332 on Build GRCh38
rs575626060 Chr.11: 86256666-86256666 on Build GRCh38
rs768551883 Chr.11: 86256434-86256434 on Build GRCh38
rs551321312 Chr.11: 86256577-86256577 on Build GRCh38
rs770386903 Chr.11: 86256519-86256519 on Build GRCh38
rs941697847 Chr.11: 86256635-86256635 on Build GRCh38
rs74971473 Chr.11: 86256628-86256628 on Build GRCh38
rs940022815 Chr.11: 86256616-86256616 on Build GRCh38
rs1055212893 Chr.11: 86256706-86256706 on Build GRCh38
rs757539279 Chr.11: 86256405-86256405 on Build GRCh38
rs555463990 Chr.11: 86256657-86256657 on Build GRCh38
rs763427179 Chr.11: 86256533-86256533 on Build GRCh38
rs781178324 Chr.11: 86256407-86256407 on Build GRCh38
rs750525883 Chr.11: 86256412-86256412 on Build GRCh38
rs555779960 Chr.11: 86256614-86256614 on Build GRCh38
rs767745744 Chr.11: 86256538-86256538 on Build GRCh38
rs535808704 Chr.11: 86256633-86256633 on Build GRCh38
rs759973779 Chr.11: 86256338-86256339 on Build GRCh38
rs776040537 Chr.11: 86256522-86256522 on Build GRCh38
rs1013740338 Chr.11: 86256752-86256752 on Build GRCh38
rs771855344 Chr.11: 86256482-86256482 on Build GRCh38
rs918777317 Chr.11: 86256570-86256570 on Build GRCh38
rs752963650 Chr.11: 86256343-86256343 on Build GRCh38
rs369959606 Chr.11: 86256559-86256559 on Build GRCh38
rs907767827 Chr.11: 86256607-86256607 on Build GRCh38
rs983179562 Chr.11: 86256603-86256603 on Build GRCh38
rs556684546 Chr.11: 86256491-86256491 on Build GRCh38
rs764435402 Chr.11: 86256534-86256534 on Build GRCh38
rs147440081 Chr.11: 86256492-86256492 on Build GRCh38
rs778786820 Chr.11: 86256561-86256561 on Build GRCh38
rs372688306 Chr.11: 86256539-86256539 on Build GRCh38
rs999635570 Chr.11: 86256352-86256352 on Build GRCh38
rs772359364 Chr.11: 86256357-86256359 on Build GRCh38
rs776135997 Chr.11: 86256316-86256316 on Build GRCh38
rs10792842 Chr.11: 86256667-86256667 on Build GRCh38
rs765329415 Chr.11: 86256359-86256360 on Build GRCh38
rs920015875 Chr.11: 86256643-86256643 on Build GRCh38
rs564578352 Chr.11: 86256719-86256719 on Build GRCh38

View Full Product Details