Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86810800-86811337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS23
Entrez Gene ID
11098
Gene Name
protease, serine 23
Gene Aliases
SIG13, SPUVE, ZSIG13
UniGene
Hs.25338
Gene Chromosome Location
Chr.11: 86791059-86952910 on Build GRCh38
Transcripts
NM_001293178.1 NM_001293179.1 NM_001293180.1 NM_007173.5 NR_120591.1 NR_120592.1 NR_120593.1

SNP Detail

SNP ID Chromosome Location
rs3177411 Chr.11: 86811228-86811228 on Build GRCh38
rs1052113903 Chr.11: 86810846-86810846 on Build GRCh38
rs924218980 Chr.11: 86811182-86811182 on Build GRCh38
rs916615308 Chr.11: 86811129-86811129 on Build GRCh38
rs891277058 Chr.11: 86811267-86811267 on Build GRCh38
rs542775725 Chr.11: 86811125-86811125 on Build GRCh38
rs946395315 Chr.11: 86811185-86811185 on Build GRCh38
rs1043469609 Chr.11: 86810877-86810877 on Build GRCh38
rs564885802 Chr.11: 86811166-86811166 on Build GRCh38
rs1008939851 Chr.11: 86810930-86810930 on Build GRCh38
rs946482717 Chr.11: 86810866-86810866 on Build GRCh38
rs995923189 Chr.11: 86810907-86810907 on Build GRCh38
rs927751530 Chr.11: 86811078-86811078 on Build GRCh38
rs547326225 Chr.11: 86811015-86811015 on Build GRCh38
rs982492889 Chr.11: 86811150-86811150 on Build GRCh38
rs921030842 Chr.11: 86811218-86811218 on Build GRCh38
rs1023044396 Chr.11: 86811047-86811047 on Build GRCh38
rs61159986 Chr.11: 86810921-86810921 on Build GRCh38
rs545199235 Chr.11: 86811155-86811155 on Build GRCh38
rs79270994 Chr.11: 86810854-86810854 on Build GRCh38
rs900039060 Chr.11: 86810879-86810879 on Build GRCh38
rs996032097 Chr.11: 86811042-86811042 on Build GRCh38
rs545767665 Chr.11: 86811089-86811089 on Build GRCh38
rs372442803 Chr.11: 86811116-86811116 on Build GRCh38
rs531448300 Chr.11: 86811145-86811145 on Build GRCh38
rs79072426 Chr.11: 86811248-86811248 on Build GRCh38
rs573641965 Chr.11: 86811069-86811069 on Build GRCh38
rs562570857 Chr.11: 86811143-86811143 on Build GRCh38
rs1004400371 Chr.11: 86810939-86810939 on Build GRCh38
rs952834489 Chr.11: 86811051-86811051 on Build GRCh38
rs994656644 Chr.11: 86811035-86811035 on Build GRCh38
rs865848390 Chr.11: 86811283-86811283 on Build GRCh38
rs749593577 Chr.11: 86811159-86811159 on Build GRCh38
rs200816967 Chr.11: 86810883-86810883 on Build GRCh38
rs376513904 Chr.11: 86810845-86810845 on Build GRCh38
rs1043747677 Chr.11: 86811207-86811207 on Build GRCh38
rs1053998661 Chr.11: 86811236-86811236 on Build GRCh38
rs756535991 Chr.11: 86810990-86810990 on Build GRCh38
rs780399212 Chr.11: 86811017-86811017 on Build GRCh38
rs935551980 Chr.11: 86811189-86811189 on Build GRCh38
rs997027886 Chr.11: 86810897-86810897 on Build GRCh38
rs960630931 Chr.11: 86811101-86811101 on Build GRCh38
rs890136027 Chr.11: 86810927-86810927 on Build GRCh38
rs1027425591 Chr.11: 86811039-86811039 on Build GRCh38
rs1014859043 Chr.11: 86811014-86811014 on Build GRCh38
rs1015573531 Chr.11: 86810986-86810986 on Build GRCh38
rs377016931 Chr.11: 86811135-86811135 on Build GRCh38

View Full Product Details