Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 101031126-101031626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGR
Entrez Gene ID
5241
Gene Name
progesterone receptor
Gene Aliases
NR3C3, PR
UniGene
Hs.32405
Gene Chromosome Location
Chr.11: 101029624-101130681 on Build GRCh38
Transcripts
NM_000926.4 NM_001202474.3 NM_001271161.2 NM_001271162.1 NR_073141.2 NR_073142.2 NR_073143.2

SNP Detail

SNP ID Chromosome Location
rs972682539 Chr.11: 101031324-101031324 on Build GRCh38
rs758842029 Chr.11: 101031491-101031491 on Build GRCh38
rs534367678 Chr.11: 101031381-101031381 on Build GRCh38
rs56018527 Chr.11: 101031526-101031526 on Build GRCh38
rs986602986 Chr.11: 101031176-101031176 on Build GRCh38
rs997452133 Chr.11: 101031182-101031182 on Build GRCh38
rs953379573 Chr.11: 101031297-101031297 on Build GRCh38
rs574319304 Chr.11: 101031562-101031562 on Build GRCh38
rs937029367 Chr.11: 101031361-101031361 on Build GRCh38
rs140175696 Chr.11: 101031570-101031570 on Build GRCh38
rs1016356144 Chr.11: 101031518-101031518 on Build GRCh38
rs1006260383 Chr.11: 101031508-101031508 on Build GRCh38
rs913062519 Chr.11: 101031158-101031158 on Build GRCh38
rs765261089 Chr.11: 101031335-101031335 on Build GRCh38
rs926938263 Chr.11: 101031360-101031360 on Build GRCh38
rs565617287 Chr.11: 101031364-101031364 on Build GRCh38
rs557057142 Chr.11: 101031550-101031550 on Build GRCh38
rs930076301 Chr.11: 101031268-101031268 on Build GRCh38
rs905426200 Chr.11: 101031326-101031326 on Build GRCh38
rs531041785 Chr.11: 101031267-101031267 on Build GRCh38
rs1025534306 Chr.11: 101031533-101031533 on Build GRCh38
rs79996667 Chr.11: 101031591-101031591 on Build GRCh38
rs150084437 Chr.11: 101031267-101031271 on Build GRCh38
rs551746158 Chr.11: 101031227-101031227 on Build GRCh38
rs188799198 Chr.11: 101031472-101031472 on Build GRCh38
rs537170890 Chr.11: 101031540-101031540 on Build GRCh38
rs945896824 Chr.11: 101031428-101031428 on Build GRCh38
rs917984199 Chr.11: 101031566-101031566 on Build GRCh38
rs1824125 Chr.11: 101031322-101031322 on Build GRCh38
rs1020191308 Chr.11: 101031159-101031159 on Build GRCh38
rs750874828 Chr.11: 101031456-101031456 on Build GRCh38
rs926904171 Chr.11: 101031558-101031558 on Build GRCh38
rs75260139 Chr.11: 101031592-101031592 on Build GRCh38
rs955135884 Chr.11: 101031473-101031473 on Build GRCh38
rs981060036 Chr.11: 101031354-101031354 on Build GRCh38
rs981412014 Chr.11: 101031554-101031554 on Build GRCh38
rs997822251 Chr.11: 101031490-101031490 on Build GRCh38
rs182901246 Chr.11: 101031401-101031401 on Build GRCh38
rs779145953 Chr.11: 101031413-101031413 on Build GRCh38
rs1010511203 Chr.11: 101031151-101031151 on Build GRCh38
rs529023034 Chr.11: 101031350-101031350 on Build GRCh38
rs753924407 Chr.11: 101031298-101031298 on Build GRCh38
rs962083385 Chr.11: 101031520-101031520 on Build GRCh38
rs914239501 Chr.11: 101031594-101031594 on Build GRCh38
rs918095407 Chr.11: 101031330-101031330 on Build GRCh38
rs760929776 Chr.11: 101031591-101031591 on Build GRCh38
rs901795423 Chr.11: 101031163-101031163 on Build GRCh38
rs147888503 Chr.11: 101031304-101031304 on Build GRCh38
rs548903321 Chr.11: 101031356-101031356 on Build GRCh38
rs989813080 Chr.11: 101031575-101031575 on Build GRCh38
rs1047552191 Chr.11: 101031272-101031272 on Build GRCh38
rs563253222 Chr.11: 101031544-101031544 on Build GRCh38
rs911075402 Chr.11: 101031497-101031497 on Build GRCh38
rs796565641 Chr.11: 101031497-101031497 on Build GRCh38
rs747345971 Chr.11: 101031523-101031523 on Build GRCh38
rs972182218 Chr.11: 101031528-101031528 on Build GRCh38
rs889108357 Chr.11: 101031499-101031499 on Build GRCh38
rs945759160 Chr.11: 101031595-101031595 on Build GRCh38
rs565274011 Chr.11: 101031236-101031236 on Build GRCh38
rs1009905896 Chr.11: 101031454-101031454 on Build GRCh38

View Full Product Details