Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102075741-102076281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf70
Entrez Gene ID
85016
Gene Name
chromosome 11 open reading frame 70
Gene Aliases
-
UniGene
Hs.98328
Gene Chromosome Location
Chr.11: 102047409-102084560 on Build GRCh38
Transcripts
NM_001195005.1 NM_032930.2 XM_005271713.3 XM_017018454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614101 c.517C>T p.H173Y Chr.11: 102075954-102075954 on Build GRCh38
685809 c.337G>C p.D113H Chr.11: 102075888-102075888 on Build GRCh38
6348179 c.493G>C p.E165Q Chr.11: 102075930-102075930 on Build GRCh38
4998426 c.586C>A p.L196I Chr.11: 102076023-102076023 on Build GRCh38
6067027 c.493G>T p.E165* Chr.11: 102075930-102075930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543398165 Chr.11: 102076233-102076233 on Build GRCh38
rs772384023 Chr.11: 102075833-102075833 on Build GRCh38
rs776784115 Chr.11: 102076254-102076254 on Build GRCh38
rs555467417 Chr.11: 102076227-102076227 on Build GRCh38
rs1008567031 Chr.11: 102075779-102075779 on Build GRCh38
rs745659546 Chr.11: 102075919-102075919 on Build GRCh38
rs533647204 Chr.11: 102075811-102075811 on Build GRCh38
rs752496808 Chr.11: 102075905-102075905 on Build GRCh38
rs757364853 Chr.11: 102075937-102075937 on Build GRCh38
rs376842255 Chr.11: 102075965-102075965 on Build GRCh38
rs1040011315 Chr.11: 102076126-102076126 on Build GRCh38
rs915184576 Chr.11: 102076020-102076020 on Build GRCh38
rs868371088 Chr.11: 102076200-102076200 on Build GRCh38
rs759047947 Chr.11: 102075979-102075979 on Build GRCh38
rs556290819 Chr.11: 102076146-102076146 on Build GRCh38
rs944462401 Chr.11: 102076125-102076125 on Build GRCh38
rs116126510 Chr.11: 102075866-102075866 on Build GRCh38
rs747400996 Chr.11: 102075957-102075957 on Build GRCh38
rs750519685 Chr.11: 102076028-102076028 on Build GRCh38
rs778858126 Chr.11: 102075859-102075859 on Build GRCh38
rs770407389 Chr.11: 102075967-102075967 on Build GRCh38
rs777318658 Chr.11: 102075961-102075961 on Build GRCh38
rs373573579 Chr.11: 102075950-102075950 on Build GRCh38
rs371474452 Chr.11: 102076080-102076080 on Build GRCh38
rs1019031328 Chr.11: 102075784-102075784 on Build GRCh38
rs763985305 Chr.11: 102076017-102076017 on Build GRCh38
rs746133407 Chr.11: 102075939-102075939 on Build GRCh38
rs1014240853 Chr.11: 102076129-102076129 on Build GRCh38
rs761205079 Chr.11: 102075836-102075836 on Build GRCh38
rs192420752 Chr.11: 102076107-102076107 on Build GRCh38
rs770342377 Chr.11: 102076057-102076057 on Build GRCh38
rs937913571 Chr.11: 102075999-102075999 on Build GRCh38
rs752435466 Chr.11: 102075993-102075993 on Build GRCh38
rs142524446 Chr.11: 102075925-102075925 on Build GRCh38
rs776052469 Chr.11: 102075835-102075835 on Build GRCh38
rs779190170 Chr.11: 102075938-102075938 on Build GRCh38
rs879110377 Chr.11: 102075830-102075830 on Build GRCh38
rs374908360 Chr.11: 102076193-102076193 on Build GRCh38
rs773698817 Chr.11: 102076069-102076069 on Build GRCh38
rs372015461 Chr.11: 102076081-102076081 on Build GRCh38
rs775160676 Chr.11: 102076089-102076089 on Build GRCh38
rs770229788 Chr.11: 102075958-102075958 on Build GRCh38
rs753890578 Chr.11: 102075931-102075931 on Build GRCh38
rs1005003069 Chr.11: 102076209-102076209 on Build GRCh38
rs767227461 Chr.11: 102075983-102075983 on Build GRCh38
rs866788183 Chr.11: 102075990-102075990 on Build GRCh38
rs772351721 Chr.11: 102075943-102075943 on Build GRCh38
rs267602669 Chr.11: 102075903-102075903 on Build GRCh38
rs145316880 Chr.11: 102075991-102075991 on Build GRCh38
rs758501351 Chr.11: 102076030-102076030 on Build GRCh38
rs767163004 Chr.11: 102075898-102075898 on Build GRCh38
rs571495259 Chr.11: 102076095-102076095 on Build GRCh38
rs755462053 Chr.11: 102076041-102076041 on Build GRCh38
rs563349596 Chr.11: 102075992-102075992 on Build GRCh38
rs952198100 Chr.11: 102075893-102075893 on Build GRCh38
rs777707051 Chr.11: 102075924-102075924 on Build GRCh38
rs529008192 Chr.11: 102076004-102076004 on Build GRCh38
rs1023864860 Chr.11: 102076173-102076173 on Build GRCh38
rs756002464 Chr.11: 102075922-102075922 on Build GRCh38
rs773930289 Chr.11: 102075975-102075975 on Build GRCh38
rs778982974 Chr.11: 102076201-102076201 on Build GRCh38
rs112363908 Chr.11: 102076216-102076216 on Build GRCh38
rs771830038 Chr.11: 102075955-102075955 on Build GRCh38
rs541304615 Chr.11: 102075842-102075842 on Build GRCh38
rs748610887 Chr.11: 102076056-102076056 on Build GRCh38
rs759364993 Chr.11: 102075892-102075892 on Build GRCh38
rs995881611 Chr.11: 102076188-102076188 on Build GRCh38
rs935335739 Chr.11: 102075772-102075772 on Build GRCh38
rs141029969 Chr.11: 102076176-102076176 on Build GRCh38
rs376333708 Chr.11: 102076022-102076022 on Build GRCh38
rs923646456 Chr.11: 102076083-102076087 on Build GRCh38
rs151324884 Chr.11: 102076034-102076034 on Build GRCh38
rs1048840779 Chr.11: 102075942-102075942 on Build GRCh38
rs766025387 Chr.11: 102075855-102075855 on Build GRCh38
rs927291742 Chr.11: 102075962-102075962 on Build GRCh38
rs549559265 Chr.11: 102075959-102075959 on Build GRCh38
rs564661356 Chr.11: 102075843-102075843 on Build GRCh38
rs781762004 Chr.11: 102076052-102076052 on Build GRCh38

View Full Product Details