Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 103051005-103051503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D5
Entrez Gene ID
84259
Gene Name
defective in cullin neddylation 1 domain containing 5
Gene Aliases
SCCRO5
UniGene
Hs.503716
Gene Chromosome Location
Chr.11: 103050684-103092215 on Build GRCh38
Transcripts
NM_001318739.1 NM_001318740.1 NM_001318741.1 NM_032299.3

SNP Detail

SNP ID Chromosome Location
rs569498010 Chr.11: 103051213-103051213 on Build GRCh38
rs533423080 Chr.11: 103051072-103051072 on Build GRCh38
rs983273631 Chr.11: 103051351-103051351 on Build GRCh38
rs552840866 Chr.11: 103051382-103051382 on Build GRCh38
rs544113841 Chr.11: 103051461-103051461 on Build GRCh38
rs867069292 Chr.11: 103051156-103051156 on Build GRCh38
rs180929216 Chr.11: 103051454-103051454 on Build GRCh38
rs1018820780 Chr.11: 103051397-103051398 on Build GRCh38
rs911682225 Chr.11: 103051079-103051079 on Build GRCh38
rs373729186 Chr.11: 103051402-103051403 on Build GRCh38
rs1021777684 Chr.11: 103051128-103051128 on Build GRCh38
rs903343949 Chr.11: 103051150-103051150 on Build GRCh38
rs72987590 Chr.11: 103051363-103051363 on Build GRCh38
rs975551558 Chr.11: 103051411-103051411 on Build GRCh38
rs544979118 Chr.11: 103051403-103051403 on Build GRCh38
rs931287174 Chr.11: 103051198-103051198 on Build GRCh38
rs142797026 Chr.11: 103051189-103051189 on Build GRCh38
rs185862342 Chr.11: 103051209-103051209 on Build GRCh38
rs151104778 Chr.11: 103051384-103051384 on Build GRCh38
rs894708087 Chr.11: 103051116-103051116 on Build GRCh38
rs564422797 Chr.11: 103051055-103051055 on Build GRCh38
rs1001998833 Chr.11: 103051319-103051319 on Build GRCh38
rs571758803 Chr.11: 103051335-103051335 on Build GRCh38
rs190919709 Chr.11: 103051362-103051362 on Build GRCh38
rs1055978379 Chr.11: 103051114-103051114 on Build GRCh38
rs566415932 Chr.11: 103051319-103051319 on Build GRCh38
rs929151640 Chr.11: 103051347-103051347 on Build GRCh38
rs966576320 Chr.11: 103051046-103051046 on Build GRCh38
rs931360939 Chr.11: 103051429-103051429 on Build GRCh38
rs558224191 Chr.11: 103051388-103051388 on Build GRCh38
rs561814802 Chr.11: 103051441-103051441 on Build GRCh38
rs528673974 Chr.11: 103051331-103051331 on Build GRCh38
rs566900516 Chr.11: 103051365-103051365 on Build GRCh38
rs913803503 Chr.11: 103051044-103051044 on Build GRCh38
rs906655255 Chr.11: 103051318-103051318 on Build GRCh38
rs551903735 Chr.11: 103051334-103051334 on Build GRCh38
rs776696699 Chr.11: 103051135-103051135 on Build GRCh38
rs1011766627 Chr.11: 103051125-103051125 on Build GRCh38
rs535881125 Chr.11: 103051366-103051366 on Build GRCh38
rs117470978 Chr.11: 103051261-103051261 on Build GRCh38
rs772546387 Chr.11: 103051280-103051280 on Build GRCh38
rs762571093 Chr.11: 103051309-103051309 on Build GRCh38
rs117918253 Chr.11: 103051420-103051420 on Build GRCh38
rs529331372 Chr.11: 103051255-103051255 on Build GRCh38

View Full Product Details