Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112170369-112170906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX12
Entrez Gene ID
56158
Gene Name
testis expressed 12
Gene Aliases
-
UniGene
Hs.524039
Gene Chromosome Location
Chr.11: 112167372-112172556 on Build GRCh38
Transcripts
NM_031275.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
923095 c.162G>T p.E54D Chr.11: 112170517-112170517 on Build GRCh38
2102791 c.64-7C>T p.? Chr.11: 112170412-112170412 on Build GRCh38
361626 c.76G>C p.D26H Chr.11: 112170431-112170431 on Build GRCh38
1351285 c.97C>T p.L33F Chr.11: 112170452-112170452 on Build GRCh38
6890299 c.101G>A p.G34E Chr.11: 112170456-112170456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202187323 Chr.11: 112170624-112170624 on Build GRCh38
rs914181691 Chr.11: 112170847-112170847 on Build GRCh38
rs542867232 Chr.11: 112170571-112170571 on Build GRCh38
rs761387860 Chr.11: 112170657-112170657 on Build GRCh38
rs11214107 Chr.11: 112170874-112170874 on Build GRCh38
rs771310058 Chr.11: 112170641-112170641 on Build GRCh38
rs763222204 Chr.11: 112170440-112170440 on Build GRCh38
rs1010616661 Chr.11: 112170520-112170520 on Build GRCh38
rs920607377 Chr.11: 112170727-112170727 on Build GRCh38
rs1043455141 Chr.11: 112170763-112170763 on Build GRCh38
rs763806920 Chr.11: 112170654-112170654 on Build GRCh38
rs149903853 Chr.11: 112170406-112170414 on Build GRCh38
rs752290155 Chr.11: 112170444-112170444 on Build GRCh38
rs754816856 Chr.11: 112170696-112170696 on Build GRCh38
rs74682721 Chr.11: 112170408-112170408 on Build GRCh38
rs188352044 Chr.11: 112170686-112170686 on Build GRCh38
rs532812476 Chr.11: 112170786-112170786 on Build GRCh38
rs946971865 Chr.11: 112170849-112170849 on Build GRCh38
rs981269072 Chr.11: 112170758-112170758 on Build GRCh38
rs777306411 Chr.11: 112170400-112170400 on Build GRCh38
rs372394612 Chr.11: 112170539-112170539 on Build GRCh38
rs779943549 Chr.11: 112170584-112170584 on Build GRCh38
rs750408143 Chr.11: 112170668-112170668 on Build GRCh38
rs774749655 Chr.11: 112170645-112170645 on Build GRCh38
rs763155830 Chr.11: 112170601-112170602 on Build GRCh38
rs1007400549 Chr.11: 112170808-112170808 on Build GRCh38
rs558166091 Chr.11: 112170443-112170443 on Build GRCh38
rs750839844 Chr.11: 112170431-112170431 on Build GRCh38
rs777673011 Chr.11: 112170719-112170719 on Build GRCh38
rs375324229 Chr.11: 112170507-112170507 on Build GRCh38
rs1052695795 Chr.11: 112170506-112170506 on Build GRCh38
rs181224832 Chr.11: 112170731-112170731 on Build GRCh38
rs12294058 Chr.11: 112170487-112170487 on Build GRCh38
rs778688126 Chr.11: 112170475-112170475 on Build GRCh38
rs776204431 Chr.11: 112170655-112170655 on Build GRCh38
rs200180264 Chr.11: 112170570-112170570 on Build GRCh38
rs763556873 Chr.11: 112170462-112170462 on Build GRCh38
rs184402971 Chr.11: 112170536-112170536 on Build GRCh38
rs373811165 Chr.11: 112170690-112170690 on Build GRCh38
rs374971784 Chr.11: 112170559-112170559 on Build GRCh38
rs951633675 Chr.11: 112170863-112170863 on Build GRCh38
rs756571 Chr.11: 112170396-112170396 on Build GRCh38
rs755018858 Chr.11: 112170405-112170406 on Build GRCh38
rs111833972 Chr.11: 112170816-112170816 on Build GRCh38
rs746777351 Chr.11: 112170587-112170587 on Build GRCh38
rs755627915 Chr.11: 112170449-112170449 on Build GRCh38
rs781552926 Chr.11: 112170522-112170522 on Build GRCh38
rs765230608 Chr.11: 112170568-112170568 on Build GRCh38
rs200513498 Chr.11: 112170413-112170413 on Build GRCh38
rs763717628 Chr.11: 112170549-112170549 on Build GRCh38
rs928084045 Chr.11: 112170793-112170793 on Build GRCh38
rs757272178 Chr.11: 112170472-112170472 on Build GRCh38
rs186507787 Chr.11: 112170854-112170854 on Build GRCh38
rs201079014 Chr.11: 112170411-112170411 on Build GRCh38
rs904489733 Chr.11: 112170785-112170785 on Build GRCh38
rs756238701 Chr.11: 112170718-112170718 on Build GRCh38
rs768878525 Chr.11: 112170493-112170493 on Build GRCh38
rs749354799 Chr.11: 112170721-112170721 on Build GRCh38
rs781560550 Chr.11: 112170600-112170600 on Build GRCh38
rs376738969 Chr.11: 112170408-112170416 on Build GRCh38
rs1025765431 Chr.11: 112170829-112170829 on Build GRCh38
rs747192340 Chr.11: 112170491-112170491 on Build GRCh38
rs1045208243 Chr.11: 112170479-112170479 on Build GRCh38
rs759866728 Chr.11: 112170647-112170647 on Build GRCh38
rs113640749 Chr.11: 112170498-112170498 on Build GRCh38
rs773317774 Chr.11: 112170577-112170579 on Build GRCh38
rs372414148 Chr.11: 112170477-112170477 on Build GRCh38
rs762839951 Chr.11: 112170684-112170684 on Build GRCh38
rs1052256451 Chr.11: 112170723-112170723 on Build GRCh38
rs771197438 Chr.11: 112170639-112170643 on Build GRCh38
rs202099347 Chr.11: 112170412-112170412 on Build GRCh38
rs939534445 Chr.11: 112170815-112170815 on Build GRCh38
rs961999041 Chr.11: 112170666-112170666 on Build GRCh38
rs763390094 Chr.11: 112170535-112170535 on Build GRCh38
rs780426080 Chr.11: 112170488-112170488 on Build GRCh38
rs756721256 Chr.11: 112170560-112170560 on Build GRCh38
rs748317314 Chr.11: 112170607-112170607 on Build GRCh38
rs765638641 Chr.11: 112170428-112170428 on Build GRCh38
rs998342865 Chr.11: 112170818-112170818 on Build GRCh38
rs773775176 Chr.11: 112170623-112170623 on Build GRCh38
rs769706405 Chr.11: 112170619-112170619 on Build GRCh38
rs755290258 Chr.11: 112170598-112170598 on Build GRCh38
rs78087179 Chr.11: 112170853-112170853 on Build GRCh38
rs570103428 Chr.11: 112170746-112170746 on Build GRCh38
rs781568954 Chr.11: 112170709-112170709 on Build GRCh38
rs753798548 Chr.11: 112170469-112170469 on Build GRCh38

View Full Product Details