Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113981089-113981589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR3A
Entrez Gene ID
3359
Gene Name
5-hydroxytryptamine receptor 3A
Gene Aliases
5-HT-3, 5-HT3A, 5-HT3R, 5HT3R, HTR3
UniGene
Hs.413899
Gene Chromosome Location
Chr.11: 113975075-113990313 on Build GRCh38
Transcripts
NM_000869.5 NM_001161772.2 NM_213621.3 NR_046363.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2103634 c.366G>T p.W122C Chr.11: 113981286-113981286 on Build GRCh38
4835813 c.381C>T p.L127L Chr.11: 113981301-113981301 on Build GRCh38
7371638 c.353C>A p.T118K Chr.11: 113981273-113981273 on Build GRCh38
5341403 c.353C>T p.T118M Chr.11: 113981273-113981273 on Build GRCh38
3443603 c.369C>T p.V123V Chr.11: 113981289-113981289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532408701 Chr.11: 113981129-113981129 on Build GRCh38
rs1032828073 Chr.11: 113981476-113981476 on Build GRCh38
rs868498726 Chr.11: 113981558-113981558 on Build GRCh38
rs891070500 Chr.11: 113981247-113981247 on Build GRCh38
rs556194700 Chr.11: 113981170-113981170 on Build GRCh38
rs147851856 Chr.11: 113981368-113981368 on Build GRCh38
rs369511862 Chr.11: 113981322-113981322 on Build GRCh38
rs114988336 Chr.11: 113981369-113981369 on Build GRCh38
rs193065897 Chr.11: 113981330-113981330 on Build GRCh38
rs567222424 Chr.11: 113981166-113981166 on Build GRCh38
rs879828515 Chr.11: 113981424-113981424 on Build GRCh38
rs945908825 Chr.11: 113981111-113981111 on Build GRCh38
rs894672679 Chr.11: 113981148-113981148 on Build GRCh38
rs761265582 Chr.11: 113981475-113981475 on Build GRCh38
rs187977954 Chr.11: 113981134-113981134 on Build GRCh38
rs371565901 Chr.11: 113981329-113981329 on Build GRCh38
rs765960970 Chr.11: 113981267-113981267 on Build GRCh38
rs777137174 Chr.11: 113981312-113981312 on Build GRCh38
rs375271884 Chr.11: 113981196-113981196 on Build GRCh38
rs759559858 Chr.11: 113981186-113981186 on Build GRCh38
rs372830188 Chr.11: 113981295-113981295 on Build GRCh38
rs774578610 Chr.11: 113981171-113981171 on Build GRCh38
rs898114198 Chr.11: 113981210-113981210 on Build GRCh38
rs981035182 Chr.11: 113981327-113981327 on Build GRCh38
rs139883280 Chr.11: 113981274-113981274 on Build GRCh38
rs536095755 Chr.11: 113981169-113981169 on Build GRCh38
rs575904069 Chr.11: 113981469-113981470 on Build GRCh38
rs2741938 Chr.11: 113981479-113981479 on Build GRCh38
rs776183251 Chr.11: 113981208-113981208 on Build GRCh38
rs775695801 Chr.11: 113981301-113981301 on Build GRCh38
rs751173809 Chr.11: 113981273-113981273 on Build GRCh38
rs754561172 Chr.11: 113981488-113981488 on Build GRCh38
rs1038130014 Chr.11: 113981207-113981207 on Build GRCh38
rs748143851 Chr.11: 113981277-113981277 on Build GRCh38
rs369716906 Chr.11: 113981344-113981344 on Build GRCh38
rs955028170 Chr.11: 113981317-113981317 on Build GRCh38
rs372569767 Chr.11: 113981476-113981477 on Build GRCh38
rs746074248 Chr.11: 113981298-113981298 on Build GRCh38
rs752390838 Chr.11: 113981167-113981167 on Build GRCh38
rs749051731 Chr.11: 113981294-113981294 on Build GRCh38
rs767707668 Chr.11: 113981371-113981371 on Build GRCh38
rs533725521 Chr.11: 113981135-113981135 on Build GRCh38
rs1037890880 Chr.11: 113981484-113981484 on Build GRCh38
rs79452715 Chr.11: 113981321-113981321 on Build GRCh38
rs990002549 Chr.11: 113981341-113981341 on Build GRCh38
rs937059531 Chr.11: 113981191-113981191 on Build GRCh38
rs1176719 Chr.11: 113981465-113981465 on Build GRCh38
rs746965937 Chr.11: 113981214-113981214 on Build GRCh38
rs546002178 Chr.11: 113981284-113981284 on Build GRCh38
rs764924298 Chr.11: 113981535-113981535 on Build GRCh38
rs767172252 Chr.11: 113981167-113981169 on Build GRCh38
rs115066568 Chr.11: 113981480-113981480 on Build GRCh38
rs955075305 Chr.11: 113981540-113981540 on Build GRCh38
rs769287523 Chr.11: 113981306-113981306 on Build GRCh38
rs11607240 Chr.11: 113981131-113981131 on Build GRCh38
rs538612689 Chr.11: 113981559-113981559 on Build GRCh38
rs775297863 Chr.11: 113981446-113981446 on Build GRCh38
rs764607948 Chr.11: 113981216-113981216 on Build GRCh38
rs556396599 Chr.11: 113981538-113981538 on Build GRCh38
rs563840621 Chr.11: 113981431-113981431 on Build GRCh38
rs777623428 Chr.11: 113981151-113981151 on Build GRCh38
rs140878412 Chr.11: 113981229-113981229 on Build GRCh38
rs770586926 Chr.11: 113981297-113981297 on Build GRCh38
rs973623387 Chr.11: 113981549-113981549 on Build GRCh38
rs376826915 Chr.11: 113981456-113981456 on Build GRCh38
rs368237294 Chr.11: 113981212-113981212 on Build GRCh38
rs777621564 Chr.11: 113981291-113981291 on Build GRCh38
rs757571267 Chr.11: 113981238-113981238 on Build GRCh38
rs184833515 Chr.11: 113981518-113981518 on Build GRCh38
rs2276303 Chr.11: 113981477-113981477 on Build GRCh38
rs371778174 Chr.11: 113981302-113981302 on Build GRCh38
rs565827821 Chr.11: 113981470-113981470 on Build GRCh38
rs942182782 Chr.11: 113981547-113981547 on Build GRCh38
rs767668561 Chr.11: 113981200-113981200 on Build GRCh38
rs553593574 Chr.11: 113981144-113981144 on Build GRCh38
rs752208391 Chr.11: 113981332-113981332 on Build GRCh38
rs756043302 Chr.11: 113981280-113981280 on Build GRCh38
rs1046738984 Chr.11: 113981372-113981372 on Build GRCh38
rs544050111 Chr.11: 113981391-113981391 on Build GRCh38
rs569880486 Chr.11: 113981546-113981546 on Build GRCh38
rs114014601 Chr.11: 113981299-113981299 on Build GRCh38
rs113380330 Chr.11: 113981118-113981118 on Build GRCh38
rs772979890 Chr.11: 113981152-113981152 on Build GRCh38

View Full Product Details