Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114156233-114156728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB16
Entrez Gene ID
7704
Gene Name
zinc finger and BTB domain containing 16
Gene Aliases
PLZF, ZNF145
UniGene
Hs.591945
Gene Chromosome Location
Chr.11: 114059576-114256765 on Build GRCh38
Transcripts
NM_001018011.1 NM_006006.4 XM_005271658.4 XM_017018257.1 XM_017018258.1 XM_017018259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1351411 c.1297G>A p.G433R Chr.11: 114156365-114156365 on Build GRCh38
2103728 c.1336C>T p.R446W Chr.11: 114156404-114156404 on Build GRCh38
4017984 c.1319G>A p.R440Q Chr.11: 114156387-114156387 on Build GRCh38
923277 c.1292C>T p.T431M Chr.11: 114156360-114156360 on Build GRCh38
5569964 c.1293G>A p.T431T Chr.11: 114156361-114156361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775928369 Chr.11: 114156293-114156293 on Build GRCh38
rs973918061 Chr.11: 114156570-114156570 on Build GRCh38
rs765684473 Chr.11: 114156439-114156439 on Build GRCh38
rs970063190 Chr.11: 114156351-114156351 on Build GRCh38
rs1000015733 Chr.11: 114156266-114156266 on Build GRCh38
rs751235828 Chr.11: 114156329-114156329 on Build GRCh38
rs776974563 Chr.11: 114156416-114156416 on Build GRCh38
rs775652022 Chr.11: 114156391-114156391 on Build GRCh38
rs779160251 Chr.11: 114156370-114156370 on Build GRCh38
rs375573362 Chr.11: 114156361-114156361 on Build GRCh38
rs761248844 Chr.11: 114156298-114156298 on Build GRCh38
rs747010083 Chr.11: 114156405-114156405 on Build GRCh38
rs778683539 Chr.11: 114156483-114156483 on Build GRCh38
rs936901482 Chr.11: 114156660-114156660 on Build GRCh38
rs754594119 Chr.11: 114156330-114156330 on Build GRCh38
rs555396925 Chr.11: 114156498-114156498 on Build GRCh38
rs750447749 Chr.11: 114156355-114156355 on Build GRCh38
rs376503099 Chr.11: 114156451-114156451 on Build GRCh38
rs573180791 Chr.11: 114156280-114156280 on Build GRCh38
rs774522698 Chr.11: 114156289-114156289 on Build GRCh38
rs369356903 Chr.11: 114156312-114156312 on Build GRCh38
rs904451169 Chr.11: 114156265-114156265 on Build GRCh38
rs935941341 Chr.11: 114156536-114156536 on Build GRCh38
rs750371545 Chr.11: 114156480-114156482 on Build GRCh38
rs373467537 Chr.11: 114156510-114156510 on Build GRCh38
rs983581780 Chr.11: 114156442-114156442 on Build GRCh38
rs966213944 Chr.11: 114156683-114156683 on Build GRCh38
rs551098472 Chr.11: 114156539-114156539 on Build GRCh38
rs1015594040 Chr.11: 114156673-114156673 on Build GRCh38
rs1027651132 Chr.11: 114156639-114156639 on Build GRCh38
rs777073873 Chr.11: 114156321-114156321 on Build GRCh38
rs772121679 Chr.11: 114156292-114156292 on Build GRCh38
rs1020017730 Chr.11: 114156497-114156497 on Build GRCh38
rs754268555 Chr.11: 114156543-114156543 on Build GRCh38
rs148436647 Chr.11: 114156364-114156364 on Build GRCh38
rs1001472649 Chr.11: 114156611-114156611 on Build GRCh38
rs17116545 Chr.11: 114156555-114156555 on Build GRCh38
rs373139030 Chr.11: 114156441-114156441 on Build GRCh38
rs1024815188 Chr.11: 114156340-114156340 on Build GRCh38
rs912641146 Chr.11: 114156613-114156613 on Build GRCh38
rs752084825 Chr.11: 114156334-114156334 on Build GRCh38
rs1053942028 Chr.11: 114156666-114156666 on Build GRCh38
rs797011179 Chr.11: 114156387-114156387 on Build GRCh38
rs586486 Chr.11: 114156647-114156647 on Build GRCh38
rs879504234 Chr.11: 114156554-114156554 on Build GRCh38
rs371953230 Chr.11: 114156347-114156347 on Build GRCh38
rs767023648 Chr.11: 114156447-114156447 on Build GRCh38
rs538741723 Chr.11: 114156365-114156365 on Build GRCh38
rs563129809 Chr.11: 114156591-114156591 on Build GRCh38
rs929808150 Chr.11: 114156600-114156600 on Build GRCh38
rs765467140 Chr.11: 114156578-114156578 on Build GRCh38
rs559127482 Chr.11: 114156699-114156699 on Build GRCh38
rs746081118 Chr.11: 114156380-114156380 on Build GRCh38
rs983301434 Chr.11: 114156601-114156601 on Build GRCh38
rs113500641 Chr.11: 114156579-114156579 on Build GRCh38
rs141621748 Chr.11: 114156386-114156386 on Build GRCh38
rs952289419 Chr.11: 114156646-114156646 on Build GRCh38
rs904420692 Chr.11: 114156520-114156520 on Build GRCh38
rs370138532 Chr.11: 114156612-114156612 on Build GRCh38
rs757106053 Chr.11: 114156466-114156466 on Build GRCh38
rs762318942 Chr.11: 114156426-114156426 on Build GRCh38
rs762230974 Chr.11: 114156322-114156322 on Build GRCh38
rs369917864 Chr.11: 114156409-114156409 on Build GRCh38
rs372815069 Chr.11: 114156459-114156459 on Build GRCh38
rs766176520 Chr.11: 114156323-114156323 on Build GRCh38
rs532334827 Chr.11: 114156604-114156604 on Build GRCh38
rs1049449742 Chr.11: 114156515-114156515 on Build GRCh38
rs376433626 Chr.11: 114156303-114156303 on Build GRCh38
rs971503724 Chr.11: 114156512-114156512 on Build GRCh38
rs976489254 Chr.11: 114156454-114156454 on Build GRCh38
rs1039968348 Chr.11: 114156369-114156369 on Build GRCh38
rs905971786 Chr.11: 114156589-114156589 on Build GRCh38
rs182180285 Chr.11: 114156452-114156452 on Build GRCh38
rs767053945 Chr.11: 114156333-114156333 on Build GRCh38
rs375580497 Chr.11: 114156446-114156446 on Build GRCh38

View Full Product Details