Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1179105-1179603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL10
Entrez Gene ID
254173
Gene Name
tubulin tyrosine ligase like 10
Gene Aliases
TTLL5
UniGene
Hs.454835
Gene Chromosome Location
Chr.1: 1173709-1197936 on Build GRCh38
Transcripts
NM_001130045.1 XM_011541177.2 XM_017000906.1 XM_017000907.1 XM_017000908.1 XM_017000909.1 XM_017000910.1 XM_017000911.1 XM_017000912.1

Gene Symbol
TTLL10-AS1
Entrez Gene ID
100506376
Gene Name
TTLL10 antisense RNA 1
Gene Aliases
-
UniGene
Hs.672618
Gene Chromosome Location
Chr.1: 1173055-1179648 on Build GRCh38
Transcripts
NM_001130045.1 XM_011541177.2 XM_017000906.1 XM_017000907.1 XM_017000908.1 XM_017000909.1 XM_017000910.1 XM_017000911.1 XM_017000912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4888909 c.4G>T p.D2Y Chr.1: 1179219-1179219 on Build GRCh38
3417858 c.104G>A p.R35Q Chr.1: 1179319-1179319 on Build GRCh38
7327261 c.60C>T p.A20A Chr.1: 1179275-1179275 on Build GRCh38
1332545 c.103C>T p.R35W Chr.1: 1179318-1179318 on Build GRCh38
5349940 c.66C>T p.F22F Chr.1: 1179281-1179281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980751638 Chr.1: 1179324-1179324 on Build GRCh38
rs113718908 Chr.1: 1179538-1179538 on Build GRCh38
rs925210965 Chr.1: 1179381-1179381 on Build GRCh38
rs113596156 Chr.1: 1179319-1179319 on Build GRCh38
rs1029710479 Chr.1: 1179230-1179230 on Build GRCh38
rs894844270 Chr.1: 1179403-1179403 on Build GRCh38
rs995788046 Chr.1: 1179264-1179264 on Build GRCh38
rs1010732597 Chr.1: 1179531-1179531 on Build GRCh38
rs960740026 Chr.1: 1179568-1179568 on Build GRCh38
rs941573468 Chr.1: 1179250-1179250 on Build GRCh38
rs750373348 Chr.1: 1179247-1179247 on Build GRCh38
rs936499613 Chr.1: 1179561-1179561 on Build GRCh38
rs994316672 Chr.1: 1179261-1179261 on Build GRCh38
rs925157840 Chr.1: 1179560-1179560 on Build GRCh38
rs1050079403 Chr.1: 1179268-1179268 on Build GRCh38
rs982897467 Chr.1: 1179393-1179393 on Build GRCh38
rs1010784894 Chr.1: 1179298-1179298 on Build GRCh38
rs555277428 Chr.1: 1179271-1179271 on Build GRCh38
rs6668667 Chr.1: 1179288-1179288 on Build GRCh38
rs771603493 Chr.1: 1179331-1179331 on Build GRCh38
rs775787028 Chr.1: 1179463-1179463 on Build GRCh38
rs920162735 Chr.1: 1179173-1179173 on Build GRCh38
rs941533867 Chr.1: 1179421-1179421 on Build GRCh38
rs1041525528 Chr.1: 1179207-1179207 on Build GRCh38
rs983225144 Chr.1: 1179578-1179578 on Build GRCh38
rs1052188095 Chr.1: 1179519-1179519 on Build GRCh38
rs961840188 Chr.1: 1179165-1179165 on Build GRCh38
rs903756735 Chr.1: 1179221-1179221 on Build GRCh38
rs572857820 Chr.1: 1179382-1179382 on Build GRCh38
rs911267397 Chr.1: 1179287-1179292 on Build GRCh38
rs866512655 Chr.1: 1179423-1179423 on Build GRCh38
rs545062696 Chr.1: 1179420-1179420 on Build GRCh38
rs1018931430 Chr.1: 1179487-1179487 on Build GRCh38
rs747906470 Chr.1: 1179445-1179445 on Build GRCh38
rs755176679 Chr.1: 1179424-1179424 on Build GRCh38
rs1013656546 Chr.1: 1179414-1179414 on Build GRCh38
rs367795669 Chr.1: 1179276-1179276 on Build GRCh38
rs969270942 Chr.1: 1179537-1179537 on Build GRCh38
rs1013375048 Chr.1: 1179249-1179249 on Build GRCh38
rs752671231 Chr.1: 1179235-1179235 on Build GRCh38
rs188570709 Chr.1: 1179195-1179195 on Build GRCh38
rs77946374 Chr.1: 1179148-1179148 on Build GRCh38
rs992746328 Chr.1: 1179147-1179147 on Build GRCh38
rs758520312 Chr.1: 1179241-1179241 on Build GRCh38
rs538573333 Chr.1: 1179270-1179270 on Build GRCh38
rs763155019 Chr.1: 1179480-1179480 on Build GRCh38
rs544129424 Chr.1: 1179459-1179459 on Build GRCh38
rs765387292 Chr.1: 1179451-1179451 on Build GRCh38
rs923164712 Chr.1: 1179208-1179208 on Build GRCh38
rs977898869 Chr.1: 1179533-1179533 on Build GRCh38
rs1042312220 Chr.1: 1179210-1179210 on Build GRCh38
rs990779819 Chr.1: 1179584-1179584 on Build GRCh38
rs565046422 Chr.1: 1179138-1179138 on Build GRCh38
rs367773856 Chr.1: 1179199-1179199 on Build GRCh38
rs530077297 Chr.1: 1179526-1179526 on Build GRCh38
rs368507121 Chr.1: 1179144-1179144 on Build GRCh38
rs1047653809 Chr.1: 1179211-1179211 on Build GRCh38
rs1015856529 Chr.1: 1179130-1179130 on Build GRCh38
rs778717464 Chr.1: 1179318-1179318 on Build GRCh38
rs956810634 Chr.1: 1179192-1179192 on Build GRCh38
rs546679547 Chr.1: 1179548-1179548 on Build GRCh38
rs150461572 Chr.1: 1179139-1179139 on Build GRCh38
rs75179781 Chr.1: 1179367-1179367 on Build GRCh38
rs1002136068 Chr.1: 1179547-1179547 on Build GRCh38
rs539075182 Chr.1: 1179200-1179200 on Build GRCh38
rs1022161915 Chr.1: 1179534-1179534 on Build GRCh38
rs564951743 Chr.1: 1179429-1179429 on Build GRCh38
rs780112601 Chr.1: 1179299-1179299 on Build GRCh38
rs977136982 Chr.1: 1179509-1179509 on Build GRCh38
rs147449454 Chr.1: 1179583-1179583 on Build GRCh38
rs113258684 Chr.1: 1179431-1179431 on Build GRCh38
rs765475689 Chr.1: 1179205-1179205 on Build GRCh38
rs1050025785 Chr.1: 1179471-1179471 on Build GRCh38
rs900131007 Chr.1: 1179259-1179259 on Build GRCh38
rs560847385 Chr.1: 1179496-1179496 on Build GRCh38
rs1038589346 Chr.1: 1179258-1179258 on Build GRCh38
rs374835744 Chr.1: 1179172-1179172 on Build GRCh38
rs569439371 Chr.1: 1179246-1179246 on Build GRCh38
rs771467462 Chr.1: 1179388-1179388 on Build GRCh38
rs558907012 Chr.1: 1179204-1179204 on Build GRCh38
rs566843710 Chr.1: 1179196-1179196 on Build GRCh38
rs927971096 Chr.1: 1179326-1179326 on Build GRCh38
rs759721824 Chr.1: 1179202-1179202 on Build GRCh38
rs1027256656 Chr.1: 1179265-1179265 on Build GRCh38
rs12401465 Chr.1: 1179358-1179358 on Build GRCh38
rs375453018 Chr.1: 1179143-1179143 on Build GRCh38
rs908724920 Chr.1: 1179234-1179234 on Build GRCh38
rs939476634 Chr.1: 1179209-1179209 on Build GRCh38
rs976893386 Chr.1: 1179305-1179305 on Build GRCh38
rs1030857090 Chr.1: 1179174-1179174 on Build GRCh38
rs908662321 Chr.1: 1179398-1179398 on Build GRCh38

View Full Product Details