Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1334025-1334514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAS1R3
Entrez Gene ID
83756
Gene Name
taste 1 receptor member 3
Gene Aliases
T1R3
UniGene
Hs.74375
Gene Chromosome Location
Chr.1: 1331324-1335320 on Build GRCh38
Transcripts
NM_152228.2 XM_017002435.1 XM_017002436.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4004751 c.2423C>T p.A808V Chr.1: 1334328-1334328 on Build GRCh38
3472483 c.2439G>A p.R813R Chr.1: 1334344-1334344 on Build GRCh38
6373413 c.2139G>T p.M713I Chr.1: 1334044-1334044 on Build GRCh38
4386328 c.2311T>C p.Y771H Chr.1: 1334216-1334216 on Build GRCh38
5765782 c.2285G>A p.R762H Chr.1: 1334190-1334190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759018969 Chr.1: 1334053-1334053 on Build GRCh38
rs777698817 Chr.1: 1334085-1334085 on Build GRCh38
rs150053882 Chr.1: 1334353-1334353 on Build GRCh38
rs188379812 Chr.1: 1334359-1334359 on Build GRCh38
rs747310953 Chr.1: 1334224-1334224 on Build GRCh38
rs529036184 Chr.1: 1334160-1334160 on Build GRCh38
rs772170812 Chr.1: 1334451-1334451 on Build GRCh38
rs962929546 Chr.1: 1334193-1334193 on Build GRCh38
rs528566692 Chr.1: 1334175-1334175 on Build GRCh38
rs750640247 Chr.1: 1334127-1334127 on Build GRCh38
rs542365388 Chr.1: 1334487-1334487 on Build GRCh38
rs1026966893 Chr.1: 1334215-1334215 on Build GRCh38
rs775499172 Chr.1: 1334248-1334248 on Build GRCh38
rs141736258 Chr.1: 1334361-1334361 on Build GRCh38
rs139727182 Chr.1: 1334089-1334089 on Build GRCh38
rs754031951 Chr.1: 1334138-1334138 on Build GRCh38
rs1013490755 Chr.1: 1334076-1334076 on Build GRCh38
rs748069550 Chr.1: 1334079-1334079 on Build GRCh38
rs749544965 Chr.1: 1334382-1334382 on Build GRCh38
rs897096238 Chr.1: 1334402-1334402 on Build GRCh38
rs370778300 Chr.1: 1334059-1334059 on Build GRCh38
rs372183956 Chr.1: 1334187-1334187 on Build GRCh38
rs553610288 Chr.1: 1334349-1334349 on Build GRCh38
rs936431435 Chr.1: 1334358-1334358 on Build GRCh38
rs754117961 Chr.1: 1334179-1334179 on Build GRCh38
rs146326046 Chr.1: 1334275-1334275 on Build GRCh38
rs759693381 Chr.1: 1334314-1334314 on Build GRCh38
rs150645035 Chr.1: 1334065-1334065 on Build GRCh38
rs12030797 Chr.1: 1334372-1334372 on Build GRCh38
rs149300188 Chr.1: 1334190-1334190 on Build GRCh38
rs774483869 Chr.1: 1334384-1334384 on Build GRCh38
rs750869261 Chr.1: 1334486-1334486 on Build GRCh38
rs199779671 Chr.1: 1334243-1334243 on Build GRCh38
rs148758835 Chr.1: 1334422-1334422 on Build GRCh38
rs536836467 Chr.1: 1334344-1334344 on Build GRCh38
rs1027105596 Chr.1: 1334091-1334091 on Build GRCh38
rs748687102 Chr.1: 1334214-1334214 on Build GRCh38
rs967632727 Chr.1: 1334489-1334489 on Build GRCh38
rs776554702 Chr.1: 1334401-1334401 on Build GRCh38
rs760649453 Chr.1: 1334325-1334325 on Build GRCh38
rs771454609 Chr.1: 1334228-1334228 on Build GRCh38
rs769442638 Chr.1: 1334458-1334458 on Build GRCh38
rs34810828 Chr.1: 1334343-1334343 on Build GRCh38
rs141275308 Chr.1: 1334153-1334153 on Build GRCh38
rs762705846 Chr.1: 1334411-1334411 on Build GRCh38
rs987022778 Chr.1: 1334263-1334265 on Build GRCh38
rs1054964909 Chr.1: 1334071-1334071 on Build GRCh38
rs764262865 Chr.1: 1334317-1334317 on Build GRCh38
rs772348745 Chr.1: 1334194-1334196 on Build GRCh38
rs776502934 Chr.1: 1334452-1334452 on Build GRCh38
rs761362622 Chr.1: 1334476-1334476 on Build GRCh38
rs762196026 Chr.1: 1334242-1334242 on Build GRCh38
rs766214334 Chr.1: 1334268-1334268 on Build GRCh38
rs528530145 Chr.1: 1334492-1334492 on Build GRCh38
rs542451080 Chr.1: 1334162-1334162 on Build GRCh38
rs777140245 Chr.1: 1334068-1334068 on Build GRCh38
rs767338196 Chr.1: 1334188-1334188 on Build GRCh38
rs752270915 Chr.1: 1334409-1334409 on Build GRCh38
rs201702540 Chr.1: 1334292-1334292 on Build GRCh38
rs749470074 Chr.1: 1334442-1334442 on Build GRCh38
rs368525536 Chr.1: 1334441-1334441 on Build GRCh38
rs758478254 Chr.1: 1334084-1334084 on Build GRCh38
rs754466080 Chr.1: 1334209-1334209 on Build GRCh38
rs367565528 Chr.1: 1334159-1334159 on Build GRCh38
rs150371886 Chr.1: 1334416-1334416 on Build GRCh38
rs757402959 Chr.1: 1334062-1334062 on Build GRCh38
rs774906282 Chr.1: 1334212-1334212 on Build GRCh38
rs756382374 Chr.1: 1334434-1334434 on Build GRCh38
rs139536001 Chr.1: 1334281-1334281 on Build GRCh38
rs147581582 Chr.1: 1334380-1334380 on Build GRCh38
rs746404685 Chr.1: 1334426-1334426 on Build GRCh38
rs145890591 Chr.1: 1334303-1334303 on Build GRCh38
rs765104153 Chr.1: 1334408-1334408 on Build GRCh38
rs777786663 Chr.1: 1334146-1334146 on Build GRCh38
rs140102435 Chr.1: 1334285-1334285 on Build GRCh38
rs760850245 Chr.1: 1334137-1334137 on Build GRCh38
rs751985178 Chr.1: 1334060-1334060 on Build GRCh38
rs531899606 Chr.1: 1334233-1334235 on Build GRCh38
rs773785119 Chr.1: 1334221-1334221 on Build GRCh38
rs775420933 Chr.1: 1334465-1334465 on Build GRCh38
rs145836110 Chr.1: 1334360-1334360 on Build GRCh38
rs868194045 Chr.1: 1334430-1334430 on Build GRCh38
rs307377 Chr.1: 1334174-1334174 on Build GRCh38
rs772704578 Chr.1: 1334278-1334278 on Build GRCh38
rs146312553 Chr.1: 1334143-1334143 on Build GRCh38
rs146800023 Chr.1: 1334095-1334095 on Build GRCh38
rs746761454 Chr.1: 1334086-1334086 on Build GRCh38
rs143563187 Chr.1: 1334350-1334350 on Build GRCh38
rs766677901 Chr.1: 1334258-1334281 on Build GRCh38
rs79148073 Chr.1: 1334151-1334151 on Build GRCh38
rs760940327 Chr.1: 1334238-1334239 on Build GRCh38
rs371907990 Chr.1: 1334470-1334470 on Build GRCh38
rs147583678 Chr.1: 1334294-1334294 on Build GRCh38
rs774847924 Chr.1: 1334116-1334116 on Build GRCh38
rs1053089249 Chr.1: 1334152-1334152 on Build GRCh38
rs12030791 Chr.1: 1334351-1334351 on Build GRCh38
rs200098251 Chr.1: 1334369-1334369 on Build GRCh38
rs147441599 Chr.1: 1334052-1334052 on Build GRCh38
rs745528793 Chr.1: 1334096-1334096 on Build GRCh38
rs150531289 Chr.1: 1334479-1334479 on Build GRCh38
rs759569168 Chr.1: 1334456-1334456 on Build GRCh38
rs142104610 Chr.1: 1334276-1334276 on Build GRCh38
rs756175053 Chr.1: 1334073-1334073 on Build GRCh38
rs781735367 Chr.1: 1334427-1334427 on Build GRCh38
rs750377258 Chr.1: 1334263-1334263 on Build GRCh38
rs760596649 Chr.1: 1334397-1334397 on Build GRCh38
rs768983840 Chr.1: 1334443-1334443 on Build GRCh38
rs143005794 Chr.1: 1334400-1334400 on Build GRCh38
rs757930984 Chr.1: 1334282-1334282 on Build GRCh38
rs149196451 Chr.1: 1334123-1334123 on Build GRCh38
rs750240840 Chr.1: 1334189-1334189 on Build GRCh38
rs35913253 Chr.1: 1334399-1334399 on Build GRCh38
rs748967541 Chr.1: 1334200-1334200 on Build GRCh38
rs748734474 Chr.1: 1334168-1334168 on Build GRCh38
rs778103506 Chr.1: 1334287-1334287 on Build GRCh38
rs765367938 Chr.1: 1334326-1334327 on Build GRCh38
rs764684128 Chr.1: 1334056-1334056 on Build GRCh38
rs773082025 Chr.1: 1334320-1334320 on Build GRCh38
rs779124912 Chr.1: 1334445-1334445 on Build GRCh38
rs771399309 Chr.1: 1334293-1334293 on Build GRCh38
rs762870323 Chr.1: 1334315-1334315 on Build GRCh38
rs768155259 Chr.1: 1334169-1334169 on Build GRCh38
rs370702689 Chr.1: 1334110-1334110 on Build GRCh38
rs778475973 Chr.1: 1334279-1334279 on Build GRCh38
rs143394629 Chr.1: 1334182-1334182 on Build GRCh38
rs556963807 Chr.1: 1334447-1334447 on Build GRCh38
rs924936837 Chr.1: 1334346-1334346 on Build GRCh38
rs760739124 Chr.1: 1334318-1334318 on Build GRCh38
rs112507608 Chr.1: 1334108-1334108 on Build GRCh38
rs1002443009 Chr.1: 1334387-1334387 on Build GRCh38
rs769967165 Chr.1: 1334311-1334311 on Build GRCh38
rs901247435 Chr.1: 1334464-1334464 on Build GRCh38
rs373966526 Chr.1: 1334104-1334104 on Build GRCh38
rs774147677 Chr.1: 1334475-1334475 on Build GRCh38
rs200184398 Chr.1: 1334488-1334488 on Build GRCh38
rs781679369 Chr.1: 1334363-1334363 on Build GRCh38
rs780332472 Chr.1: 1334078-1334078 on Build GRCh38
rs543437187 Chr.1: 1334432-1334432 on Build GRCh38
rs779611258 Chr.1: 1334191-1334191 on Build GRCh38
rs536882133 Chr.1: 1334058-1334058 on Build GRCh38
rs376388007 Chr.1: 1334473-1334473 on Build GRCh38
rs767295745 Chr.1: 1334261-1334261 on Build GRCh38
rs780896820 Chr.1: 1334158-1334158 on Build GRCh38
rs982835883 Chr.1: 1334192-1334192 on Build GRCh38
rs766160710 Chr.1: 1334338-1334338 on Build GRCh38
rs377356981 Chr.1: 1334107-1334107 on Build GRCh38
rs776931966 Chr.1: 1334306-1334307 on Build GRCh38
rs368748861 Chr.1: 1334184-1334184 on Build GRCh38
rs750646328 Chr.1: 1334072-1334072 on Build GRCh38
rs577836548 Chr.1: 1334122-1334122 on Build GRCh38
rs746083552 Chr.1: 1334308-1334308 on Build GRCh38
rs934790028 Chr.1: 1334280-1334280 on Build GRCh38
rs145408540 Chr.1: 1334312-1334312 on Build GRCh38
rs772453723 Chr.1: 1334171-1334171 on Build GRCh38
rs200756454 Chr.1: 1334295-1334295 on Build GRCh38
rs751172616 Chr.1: 1334417-1334417 on Build GRCh38
rs776944125 Chr.1: 1334230-1334230 on Build GRCh38
rs543993276 Chr.1: 1334272-1334272 on Build GRCh38
rs765428998 Chr.1: 1334183-1334183 on Build GRCh38
rs375007877 Chr.1: 1334103-1334103 on Build GRCh38
rs139530347 Chr.1: 1334383-1334383 on Build GRCh38
rs761792916 Chr.1: 1334260-1334260 on Build GRCh38
rs775925398 Chr.1: 1334057-1334057 on Build GRCh38
rs755859716 Chr.1: 1334266-1334266 on Build GRCh38
rs375596902 Chr.1: 1334170-1334170 on Build GRCh38

View Full Product Details