Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130404832-130405326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS8
Entrez Gene ID
11095
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 8
Gene Aliases
ADAM-TS8, METH2
UniGene
Hs.271605
Gene Chromosome Location
Chr.11: 130404923-130428993 on Build GRCh38
Transcripts
NM_007037.5 XM_017017145.1

SNP Detail

SNP ID Chromosome Location
rs931777255 Chr.11: 130405033-130405033 on Build GRCh38
rs936480120 Chr.11: 130405198-130405198 on Build GRCh38
rs1015656465 Chr.11: 130405128-130405128 on Build GRCh38
rs555377340 Chr.11: 130405152-130405152 on Build GRCh38
rs548611210 Chr.11: 130404900-130404900 on Build GRCh38
rs552772675 Chr.11: 130404867-130404867 on Build GRCh38
rs766721394 Chr.11: 130405197-130405197 on Build GRCh38
rs1044899078 Chr.11: 130404933-130404933 on Build GRCh38
rs769399114 Chr.11: 130405283-130405284 on Build GRCh38
rs540996572 Chr.11: 130405260-130405260 on Build GRCh38
rs749355879 Chr.11: 130404961-130404961 on Build GRCh38
rs116377510 Chr.11: 130405140-130405140 on Build GRCh38
rs116078696 Chr.11: 130405075-130405075 on Build GRCh38
rs564049570 Chr.11: 130405160-130405160 on Build GRCh38
rs947975120 Chr.11: 130404904-130404904 on Build GRCh38
rs1007825232 Chr.11: 130405002-130405002 on Build GRCh38
rs1052165249 Chr.11: 130404990-130404990 on Build GRCh38
rs903725399 Chr.11: 130404934-130404934 on Build GRCh38
rs3214972 Chr.11: 130405224-130405224 on Build GRCh38
rs947996463 Chr.11: 130405193-130405193 on Build GRCh38
rs77754229 Chr.11: 130404958-130404958 on Build GRCh38
rs527812784 Chr.11: 130405032-130405032 on Build GRCh38
rs763018078 Chr.11: 130405167-130405168 on Build GRCh38
rs751920483 Chr.11: 130405226-130405226 on Build GRCh38
rs545547625 Chr.11: 130405037-130405037 on Build GRCh38
rs1035818421 Chr.11: 130405223-130405223 on Build GRCh38
rs755421197 Chr.11: 130405285-130405285 on Build GRCh38
rs992796048 Chr.11: 130405151-130405151 on Build GRCh38
rs769032179 Chr.11: 130405029-130405029 on Build GRCh38
rs981120894 Chr.11: 130405195-130405195 on Build GRCh38
rs571927859 Chr.11: 130405251-130405251 on Build GRCh38
rs924609126 Chr.11: 130404991-130404991 on Build GRCh38
rs944178094 Chr.11: 130405001-130405001 on Build GRCh38
rs943797707 Chr.11: 130405218-130405218 on Build GRCh38
rs1019700560 Chr.11: 130405015-130405015 on Build GRCh38
rs977360652 Chr.11: 130404970-130404970 on Build GRCh38
rs531493745 Chr.11: 130405215-130405215 on Build GRCh38
rs982849561 Chr.11: 130405078-130405078 on Build GRCh38
rs569660554 Chr.11: 130405076-130405076 on Build GRCh38
rs914803486 Chr.11: 130404915-130404915 on Build GRCh38
rs371528863 Chr.11: 130405038-130405038 on Build GRCh38
rs958920233 Chr.11: 130405283-130405283 on Build GRCh38
rs76580375 Chr.11: 130404884-130404884 on Build GRCh38
rs1005899826 Chr.11: 130405048-130405048 on Build GRCh38
rs539255392 Chr.11: 130405069-130405069 on Build GRCh38
rs959113809 Chr.11: 130404879-130404879 on Build GRCh38
rs1041505796 Chr.11: 130405008-130405008 on Build GRCh38
rs938691306 Chr.11: 130404860-130404860 on Build GRCh38
rs892176656 Chr.11: 130405216-130405216 on Build GRCh38
rs547597525 Chr.11: 130405034-130405034 on Build GRCh38

View Full Product Details