Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 192312-192809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A12
Entrez Gene ID
6539
Gene Name
solute carrier family 6 member 12
Gene Aliases
BGT-1, BGT1, GAT2
UniGene
Hs.437174
Gene Chromosome Location
Chr.12: 190077-214523 on Build GRCh38
Transcripts
NM_001122847.2 NM_001122848.2 NM_001206931.1 NM_003044.4 XM_005253747.4 XM_005253748.4 XM_006719005.3 XM_011521010.1 XM_017019841.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2205006 c.1659C>T p.F553F Chr.12: 192520-192520 on Build GRCh38
3460059 c.1601G>A p.W534* Chr.12: 192578-192578 on Build GRCh38
3704059 c.1547C>G p.S516C Chr.12: 192632-192632 on Build GRCh38
6783177 c.1567delC p.L523fs*37 Chr.12: 192612-192612 on Build GRCh38
4469652 c.1611C>T p.S537S Chr.12: 192568-192568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201448258 Chr.12: 192499-192499 on Build GRCh38
rs746418795 Chr.12: 192558-192558 on Build GRCh38
rs1021463051 Chr.12: 192455-192455 on Build GRCh38
rs763174593 Chr.12: 192464-192465 on Build GRCh38
rs548400421 Chr.12: 192763-192763 on Build GRCh38
rs935165598 Chr.12: 192540-192540 on Build GRCh38
rs191081528 Chr.12: 192705-192705 on Build GRCh38
rs941116377 Chr.12: 192615-192615 on Build GRCh38
rs646569 Chr.12: 192677-192677 on Build GRCh38
rs560878368 Chr.12: 192589-192589 on Build GRCh38
rs1034994396 Chr.12: 192365-192365 on Build GRCh38
rs543757957 Chr.12: 192404-192404 on Build GRCh38
rs202182580 Chr.12: 192620-192620 on Build GRCh38
rs535598322 Chr.12: 192755-192755 on Build GRCh38
rs577163007 Chr.12: 192494-192494 on Build GRCh38
rs576360403 Chr.12: 192399-192399 on Build GRCh38
rs1026246317 Chr.12: 192407-192407 on Build GRCh38
rs1000929911 Chr.12: 192750-192750 on Build GRCh38
rs751282302 Chr.12: 192505-192505 on Build GRCh38
rs763653166 Chr.12: 192490-192490 on Build GRCh38
rs530514022 Chr.12: 192646-192646 on Build GRCh38
rs770954629 Chr.12: 192551-192551 on Build GRCh38
rs1038190439 Chr.12: 192636-192636 on Build GRCh38
rs976315536 Chr.12: 192535-192535 on Build GRCh38
rs984801145 Chr.12: 192429-192429 on Build GRCh38
rs757446594 Chr.12: 192626-192626 on Build GRCh38
rs951555173 Chr.12: 192416-192417 on Build GRCh38
rs774772049 Chr.12: 192442-192442 on Build GRCh38
rs772698268 Chr.12: 192675-192675 on Build GRCh38
rs570170465 Chr.12: 192765-192765 on Build GRCh38
rs745940875 Chr.12: 192654-192654 on Build GRCh38
rs186392113 Chr.12: 192468-192468 on Build GRCh38
rs773266352 Chr.12: 192458-192458 on Build GRCh38
rs762988486 Chr.12: 192612-192612 on Build GRCh38
rs761369474 Chr.12: 192759-192759 on Build GRCh38
rs143357433 Chr.12: 192519-192519 on Build GRCh38
rs1009622029 Chr.12: 192441-192441 on Build GRCh38
rs559539489 Chr.12: 192502-192502 on Build GRCh38
rs563905232 Chr.12: 192688-192688 on Build GRCh38
rs896493990 Chr.12: 192423-192423 on Build GRCh38
rs779000283 Chr.12: 192641-192641 on Build GRCh38
rs370788960 Chr.12: 192493-192493 on Build GRCh38
rs749931281 Chr.12: 192600-192600 on Build GRCh38
rs138858909 Chr.12: 192520-192520 on Build GRCh38
rs775691317 Chr.12: 192482-192482 on Build GRCh38
rs1017642760 Chr.12: 192747-192747 on Build GRCh38
rs757339294 Chr.12: 192440-192440 on Build GRCh38
rs747063769 Chr.12: 192371-192371 on Build GRCh38
rs568283919 Chr.12: 192713-192713 on Build GRCh38
rs374653199 Chr.12: 192689-192689 on Build GRCh38
rs746086040 Chr.12: 192566-192566 on Build GRCh38
rs754008030 Chr.12: 192619-192619 on Build GRCh38
rs760411201 Chr.12: 192489-192489 on Build GRCh38
rs770140155 Chr.12: 192584-192584 on Build GRCh38
rs377552047 Chr.12: 192474-192474 on Build GRCh38
rs367825245 Chr.12: 192613-192613 on Build GRCh38
rs1039686086 Chr.12: 192648-192648 on Build GRCh38
rs775993391 Chr.12: 192582-192582 on Build GRCh38
rs750346557 Chr.12: 192496-192496 on Build GRCh38
rs767871917 Chr.12: 192481-192481 on Build GRCh38
rs771455437 Chr.12: 192712-192712 on Build GRCh38
rs75000134 Chr.12: 192583-192583 on Build GRCh38
rs376565192 Chr.12: 192542-192542 on Build GRCh38
rs766519638 Chr.12: 192683-192683 on Build GRCh38
rs756416342 Chr.12: 192517-192517 on Build GRCh38
rs772424014 Chr.12: 192567-192567 on Build GRCh38
rs1017705396 Chr.12: 192754-192754 on Build GRCh38
rs771438966 Chr.12: 192436-192436 on Build GRCh38
rs374806665 Chr.12: 192495-192495 on Build GRCh38
rs779416925 Chr.12: 192555-192555 on Build GRCh38
rs145636883 Chr.12: 192598-192598 on Build GRCh38
rs3741968 Chr.12: 192709-192709 on Build GRCh38
rs970366316 Chr.12: 192772-192772 on Build GRCh38
rs965434828 Chr.12: 192383-192383 on Build GRCh38
rs979562773 Chr.12: 192470-192470 on Build GRCh38
rs780371747 Chr.12: 192676-192676 on Build GRCh38
rs563357289 Chr.12: 192597-192597 on Build GRCh38
rs528428292 Chr.12: 192693-192693 on Build GRCh38
rs868867087 Chr.12: 192639-192639 on Build GRCh38

View Full Product Details