Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6841121-6841617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB3
Entrez Gene ID
2784
Gene Name
G protein subunit beta 3
Gene Aliases
CSNB1H
UniGene
Hs.631657
Gene Chromosome Location
Chr.12: 6840854-6847400 on Build GRCh38
Transcripts
NM_001297571.1 NM_002075.3 XM_011520953.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3464530 c.12G>A p.M4I Chr.12: 6841299-6841299 on Build GRCh38
1128481 c.59A>G p.D20G Chr.12: 6841587-6841587 on Build GRCh38
1363931 c.15G>A p.E5E Chr.12: 6841302-6841302 on Build GRCh38
695316 c.70G>C p.A24P Chr.12: 6841598-6841598 on Build GRCh38
4856354 c.33G>A p.A11A Chr.12: 6841320-6841320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782072645 Chr.12: 6841535-6841535 on Build GRCh38
rs782218646 Chr.12: 6841226-6841226 on Build GRCh38
rs782520640 Chr.12: 6841328-6841328 on Build GRCh38
rs782565385 Chr.12: 6841413-6841413 on Build GRCh38
rs913856195 Chr.12: 6841551-6841551 on Build GRCh38
rs782259270 Chr.12: 6841242-6841242 on Build GRCh38
rs782242015 Chr.12: 6841373-6841373 on Build GRCh38
rs781984234 Chr.12: 6841473-6841473 on Build GRCh38
rs566098814 Chr.12: 6841320-6841320 on Build GRCh38
rs942512229 Chr.12: 6841266-6841266 on Build GRCh38
rs782750899 Chr.12: 6841309-6841309 on Build GRCh38
rs779469264 Chr.12: 6841459-6841459 on Build GRCh38
rs782103240 Chr.12: 6841292-6841292 on Build GRCh38
rs782638077 Chr.12: 6841419-6841419 on Build GRCh38
rs782550076 Chr.12: 6841319-6841319 on Build GRCh38
rs925371824 Chr.12: 6841298-6841298 on Build GRCh38
rs782817413 Chr.12: 6841386-6841386 on Build GRCh38
rs13306408 Chr.12: 6841577-6841577 on Build GRCh38
rs782393579 Chr.12: 6841170-6841170 on Build GRCh38
rs925338623 Chr.12: 6841496-6841496 on Build GRCh38
rs782310728 Chr.12: 6841251-6841251 on Build GRCh38
rs370718226 Chr.12: 6841276-6841276 on Build GRCh38
rs781896099 Chr.12: 6841417-6841417 on Build GRCh38
rs782102378 Chr.12: 6841559-6841559 on Build GRCh38
rs995164982 Chr.12: 6841405-6841405 on Build GRCh38
rs199531660 Chr.12: 6841310-6841310 on Build GRCh38
rs781958487 Chr.12: 6841389-6841389 on Build GRCh38
rs912501024 Chr.12: 6841445-6841445 on Build GRCh38
rs146061530 Chr.12: 6841371-6841371 on Build GRCh38
rs28395781 Chr.12: 6841239-6841239 on Build GRCh38
rs531431756 Chr.12: 6841278-6841278 on Build GRCh38
rs782360979 Chr.12: 6841474-6841474 on Build GRCh38
rs782378563 Chr.12: 6841243-6841243 on Build GRCh38
rs782102571 Chr.12: 6841195-6841195 on Build GRCh38
rs755475906 Chr.12: 6841391-6841391 on Build GRCh38
rs782315249 Chr.12: 6841541-6841541 on Build GRCh38
rs13306409 Chr.12: 6841198-6841198 on Build GRCh38
rs754210939 Chr.12: 6841315-6841315 on Build GRCh38
rs781821917 Chr.12: 6841357-6841357 on Build GRCh38
rs189513536 Chr.12: 6841197-6841197 on Build GRCh38
rs782193272 Chr.12: 6841401-6841401 on Build GRCh38
rs963843562 Chr.12: 6841456-6841456 on Build GRCh38
rs782556422 Chr.12: 6841238-6841238 on Build GRCh38
rs561819792 Chr.12: 6841549-6841549 on Build GRCh38
rs376645329 Chr.12: 6841245-6841245 on Build GRCh38
rs781937681 Chr.12: 6841515-6841515 on Build GRCh38
rs782311919 Chr.12: 6841540-6841540 on Build GRCh38
rs782624468 Chr.12: 6841214-6841214 on Build GRCh38
rs1014286127 Chr.12: 6841360-6841360 on Build GRCh38
rs140043428 Chr.12: 6841418-6841418 on Build GRCh38
rs1025771772 Chr.12: 6841408-6841408 on Build GRCh38
rs1052437296 Chr.12: 6841518-6841518 on Build GRCh38
rs782605565 Chr.12: 6841504-6841504 on Build GRCh38
rs782144995 Chr.12: 6841275-6841275 on Build GRCh38
rs1046653764 Chr.12: 6841362-6841362 on Build GRCh38
rs782576144 Chr.12: 6841368-6841368 on Build GRCh38
rs781893197 Chr.12: 6841330-6841330 on Build GRCh38
rs951322641 Chr.12: 6841412-6841412 on Build GRCh38
rs180759247 Chr.12: 6841365-6841365 on Build GRCh38
rs782761828 Chr.12: 6841187-6841187 on Build GRCh38
rs201491226 Chr.12: 6841343-6841343 on Build GRCh38
rs372847142 Chr.12: 6841250-6841250 on Build GRCh38
rs782033753 Chr.12: 6841273-6841273 on Build GRCh38
rs781894685 Chr.12: 6841324-6841324 on Build GRCh38
rs367711032 Chr.12: 6841567-6841567 on Build GRCh38
rs782820356 Chr.12: 6841547-6841547 on Build GRCh38
rs34879671 Chr.12: 6841171-6841172 on Build GRCh38
rs962464992 Chr.12: 6841380-6841380 on Build GRCh38
rs782353867 Chr.12: 6841379-6841379 on Build GRCh38
rs892715161 Chr.12: 6841358-6841358 on Build GRCh38
rs782585832 Chr.12: 6841400-6841400 on Build GRCh38
rs971110533 Chr.12: 6841165-6841165 on Build GRCh38
rs371356560 Chr.12: 6841246-6841246 on Build GRCh38
rs975272732 Chr.12: 6841457-6841457 on Build GRCh38

View Full Product Details