Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6843475-6843984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB3
Entrez Gene ID
2784
Gene Name
G protein subunit beta 3
Gene Aliases
CSNB1H
UniGene
Hs.631657
Gene Chromosome Location
Chr.12: 6840854-6847400 on Build GRCh38
Transcripts
NM_001297571.1 NM_002075.3 XM_011520953.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
942957 c.525G>A p.Q175Q Chr.12: 6843804-6843804 on Build GRCh38
942958 c.691G>A p.A231T Chr.12: 6843970-6843970 on Build GRCh38
5841803 c.487G>A p.D163N Chr.12: 6843688-6843688 on Build GRCh38
5889203 c.424C>T p.H142Y Chr.12: 6843519-6843519 on Build GRCh38
1208412 c.512T>G p.I171S Chr.12: 6843791-6843791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201972812 Chr.12: 6843766-6843766 on Build GRCh38
rs45476395 Chr.12: 6843936-6843936 on Build GRCh38
rs782750354 Chr.12: 6843833-6843833 on Build GRCh38
rs200309858 Chr.12: 6843639-6843639 on Build GRCh38
rs782769083 Chr.12: 6843959-6843959 on Build GRCh38
rs28395777 Chr.12: 6843540-6843540 on Build GRCh38
rs75977062 Chr.12: 6843684-6843684 on Build GRCh38
rs782227813 Chr.12: 6843943-6843943 on Build GRCh38
rs372997527 Chr.12: 6843742-6843742 on Build GRCh38
rs113742693 Chr.12: 6843578-6843578 on Build GRCh38
rs201117275 Chr.12: 6843500-6843500 on Build GRCh38
rs28395779 Chr.12: 6843756-6843756 on Build GRCh38
rs782351294 Chr.12: 6843663-6843663 on Build GRCh38
rs782433943 Chr.12: 6843890-6843890 on Build GRCh38
rs781918900 Chr.12: 6843825-6843825 on Build GRCh38
rs1023757128 Chr.12: 6843938-6843938 on Build GRCh38
rs781988763 Chr.12: 6843659-6843659 on Build GRCh38
rs368740450 Chr.12: 6843648-6843648 on Build GRCh38
rs782433777 Chr.12: 6843931-6843931 on Build GRCh38
rs782159117 Chr.12: 6843826-6843826 on Build GRCh38
rs782113998 Chr.12: 6843832-6843832 on Build GRCh38
rs781940063 Chr.12: 6843666-6843666 on Build GRCh38
rs940510787 Chr.12: 6843515-6843515 on Build GRCh38
rs782008185 Chr.12: 6843894-6843894 on Build GRCh38
rs782013128 Chr.12: 6843539-6843539 on Build GRCh38
rs782530958 Chr.12: 6843741-6843741 on Build GRCh38
rs781826431 Chr.12: 6843835-6843835 on Build GRCh38
rs782199265 Chr.12: 6843641-6843641 on Build GRCh38
rs781821386 Chr.12: 6843502-6843502 on Build GRCh38
rs782538568 Chr.12: 6843859-6843859 on Build GRCh38
rs367934996 Chr.12: 6843919-6843919 on Build GRCh38
rs782054708 Chr.12: 6843673-6843673 on Build GRCh38
rs782728633 Chr.12: 6843923-6843923 on Build GRCh38
rs781988924 Chr.12: 6843657-6843657 on Build GRCh38
rs782409537 Chr.12: 6843527-6843527 on Build GRCh38
rs782615277 Chr.12: 6843763-6843763 on Build GRCh38
rs533829744 Chr.12: 6843739-6843739 on Build GRCh38
rs782088624 Chr.12: 6843906-6843906 on Build GRCh38
rs781903842 Chr.12: 6843846-6843846 on Build GRCh38
rs782654668 Chr.12: 6843867-6843867 on Build GRCh38
rs782266619 Chr.12: 6843934-6843934 on Build GRCh38
rs371387315 Chr.12: 6843830-6843830 on Build GRCh38
rs782471427 Chr.12: 6843836-6843836 on Build GRCh38
rs782697401 Chr.12: 6843701-6843701 on Build GRCh38
rs369737617 Chr.12: 6843571-6843571 on Build GRCh38
rs138319468 Chr.12: 6843783-6843783 on Build GRCh38
rs782388688 Chr.12: 6843650-6843650 on Build GRCh38
rs781795582 Chr.12: 6843911-6843911 on Build GRCh38
rs782790179 Chr.12: 6843882-6843882 on Build GRCh38
rs782683626 Chr.12: 6843778-6843778 on Build GRCh38
rs147291332 Chr.12: 6843820-6843820 on Build GRCh38
rs782166953 Chr.12: 6843913-6843913 on Build GRCh38
rs782392907 Chr.12: 6843801-6843801 on Build GRCh38
rs1040460845 Chr.12: 6843557-6843557 on Build GRCh38
rs781904771 Chr.12: 6843630-6843630 on Build GRCh38
rs377593669 Chr.12: 6843623-6843623 on Build GRCh38
rs782199199 Chr.12: 6843765-6843765 on Build GRCh38
rs781943176 Chr.12: 6843948-6843948 on Build GRCh38
rs781852575 Chr.12: 6843631-6843631 on Build GRCh38
rs782808953 Chr.12: 6843918-6843918 on Build GRCh38
rs782476867 Chr.12: 6843632-6843632 on Build GRCh38
rs139141966 Chr.12: 6843837-6843837 on Build GRCh38
rs374134015 Chr.12: 6843960-6843960 on Build GRCh38
rs782317079 Chr.12: 6843775-6843775 on Build GRCh38
rs782717038 Chr.12: 6843618-6843618 on Build GRCh38
rs782092334 Chr.12: 6843617-6843617 on Build GRCh38
rs781936811 Chr.12: 6843517-6843517 on Build GRCh38
rs782673755 Chr.12: 6843932-6843932 on Build GRCh38
rs782343782 Chr.12: 6843509-6843509 on Build GRCh38
rs782313482 Chr.12: 6843792-6843793 on Build GRCh38
rs782199511 Chr.12: 6843505-6843505 on Build GRCh38
rs144008405 Chr.12: 6843791-6843791 on Build GRCh38
rs782486451 Chr.12: 6843757-6843757 on Build GRCh38
rs782712051 Chr.12: 6843910-6843910 on Build GRCh38
rs781970278 Chr.12: 6843609-6843609 on Build GRCh38
rs781804079 Chr.12: 6843928-6843928 on Build GRCh38
rs782508077 Chr.12: 6843922-6843922 on Build GRCh38
rs782249757 Chr.12: 6843881-6843881 on Build GRCh38
rs782269397 Chr.12: 6843649-6843649 on Build GRCh38
rs373443639 Chr.12: 6843504-6843504 on Build GRCh38
rs782321891 Chr.12: 6843824-6843824 on Build GRCh38
rs782002919 Chr.12: 6843841-6843841 on Build GRCh38
rs376227398 Chr.12: 6843828-6843828 on Build GRCh38
rs1041974039 Chr.12: 6843869-6843869 on Build GRCh38
rs373704098 Chr.12: 6843598-6843598 on Build GRCh38
rs200634640 Chr.12: 6843899-6843899 on Build GRCh38
rs376301702 Chr.12: 6843526-6843526 on Build GRCh38
rs782432503 Chr.12: 6843642-6843642 on Build GRCh38
rs200864177 Chr.12: 6843660-6843660 on Build GRCh38
rs903326509 Chr.12: 6843874-6843874 on Build GRCh38
rs150490302 Chr.12: 6843635-6843635 on Build GRCh38
rs782568174 Chr.12: 6843624-6843624 on Build GRCh38
rs781898297 Chr.12: 6843710-6843710 on Build GRCh38
rs373970960 Chr.12: 6843900-6843900 on Build GRCh38
rs782022033 Chr.12: 6843690-6843690 on Build GRCh38
rs782349724 Chr.12: 6843947-6843947 on Build GRCh38
rs782505727 Chr.12: 6843935-6843935 on Build GRCh38
rs782130029 Chr.12: 6843695-6843695 on Build GRCh38

View Full Product Details