Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7654189-7654731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC1
Entrez Gene ID
339
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 1
Gene Aliases
APOBEC-1, BEDP, CDAR1, HEPR
UniGene
Hs.560
Gene Chromosome Location
Chr.12: 7649400-7670599 on Build GRCh38
Transcripts
NM_001304566.1 NM_001644.4 NM_005889.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6032082 c.44+3A>C p.? Chr.12: 7654602-7654602 on Build GRCh38
468922 c.27C>T p.T9T Chr.12: 7654622-7654622 on Build GRCh38
4966408 c.40C>T p.L14L Chr.12: 7654609-7654609 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374101878 Chr.12: 7654475-7654475 on Build GRCh38
rs35741011 Chr.12: 7654389-7654390 on Build GRCh38
rs10845624 Chr.12: 7654542-7654542 on Build GRCh38
rs947671594 Chr.12: 7654615-7654615 on Build GRCh38
rs774901084 Chr.12: 7654594-7654594 on Build GRCh38
rs371967494 Chr.12: 7654641-7654641 on Build GRCh38
rs766222114 Chr.12: 7654659-7654659 on Build GRCh38
rs927079859 Chr.12: 7654503-7654503 on Build GRCh38
rs750794607 Chr.12: 7654343-7654343 on Build GRCh38
rs755623668 Chr.12: 7654684-7654684 on Build GRCh38
rs868330247 Chr.12: 7654626-7654626 on Build GRCh38
rs760987232 Chr.12: 7654622-7654622 on Build GRCh38
rs34404451 Chr.12: 7654269-7654269 on Build GRCh38
rs889838814 Chr.12: 7654707-7654707 on Build GRCh38
rs956283654 Chr.12: 7654413-7654414 on Build GRCh38
rs796289429 Chr.12: 7654373-7654374 on Build GRCh38
rs1033598900 Chr.12: 7654409-7654409 on Build GRCh38
rs184418061 Chr.12: 7654276-7654276 on Build GRCh38
rs188371284 Chr.12: 7654522-7654522 on Build GRCh38
rs780875750 Chr.12: 7654677-7654677 on Build GRCh38
rs373055650 Chr.12: 7654372-7654373 on Build GRCh38
rs376031166 Chr.12: 7654590-7654590 on Build GRCh38
rs143770665 Chr.12: 7654611-7654611 on Build GRCh38
rs1048846866 Chr.12: 7654534-7654534 on Build GRCh38
rs143925738 Chr.12: 7654571-7654571 on Build GRCh38
rs958986645 Chr.12: 7654297-7654297 on Build GRCh38
rs1003406474 Chr.12: 7654248-7654248 on Build GRCh38
rs1023519456 Chr.12: 7654371-7654371 on Build GRCh38
rs78377629 Chr.12: 7654234-7654234 on Build GRCh38
rs374976210 Chr.12: 7654679-7654679 on Build GRCh38
rs754540773 Chr.12: 7654566-7654566 on Build GRCh38
rs770317987 Chr.12: 7654416-7654416 on Build GRCh38
rs1000408388 Chr.12: 7654366-7654366 on Build GRCh38
rs984617469 Chr.12: 7654431-7654431 on Build GRCh38
rs377465246 Chr.12: 7654523-7654523 on Build GRCh38
rs954188159 Chr.12: 7654356-7654356 on Build GRCh38
rs371540354 Chr.12: 7654668-7654668 on Build GRCh38
rs971060834 Chr.12: 7654427-7654428 on Build GRCh38
rs200875462 Chr.12: 7654577-7654577 on Build GRCh38
rs961802939 Chr.12: 7654474-7654474 on Build GRCh38
rs375025844 Chr.12: 7654654-7654654 on Build GRCh38
rs772496243 Chr.12: 7654613-7654613 on Build GRCh38
rs764450370 Chr.12: 7654625-7654625 on Build GRCh38
rs907749016 Chr.12: 7654453-7654453 on Build GRCh38
rs979808468 Chr.12: 7654479-7654479 on Build GRCh38
rs916888941 Chr.12: 7654426-7654426 on Build GRCh38
rs1012939014 Chr.12: 7654311-7654311 on Build GRCh38
rs34124287 Chr.12: 7654388-7654389 on Build GRCh38
rs778537982 Chr.12: 7654668-7654668 on Build GRCh38
rs767202746 Chr.12: 7654374-7654375 on Build GRCh38
rs781757439 Chr.12: 7654344-7654344 on Build GRCh38
rs951953110 Chr.12: 7654410-7654410 on Build GRCh38
rs771691080 Chr.12: 7654562-7654562 on Build GRCh38
rs368970083 Chr.12: 7654640-7654640 on Build GRCh38
rs776970860 Chr.12: 7654630-7654630 on Build GRCh38
rs751276266 Chr.12: 7654662-7654662 on Build GRCh38
rs35969738 Chr.12: 7654342-7654343 on Build GRCh38
rs368371432 Chr.12: 7654658-7654658 on Build GRCh38
rs148640043 Chr.12: 7654621-7654621 on Build GRCh38
rs1035675499 Chr.12: 7654252-7654252 on Build GRCh38
rs765582027 Chr.12: 7654229-7654229 on Build GRCh38
rs531933238 Chr.12: 7654301-7654302 on Build GRCh38

View Full Product Details