Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10379307-10379831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLRC4-KLRK1
Entrez Gene ID
100528032
Gene Name
KLRC4-KLRK1 readthrough
Gene Aliases
-
UniGene
Hs.387787
Gene Chromosome Location
Chr.12: 10372353-10410146 on Build GRCh38
Transcripts
NM_001199805.1 NM_007360.3 NR_120430.1

Gene Symbol
KLRK1
Entrez Gene ID
22914
Gene Name
killer cell lectin like receptor K1
Gene Aliases
CD314, D12S2489E, KLR, NKG2-D, NKG2D
UniGene
Hs.387787
Gene Chromosome Location
Chr.12: 10372353-10390054 on Build GRCh38
Transcripts
NM_001199805.1 NM_007360.3 NR_120430.1

Gene Symbol
LOC101928100
Entrez Gene ID
101928100
Gene Name
uncharacterized LOC101928100
Gene Aliases
-
UniGene
Hs.638425
Gene Chromosome Location
Chr.12: 10363769-10398506 on Build GRCh38
Transcripts
NM_001199805.1 NM_007360.3 NR_120430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5970469 c.227C>A p.A76D Chr.12: 10379714-10379714 on Build GRCh38
1322040 c.253C>G p.Q85E Chr.12: 10379471-10379471 on Build GRCh38
1358384 c.164delT p.F55fs*7 Chr.12: 10379777-10379777 on Build GRCh38
332216 c.220T>C p.W74R Chr.12: 10379721-10379721 on Build GRCh38
4038207 c.177C>T p.I59I Chr.12: 10379764-10379764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776190059 Chr.12: 10379432-10379432 on Build GRCh38
rs763969539 Chr.12: 10379454-10379454 on Build GRCh38
rs780339405 Chr.12: 10379398-10379398 on Build GRCh38
rs143021072 Chr.12: 10379448-10379448 on Build GRCh38
rs758961194 Chr.12: 10379676-10379676 on Build GRCh38
rs141717786 Chr.12: 10379745-10379745 on Build GRCh38
rs564026526 Chr.12: 10379779-10379779 on Build GRCh38
rs774264165 Chr.12: 10379714-10379714 on Build GRCh38
rs987757104 Chr.12: 10379590-10379590 on Build GRCh38
rs768301645 Chr.12: 10379707-10379707 on Build GRCh38
rs774643686 Chr.12: 10379447-10379447 on Build GRCh38
rs557219486 Chr.12: 10379763-10379763 on Build GRCh38
rs530425182 Chr.12: 10379412-10379412 on Build GRCh38
rs1056080787 Chr.12: 10379361-10379361 on Build GRCh38
rs774130083 Chr.12: 10379456-10379456 on Build GRCh38
rs749014268 Chr.12: 10379705-10379705 on Build GRCh38
rs764660013 Chr.12: 10379776-10379777 on Build GRCh38
rs752648392 Chr.12: 10379358-10379358 on Build GRCh38
rs894716874 Chr.12: 10379371-10379371 on Build GRCh38
rs112348489 Chr.12: 10379419-10379419 on Build GRCh38
rs761718210 Chr.12: 10379721-10379721 on Build GRCh38
rs192224382 Chr.12: 10379545-10379545 on Build GRCh38
rs2255353 Chr.12: 10379512-10379512 on Build GRCh38
rs749551753 Chr.12: 10379406-10379406 on Build GRCh38
rs755554166 Chr.12: 10379774-10379774 on Build GRCh38
rs911903474 Chr.12: 10379604-10379604 on Build GRCh38
rs773316467 Chr.12: 10379734-10379734 on Build GRCh38
rs891691492 Chr.12: 10379355-10379355 on Build GRCh38
rs753169996 Chr.12: 10379529-10379533 on Build GRCh38
rs766532536 Chr.12: 10379744-10379744 on Build GRCh38
rs768121599 Chr.12: 10379468-10379468 on Build GRCh38
rs200989597 Chr.12: 10379764-10379764 on Build GRCh38
rs2255336 Chr.12: 10379727-10379727 on Build GRCh38
rs765505163 Chr.12: 10379746-10379746 on Build GRCh38
rs748738337 Chr.12: 10379415-10379415 on Build GRCh38
rs376770387 Chr.12: 10379646-10379646 on Build GRCh38
rs758765044 Chr.12: 10379753-10379753 on Build GRCh38
rs865824255 Chr.12: 10379731-10379731 on Build GRCh38
rs779376008 Chr.12: 10379653-10379653 on Build GRCh38
rs753001440 Chr.12: 10379750-10379750 on Build GRCh38
rs778264314 Chr.12: 10379685-10379685 on Build GRCh38
rs1021429398 Chr.12: 10379392-10379392 on Build GRCh38
rs550400769 Chr.12: 10379449-10379449 on Build GRCh38
rs201221213 Chr.12: 10379484-10379484 on Build GRCh38
rs745309364 Chr.12: 10379691-10379691 on Build GRCh38
rs778717448 Chr.12: 10379794-10379794 on Build GRCh38
rs368970673 Chr.12: 10379668-10379668 on Build GRCh38
rs182882070 Chr.12: 10379695-10379695 on Build GRCh38
rs532525963 Chr.12: 10379510-10379510 on Build GRCh38
rs770398356 Chr.12: 10379417-10379417 on Build GRCh38
rs756606329 Chr.12: 10379777-10379777 on Build GRCh38
rs755277497 Chr.12: 10379649-10379649 on Build GRCh38
rs769098245 Chr.12: 10379410-10379410 on Build GRCh38
rs545769255 Chr.12: 10379781-10379781 on Build GRCh38
rs756269038 Chr.12: 10379487-10379487 on Build GRCh38
rs377167821 Chr.12: 10379738-10379738 on Build GRCh38
rs534769105 Chr.12: 10379563-10379563 on Build GRCh38
rs963556402 Chr.12: 10379690-10379690 on Build GRCh38
rs768301216 Chr.12: 10379785-10379785 on Build GRCh38
rs953316564 Chr.12: 10379589-10379589 on Build GRCh38
rs761877770 Chr.12: 10379469-10379469 on Build GRCh38
rs201822593 Chr.12: 10379452-10379452 on Build GRCh38
rs745505791 Chr.12: 10379440-10379440 on Build GRCh38

View Full Product Details