Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10908441-10908953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRH1
Entrez Gene ID
5554
Gene Name
proline rich protein HaeIII subfamily 1
Gene Aliases
Db-s, PA, PIF-S, PRH2, Pr1/Pr2
UniGene
Hs.656965
Gene Chromosome Location
Chr.12: 10880961-11171623 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_037918.2 NM_023920.2

Gene Symbol
PRH1-PRR4
Entrez Gene ID
100533464
Gene Name
PRH1-PRR4 readthrough
Gene Aliases
PRH1
UniGene
Hs.732031
Gene Chromosome Location
Chr.12: 10845849-11171625 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_037918.2 NM_023920.2

Gene Symbol
TAS2R13
Entrez Gene ID
50838
Gene Name
taste 2 receptor member 13
Gene Aliases
T2R13, TRB3
UniGene
Hs.679407
Gene Chromosome Location
Chr.12: 10907926-10909562 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_037918.2 NM_023920.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
934928 c.838C>A p.L280I Chr.12: 10908461-10908461 on Build GRCh38
4038631 c.788A>G p.Y263C Chr.12: 10908511-10908511 on Build GRCh38
7285488 c.804G>A p.T268T Chr.12: 10908495-10908495 on Build GRCh38
4186499 c.538A>C p.K180Q Chr.12: 10908761-10908761 on Build GRCh38
934931 c.351T>A p.P117P Chr.12: 10908948-10908948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768082133 Chr.12: 10908716-10908716 on Build GRCh38
rs771960931 Chr.12: 10908640-10908640 on Build GRCh38
rs149109598 Chr.12: 10908529-10908529 on Build GRCh38
rs183634674 Chr.12: 10908862-10908862 on Build GRCh38
rs765620420 Chr.12: 10908478-10908478 on Build GRCh38
rs759545746 Chr.12: 10908660-10908660 on Build GRCh38
rs141663427 Chr.12: 10908759-10908759 on Build GRCh38
rs1015214276 Chr.12: 10908914-10908914 on Build GRCh38
rs748636420 Chr.12: 10908658-10908658 on Build GRCh38
rs148234617 Chr.12: 10908752-10908752 on Build GRCh38
rs199740114 Chr.12: 10908471-10908471 on Build GRCh38
rs753408054 Chr.12: 10908737-10908737 on Build GRCh38
rs747009314 Chr.12: 10908645-10908645 on Build GRCh38
rs1042593012 Chr.12: 10908491-10908491 on Build GRCh38
rs975168806 Chr.12: 10908778-10908778 on Build GRCh38
rs201308754 Chr.12: 10908572-10908572 on Build GRCh38
rs140528790 Chr.12: 10908687-10908687 on Build GRCh38
rs754668618 Chr.12: 10908639-10908639 on Build GRCh38
rs752111217 Chr.12: 10908850-10908850 on Build GRCh38
rs769858966 Chr.12: 10908677-10908677 on Build GRCh38
rs567360016 Chr.12: 10908594-10908594 on Build GRCh38
rs774795106 Chr.12: 10908811-10908811 on Build GRCh38
rs959035226 Chr.12: 10908569-10908569 on Build GRCh38
rs752444755 Chr.12: 10908751-10908751 on Build GRCh38
rs780727492 Chr.12: 10908643-10908643 on Build GRCh38
rs780202524 Chr.12: 10908899-10908899 on Build GRCh38
rs762435629 Chr.12: 10908713-10908713 on Build GRCh38
rs779601010 Chr.12: 10908632-10908634 on Build GRCh38
rs1015443 Chr.12: 10908523-10908523 on Build GRCh38
rs746520939 Chr.12: 10908634-10908635 on Build GRCh38
rs747751631 Chr.12: 10908637-10908637 on Build GRCh38
rs777657052 Chr.12: 10908753-10908753 on Build GRCh38
rs567736542 Chr.12: 10908680-10908680 on Build GRCh38
rs200499053 Chr.12: 10908861-10908861 on Build GRCh38
rs745787811 Chr.12: 10908765-10908765 on Build GRCh38
rs780938834 Chr.12: 10908867-10908867 on Build GRCh38
rs764844013 Chr.12: 10908847-10908847 on Build GRCh38
rs570122150 Chr.12: 10908814-10908814 on Build GRCh38
rs749272501 Chr.12: 10908903-10908903 on Build GRCh38
rs763264582 Chr.12: 10908783-10908783 on Build GRCh38
rs781172305 Chr.12: 10908760-10908760 on Build GRCh38
rs753009429 Chr.12: 10908484-10908484 on Build GRCh38
rs762206371 Chr.12: 10908812-10908812 on Build GRCh38
rs376987737 Chr.12: 10908808-10908808 on Build GRCh38
rs374041435 Chr.12: 10908490-10908490 on Build GRCh38
rs747923980 Chr.12: 10908519-10908519 on Build GRCh38
rs775398971 Chr.12: 10908769-10908769 on Build GRCh38
rs150397675 Chr.12: 10908469-10908469 on Build GRCh38
rs375488942 Chr.12: 10908721-10908721 on Build GRCh38
rs376237809 Chr.12: 10908465-10908465 on Build GRCh38
rs145310730 Chr.12: 10908869-10908869 on Build GRCh38
rs755704027 Chr.12: 10908493-10908493 on Build GRCh38
rs779745131 Chr.12: 10908619-10908619 on Build GRCh38
rs772614164 Chr.12: 10908925-10908925 on Build GRCh38
rs776650571 Chr.12: 10908549-10908549 on Build GRCh38
rs748371831 Chr.12: 10908923-10908923 on Build GRCh38
rs776703676 Chr.12: 10908656-10908656 on Build GRCh38
rs779686004 Chr.12: 10908495-10908495 on Build GRCh38
rs765385137 Chr.12: 10908561-10908561 on Build GRCh38
rs368652295 Chr.12: 10908758-10908758 on Build GRCh38
rs757706967 Chr.12: 10908518-10908519 on Build GRCh38
rs370855654 Chr.12: 10908690-10908690 on Build GRCh38
rs771911613 Chr.12: 10908522-10908522 on Build GRCh38
rs367621923 Chr.12: 10908531-10908531 on Build GRCh38
rs754120347 Chr.12: 10908475-10908475 on Build GRCh38
rs759774725 Chr.12: 10908559-10908559 on Build GRCh38
rs746721818 Chr.12: 10908754-10908754 on Build GRCh38
rs751826901 Chr.12: 10908708-10908708 on Build GRCh38
rs894927278 Chr.12: 10908819-10908819 on Build GRCh38
rs758842159 Chr.12: 10908485-10908485 on Build GRCh38
rs768386640 Chr.12: 10908509-10908509 on Build GRCh38
rs200842780 Chr.12: 10908746-10908746 on Build GRCh38
rs372765078 Chr.12: 10908784-10908784 on Build GRCh38
rs1001141532 Chr.12: 10908525-10908525 on Build GRCh38
rs769906007 Chr.12: 10908766-10908766 on Build GRCh38
rs143136566 Chr.12: 10908651-10908651 on Build GRCh38
rs754606395 Chr.12: 10908744-10908744 on Build GRCh38
rs867568209 Chr.12: 10908634-10908634 on Build GRCh38
rs777222102 Chr.12: 10908853-10908853 on Build GRCh38
rs761242032 Chr.12: 10908836-10908836 on Build GRCh38
rs1024702065 Chr.12: 10908631-10908631 on Build GRCh38
rs754694608 Chr.12: 10908630-10908630 on Build GRCh38
rs769675563 Chr.12: 10908897-10908897 on Build GRCh38
rs370308524 Chr.12: 10908577-10908577 on Build GRCh38
rs370341557 Chr.12: 10908466-10908466 on Build GRCh38
rs142394171 Chr.12: 10908908-10908908 on Build GRCh38
rs764557255 Chr.12: 10908705-10908705 on Build GRCh38
rs143948129 Chr.12: 10908515-10908515 on Build GRCh38
rs970977617 Chr.12: 10908567-10908567 on Build GRCh38
rs767994152 Chr.12: 10908822-10908822 on Build GRCh38
rs757671004 Chr.12: 10908599-10908599 on Build GRCh38
rs751029522 Chr.12: 10908829-10908829 on Build GRCh38
rs763059068 Chr.12: 10908684-10908684 on Build GRCh38
rs137927359 Chr.12: 10908882-10908882 on Build GRCh38
rs972525830 Chr.12: 10908652-10908652 on Build GRCh38
rs902722400 Chr.12: 10908833-10908833 on Build GRCh38
rs748760325 Chr.12: 10908626-10908626 on Build GRCh38
rs772862867 Chr.12: 10908643-10908643 on Build GRCh38

View Full Product Details