Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10909178-10909709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRH1
Entrez Gene ID
5554
Gene Name
proline rich protein HaeIII subfamily 1
Gene Aliases
Db-s, PA, PIF-S, PRH2, Pr1/Pr2
UniGene
Hs.656965
Gene Chromosome Location
Chr.12: 10880961-11171623 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_037918.2 NM_023920.2

Gene Symbol
PRH1-PRR4
Entrez Gene ID
100533464
Gene Name
PRH1-PRR4 readthrough
Gene Aliases
PRH1
UniGene
Hs.732031
Gene Chromosome Location
Chr.12: 10845849-11171625 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_037918.2 NM_023920.2

Gene Symbol
TAS2R13
Entrez Gene ID
50838
Gene Name
taste 2 receptor member 13
Gene Aliases
T2R13, TRB3
UniGene
Hs.679407
Gene Chromosome Location
Chr.12: 10907926-10909562 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_037918.2 NM_023920.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3967974 c.15G>A p.L5L Chr.12: 10909284-10909284 on Build GRCh38
1298903 c.34G>C p.V12L Chr.12: 10909265-10909265 on Build GRCh38
3986621 c.32T>G p.L11R Chr.12: 10909267-10909267 on Build GRCh38
934932 c.121C>T p.L41L Chr.12: 10909178-10909178 on Build GRCh38
4038633 c.90C>T p.I30I Chr.12: 10909209-10909209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912021465 Chr.12: 10909387-10909387 on Build GRCh38
rs145681875 Chr.12: 10909429-10909429 on Build GRCh38
rs947761530 Chr.12: 10909493-10909493 on Build GRCh38
rs764296940 Chr.12: 10909298-10909298 on Build GRCh38
rs746483561 Chr.12: 10909257-10909257 on Build GRCh38
rs916772358 Chr.12: 10909437-10909437 on Build GRCh38
rs895385814 Chr.12: 10909215-10909215 on Build GRCh38
rs1039537010 Chr.12: 10909528-10909528 on Build GRCh38
rs570282750 Chr.12: 10909468-10909468 on Build GRCh38
rs528117363 Chr.12: 10909351-10909351 on Build GRCh38
rs554559478 Chr.12: 10909628-10909628 on Build GRCh38
rs187860064 Chr.12: 10909503-10909503 on Build GRCh38
rs1029561426 Chr.12: 10909578-10909578 on Build GRCh38
rs928225016 Chr.12: 10909473-10909473 on Build GRCh38
rs530699969 Chr.12: 10909275-10909275 on Build GRCh38
rs558907676 Chr.12: 10909322-10909322 on Build GRCh38
rs770569229 Chr.12: 10909271-10909271 on Build GRCh38
rs35172210 Chr.12: 10909566-10909567 on Build GRCh38
rs1046731178 Chr.12: 10909513-10909513 on Build GRCh38
rs940836296 Chr.12: 10909512-10909512 on Build GRCh38
rs995093910 Chr.12: 10909639-10909639 on Build GRCh38
rs888217569 Chr.12: 10909259-10909259 on Build GRCh38
rs773002052 Chr.12: 10909220-10909220 on Build GRCh38
rs757433207 Chr.12: 10909315-10909315 on Build GRCh38
rs953407295 Chr.12: 10909376-10909376 on Build GRCh38
rs751769448 Chr.12: 10909221-10909221 on Build GRCh38
rs761791825 Chr.12: 10909556-10909556 on Build GRCh38
rs770288010 Chr.12: 10909237-10909237 on Build GRCh38
rs139430284 Chr.12: 10909235-10909235 on Build GRCh38
rs545867696 Chr.12: 10909645-10909645 on Build GRCh38
rs113595231 Chr.12: 10909302-10909302 on Build GRCh38
rs987558219 Chr.12: 10909385-10909385 on Build GRCh38
rs923836831 Chr.12: 10909465-10909465 on Build GRCh38
rs765247180 Chr.12: 10909615-10909615 on Build GRCh38
rs142346419 Chr.12: 10909583-10909583 on Build GRCh38
rs141818191 Chr.12: 10909357-10909357 on Build GRCh38
rs530334570 Chr.12: 10909394-10909394 on Build GRCh38
rs560091714 Chr.12: 10909281-10909281 on Build GRCh38
rs977983101 Chr.12: 10909463-10909463 on Build GRCh38
rs999512904 Chr.12: 10909591-10909591 on Build GRCh38
rs183174341 Chr.12: 10909471-10909471 on Build GRCh38
rs760277052 Chr.12: 10909270-10909270 on Build GRCh38
rs749835630 Chr.12: 10909332-10909332 on Build GRCh38
rs759310016 Chr.12: 10909282-10909282 on Build GRCh38
rs998037430 Chr.12: 10909572-10909572 on Build GRCh38
rs888185885 Chr.12: 10909515-10909515 on Build GRCh38
rs1008938453 Chr.12: 10909661-10909661 on Build GRCh38
rs757456465 Chr.12: 10909228-10909228 on Build GRCh38
rs923993190 Chr.12: 10909412-10909412 on Build GRCh38
rs191331862 Chr.12: 10909380-10909380 on Build GRCh38
rs370449799 Chr.12: 10909269-10909269 on Build GRCh38
rs751607662 Chr.12: 10909306-10909306 on Build GRCh38
rs756530072 Chr.12: 10909327-10909327 on Build GRCh38
rs1005737601 Chr.12: 10909522-10909522 on Build GRCh38
rs201819234 Chr.12: 10909289-10909289 on Build GRCh38
rs899697146 Chr.12: 10909552-10909552 on Build GRCh38
rs936620912 Chr.12: 10909467-10909467 on Build GRCh38
rs370825050 Chr.12: 10909330-10909330 on Build GRCh38
rs774611954 Chr.12: 10909348-10909348 on Build GRCh38
rs899382824 Chr.12: 10909619-10909619 on Build GRCh38
rs767672649 Chr.12: 10909230-10909230 on Build GRCh38
rs749785671 Chr.12: 10909240-10909240 on Build GRCh38
rs377733965 Chr.12: 10909287-10909287 on Build GRCh38
rs147654800 Chr.12: 10909497-10909497 on Build GRCh38
rs780607686 Chr.12: 10909239-10909239 on Build GRCh38
rs1018554983 Chr.12: 10909273-10909273 on Build GRCh38
rs989644186 Chr.12: 10909435-10909435 on Build GRCh38
rs750608422 Chr.12: 10909323-10909323 on Build GRCh38
rs765108797 Chr.12: 10909283-10909283 on Build GRCh38
rs906706911 Chr.12: 10909219-10909219 on Build GRCh38
rs879288594 Chr.12: 10909469-10909469 on Build GRCh38
rs1028960670 Chr.12: 10909641-10909641 on Build GRCh38
rs958104967 Chr.12: 10909685-10909685 on Build GRCh38
rs149629304 Chr.12: 10909248-10909248 on Build GRCh38
rs771622960 Chr.12: 10909243-10909243 on Build GRCh38

View Full Product Details