Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12361184-12361726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORCS5
Entrez Gene ID
118426
Gene Name
BLOC-1 related complex subunit 5
Gene Aliases
LOH12CR1, LOH1CR12
UniGene
Hs.720779
Gene Chromosome Location
Chr.12: 12357078-12469562 on Build GRCh38
Transcripts
NM_001300742.1 NM_058169.4 XM_011520551.2 XM_011520552.2 XM_011520553.2 XM_017018778.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7078166 c.122C>T p.S41F Chr.12: 12361269-12361269 on Build GRCh38
71450 c.134G>A p.R45Q Chr.12: 12361281-12361281 on Build GRCh38
5792179 c.75C>G p.A25A Chr.12: 12361222-12361222 on Build GRCh38
5840489 c.151C>T p.P51S Chr.12: 12361298-12361298 on Build GRCh38
2227215 c.188A>G p.Q63R Chr.12: 12361335-12361335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022928192 Chr.12: 12361685-12361685 on Build GRCh38
rs932060674 Chr.12: 12361295-12361295 on Build GRCh38
rs760546323 Chr.12: 12361437-12361438 on Build GRCh38
rs144877374 Chr.12: 12361281-12361281 on Build GRCh38
rs1056413788 Chr.12: 12361679-12361679 on Build GRCh38
rs763367849 Chr.12: 12361274-12361274 on Build GRCh38
rs921188233 Chr.12: 12361559-12361559 on Build GRCh38
rs368910873 Chr.12: 12361391-12361391 on Build GRCh38
rs774793904 Chr.12: 12361373-12361373 on Build GRCh38
rs368141493 Chr.12: 12361300-12361300 on Build GRCh38
rs745945846 Chr.12: 12361356-12361356 on Build GRCh38
rs371653734 Chr.12: 12361329-12361329 on Build GRCh38
rs562239513 Chr.12: 12361325-12361325 on Build GRCh38
rs896688870 Chr.12: 12361595-12361595 on Build GRCh38
rs568552400 Chr.12: 12361670-12361670 on Build GRCh38
rs764545431 Chr.12: 12361279-12361279 on Build GRCh38
rs911979192 Chr.12: 12361317-12361317 on Build GRCh38
rs905241681 Chr.12: 12361570-12361570 on Build GRCh38
rs1004405517 Chr.12: 12361645-12361645 on Build GRCh38
rs3741795 Chr.12: 12361269-12361269 on Build GRCh38
rs200415074 Chr.12: 12361294-12361294 on Build GRCh38
rs773102903 Chr.12: 12361223-12361223 on Build GRCh38
rs768127114 Chr.12: 12361377-12361377 on Build GRCh38
rs753453049 Chr.12: 12361438-12361438 on Build GRCh38
rs953258274 Chr.12: 12361429-12361429 on Build GRCh38
rs144763041 Chr.12: 12361521-12361521 on Build GRCh38
rs946162707 Chr.12: 12361616-12361616 on Build GRCh38
rs963302991 Chr.12: 12361664-12361664 on Build GRCh38
rs1000877005 Chr.12: 12361390-12361390 on Build GRCh38
rs774113374 Chr.12: 12361663-12361663 on Build GRCh38
rs748886713 Chr.12: 12361602-12361602 on Build GRCh38
rs185806839 Chr.12: 12361528-12361528 on Build GRCh38
rs769973474 Chr.12: 12361358-12361358 on Build GRCh38
rs1014435910 Chr.12: 12361428-12361428 on Build GRCh38
rs771020065 Chr.12: 12361236-12361236 on Build GRCh38
rs764555082 Chr.12: 12361370-12361370 on Build GRCh38
rs758235440 Chr.12: 12361357-12361358 on Build GRCh38
rs1014533484 Chr.12: 12361398-12361398 on Build GRCh38
rs765440523 Chr.12: 12361252-12361252 on Build GRCh38
rs892502488 Chr.12: 12361262-12361262 on Build GRCh38
rs961763457 Chr.12: 12361417-12361417 on Build GRCh38
rs1024039140 Chr.12: 12361439-12361439 on Build GRCh38
rs759756604 Chr.12: 12361248-12361248 on Build GRCh38
rs973497642 Chr.12: 12361427-12361427 on Build GRCh38
rs577623740 Chr.12: 12361285-12361285 on Build GRCh38
rs550030615 Chr.12: 12361642-12361642 on Build GRCh38
rs538024135 Chr.12: 12361637-12361638 on Build GRCh38
rs781517205 Chr.12: 12361324-12361324 on Build GRCh38
rs1002244850 Chr.12: 12361581-12361582 on Build GRCh38
rs764889107 Chr.12: 12361547-12361547 on Build GRCh38
rs944597270 Chr.12: 12361504-12361504 on Build GRCh38
rs746980108 Chr.12: 12361337-12361337 on Build GRCh38
rs752317683 Chr.12: 12361652-12361652 on Build GRCh38
rs368305277 Chr.12: 12361222-12361222 on Build GRCh38
rs372230725 Chr.12: 12361286-12361286 on Build GRCh38
rs1051762771 Chr.12: 12361308-12361308 on Build GRCh38
rs140151931 Chr.12: 12361211-12361211 on Build GRCh38
rs755239057 Chr.12: 12361487-12361487 on Build GRCh38
rs149400513 Chr.12: 12361362-12361362 on Build GRCh38
rs529268081 Chr.12: 12361652-12361652 on Build GRCh38
rs181291805 Chr.12: 12361361-12361361 on Build GRCh38
rs34063679 Chr.12: 12361264-12361264 on Build GRCh38
rs986613349 Chr.12: 12361590-12361590 on Build GRCh38
rs762427206 Chr.12: 12361376-12361376 on Build GRCh38
rs560058000 Chr.12: 12361444-12361444 on Build GRCh38
rs770786823 Chr.12: 12361339-12361339 on Build GRCh38
rs3741794 Chr.12: 12361543-12361543 on Build GRCh38
rs558171138 Chr.12: 12361238-12361238 on Build GRCh38
rs747141622 Chr.12: 12361229-12361229 on Build GRCh38
rs894616120 Chr.12: 12361395-12361395 on Build GRCh38
rs911766857 Chr.12: 12361489-12361489 on Build GRCh38
rs776703671 Chr.12: 12361340-12361340 on Build GRCh38
rs531533835 Chr.12: 12361567-12361567 on Build GRCh38
rs201288073 Chr.12: 12361321-12361321 on Build GRCh38
rs925984471 Chr.12: 12361656-12361656 on Build GRCh38
rs776550512 Chr.12: 12361371-12361371 on Build GRCh38
rs894749829 Chr.12: 12361702-12361702 on Build GRCh38
rs955169517 Chr.12: 12361585-12361585 on Build GRCh38
rs770706414 Chr.12: 12361606-12361606 on Build GRCh38
rs1056552041 Chr.12: 12361592-12361592 on Build GRCh38
rs752086313 Chr.12: 12361280-12361280 on Build GRCh38

View Full Product Details