Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22065238-22065768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMAS
Entrez Gene ID
55907
Gene Name
cytidine monophosphate N-acetylneuraminic acid synthetase
Gene Aliases
CSS
UniGene
Hs.311346
Gene Chromosome Location
Chr.12: 22046174-22065674 on Build GRCh38
Transcripts
NM_018686.5 NR_135117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1201481 c.1276G>A p.E426K Chr.12: 22065282-22065282 on Build GRCh38
5184749 c.1305G>A p.*435* Chr.12: 22065311-22065311 on Build GRCh38
279948 c.1299A>G p.Q433Q Chr.12: 22065305-22065305 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1031123334 Chr.12: 22065349-22065349 on Build GRCh38
rs931064545 Chr.12: 22065464-22065464 on Build GRCh38
rs535948379 Chr.12: 22065724-22065724 on Build GRCh38
rs947943748 Chr.12: 22065490-22065491 on Build GRCh38
rs958016801 Chr.12: 22065377-22065377 on Build GRCh38
rs868284821 Chr.12: 22065502-22065502 on Build GRCh38
rs140441792 Chr.12: 22065489-22065489 on Build GRCh38
rs953725316 Chr.12: 22065406-22065406 on Build GRCh38
rs750284542 Chr.12: 22065341-22065341 on Build GRCh38
rs1019631103 Chr.12: 22065528-22065528 on Build GRCh38
rs767473001 Chr.12: 22065339-22065339 on Build GRCh38
rs925442865 Chr.12: 22065702-22065702 on Build GRCh38
rs200809094 Chr.12: 22065333-22065333 on Build GRCh38
rs199500244 Chr.12: 22065276-22065276 on Build GRCh38
rs879525520 Chr.12: 22065632-22065633 on Build GRCh38
rs766635784 Chr.12: 22065356-22065356 on Build GRCh38
rs770948115 Chr.12: 22065347-22065349 on Build GRCh38
rs1001508930 Chr.12: 22065420-22065420 on Build GRCh38
rs1045652829 Chr.12: 22065399-22065399 on Build GRCh38
rs772573747 Chr.12: 22065269-22065269 on Build GRCh38
rs779488724 Chr.12: 22065611-22065611 on Build GRCh38
rs908466351 Chr.12: 22065603-22065603 on Build GRCh38
rs182578956 Chr.12: 22065689-22065689 on Build GRCh38
rs745847087 Chr.12: 22065566-22065566 on Build GRCh38
rs1019227309 Chr.12: 22065725-22065726 on Build GRCh38
rs756439530 Chr.12: 22065570-22065570 on Build GRCh38
rs528332780 Chr.12: 22065424-22065424 on Build GRCh38
rs14472 Chr.12: 22065449-22065449 on Build GRCh38
rs773508769 Chr.12: 22065273-22065273 on Build GRCh38
rs895013720 Chr.12: 22065313-22065313 on Build GRCh38
rs964487825 Chr.12: 22065533-22065533 on Build GRCh38
rs1011004305 Chr.12: 22065691-22065691 on Build GRCh38
rs940890700 Chr.12: 22065427-22065428 on Build GRCh38
rs1052677387 Chr.12: 22065432-22065432 on Build GRCh38
rs1009549510 Chr.12: 22065721-22065721 on Build GRCh38
rs528004368 Chr.12: 22065475-22065475 on Build GRCh38
rs7606 Chr.12: 22065633-22065633 on Build GRCh38
rs549079807 Chr.12: 22065738-22065738 on Build GRCh38
rs952614688 Chr.12: 22065585-22065585 on Build GRCh38
rs542208860 Chr.12: 22065357-22065357 on Build GRCh38
rs996967447 Chr.12: 22065534-22065534 on Build GRCh38
rs983096330 Chr.12: 22065593-22065593 on Build GRCh38
rs972448971 Chr.12: 22065430-22065430 on Build GRCh38
rs376139711 Chr.12: 22065319-22065319 on Build GRCh38
rs374284736 Chr.12: 22065320-22065320 on Build GRCh38
rs948461764 Chr.12: 22065652-22065652 on Build GRCh38
rs762649542 Chr.12: 22065314-22065314 on Build GRCh38
rs369223961 Chr.12: 22065316-22065316 on Build GRCh38
rs959654687 Chr.12: 22065605-22065605 on Build GRCh38
rs561316602 Chr.12: 22065401-22065401 on Build GRCh38
rs1048293777 Chr.12: 22065281-22065281 on Build GRCh38
rs1048499050 Chr.12: 22065488-22065488 on Build GRCh38
rs953427443 Chr.12: 22065715-22065715 on Build GRCh38
rs149995748 Chr.12: 22065568-22065568 on Build GRCh38
rs145237537 Chr.12: 22065694-22065694 on Build GRCh38
rs755871860 Chr.12: 22065350-22065350 on Build GRCh38
rs775263049 Chr.12: 22065298-22065298 on Build GRCh38
rs904050654 Chr.12: 22065404-22065404 on Build GRCh38
rs915666882 Chr.12: 22065353-22065353 on Build GRCh38
rs912028659 Chr.12: 22065424-22065425 on Build GRCh38
rs999594798 Chr.12: 22065348-22065348 on Build GRCh38
rs139686165 Chr.12: 22065382-22065382 on Build GRCh38
rs1029380852 Chr.12: 22065706-22065706 on Build GRCh38
rs1027213357 Chr.12: 22065575-22065575 on Build GRCh38
rs769615088 Chr.12: 22065288-22065288 on Build GRCh38

View Full Product Details