Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48925193-48925694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP11
Entrez Gene ID
51303
Gene Name
FK506 binding protein 11
Gene Aliases
FKBP19
UniGene
Hs.655103
Gene Chromosome Location
Chr.12: 48921959-48925547 on Build GRCh38
Transcripts
NM_001143782.1 NM_016594.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4042304 c.2T>G p.M1R Chr.12: 48925427-48925427 on Build GRCh38
5649152 c.7C>T p.L3L Chr.12: 48925422-48925422 on Build GRCh38
7368317 c.92C>A p.T31N Chr.12: 48925337-48925337 on Build GRCh38
5649153 c.7C>T p.L3L Chr.12: 48925422-48925422 on Build GRCh38
2006374 c.92C>A p.T31N Chr.12: 48925337-48925337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754136058 Chr.12: 48925327-48925327 on Build GRCh38
rs920790385 Chr.12: 48925475-48925475 on Build GRCh38
rs769672183 Chr.12: 48925359-48925359 on Build GRCh38
rs777141949 Chr.12: 48925375-48925376 on Build GRCh38
rs150129909 Chr.12: 48925669-48925669 on Build GRCh38
rs867465733 Chr.12: 48925468-48925468 on Build GRCh38
rs954246265 Chr.12: 48925274-48925274 on Build GRCh38
rs750683237 Chr.12: 48925313-48925314 on Build GRCh38
rs985592651 Chr.12: 48925302-48925302 on Build GRCh38
rs917206132 Chr.12: 48925595-48925595 on Build GRCh38
rs151002970 Chr.12: 48925592-48925592 on Build GRCh38
rs371280157 Chr.12: 48925436-48925436 on Build GRCh38
rs750527804 Chr.12: 48925295-48925295 on Build GRCh38
rs956211120 Chr.12: 48925251-48925251 on Build GRCh38
rs753992473 Chr.12: 48925270-48925270 on Build GRCh38
rs755381017 Chr.12: 48925329-48925329 on Build GRCh38
rs779926849 Chr.12: 48925474-48925474 on Build GRCh38
rs868149948 Chr.12: 48925370-48925370 on Build GRCh38
rs769584236 Chr.12: 48925473-48925473 on Build GRCh38
rs878983141 Chr.12: 48925482-48925482 on Build GRCh38
rs1044236541 Chr.12: 48925451-48925451 on Build GRCh38
rs867225503 Chr.12: 48925403-48925403 on Build GRCh38
rs940892039 Chr.12: 48925239-48925239 on Build GRCh38
rs1000288735 Chr.12: 48925458-48925458 on Build GRCh38
rs765785005 Chr.12: 48925309-48925319 on Build GRCh38
rs770487052 Chr.12: 48925351-48925351 on Build GRCh38
rs529271511 Chr.12: 48925311-48925311 on Build GRCh38
rs954290029 Chr.12: 48925504-48925504 on Build GRCh38
rs1031684634 Chr.12: 48925246-48925246 on Build GRCh38
rs948626807 Chr.12: 48925611-48925611 on Build GRCh38
rs930766440 Chr.12: 48925478-48925478 on Build GRCh38
rs1056806383 Chr.12: 48925553-48925553 on Build GRCh38
rs12829682 Chr.12: 48925253-48925253 on Build GRCh38
rs987459172 Chr.12: 48925662-48925662 on Build GRCh38
rs531693483 Chr.12: 48925373-48925373 on Build GRCh38
rs767477429 Chr.12: 48925499-48925499 on Build GRCh38
rs546294814 Chr.12: 48925275-48925275 on Build GRCh38
rs532123518 Chr.12: 48925552-48925552 on Build GRCh38
rs998235211 Chr.12: 48925665-48925665 on Build GRCh38
rs545408112 Chr.12: 48925609-48925609 on Build GRCh38
rs758861575 Chr.12: 48925331-48925332 on Build GRCh38
rs780678259 Chr.12: 48925443-48925443 on Build GRCh38
rs762924188 Chr.12: 48925369-48925369 on Build GRCh38
rs767446795 Chr.12: 48925282-48925282 on Build GRCh38
rs770717715 Chr.12: 48925376-48925384 on Build GRCh38
rs766484996 Chr.12: 48925377-48925377 on Build GRCh38
rs890529776 Chr.12: 48925238-48925238 on Build GRCh38
rs779366277 Chr.12: 48925335-48925335 on Build GRCh38
rs963183077 Chr.12: 48925528-48925528 on Build GRCh38
rs745468649 Chr.12: 48925466-48925466 on Build GRCh38
rs796984800 Chr.12: 48925458-48925485 on Build GRCh38
rs745636172 Chr.12: 48925358-48925358 on Build GRCh38
rs1051807458 Chr.12: 48925237-48925237 on Build GRCh38
rs932015132 Chr.12: 48925389-48925389 on Build GRCh38
rs918990685 Chr.12: 48925567-48925567 on Build GRCh38
rs959798896 Chr.12: 48925344-48925344 on Build GRCh38
rs1036511736 Chr.12: 48925240-48925240 on Build GRCh38
rs776223377 Chr.12: 48925250-48925250 on Build GRCh38
rs562914452 Chr.12: 48925337-48925337 on Build GRCh38
rs1028903996 Chr.12: 48925639-48925639 on Build GRCh38
rs780943424 Chr.12: 48925332-48925332 on Build GRCh38
rs973192520 Chr.12: 48925529-48925529 on Build GRCh38
rs529865967 Chr.12: 48925430-48925430 on Build GRCh38
rs775448474 Chr.12: 48925368-48925368 on Build GRCh38
rs2272312 Chr.12: 48925487-48925487 on Build GRCh38
rs752374632 Chr.12: 48925363-48925364 on Build GRCh38
rs766639419 Chr.12: 48925640-48925640 on Build GRCh38
rs776403275 Chr.12: 48925352-48925352 on Build GRCh38
rs765540935 Chr.12: 48925415-48925415 on Build GRCh38
rs1044296924 Chr.12: 48925613-48925613 on Build GRCh38
rs775618336 Chr.12: 48925270-48925270 on Build GRCh38
rs774717756 Chr.12: 48925456-48925456 on Build GRCh38
rs772814777 Chr.12: 48925215-48925215 on Build GRCh38
rs376593036 Chr.12: 48925420-48925420 on Build GRCh38
rs891855858 Chr.12: 48925661-48925661 on Build GRCh38
rs766687910 Chr.12: 48925324-48925324 on Build GRCh38
rs760871503 Chr.12: 48925634-48925634 on Build GRCh38
rs755869398 Chr.12: 48925376-48925378 on Build GRCh38
rs1017063257 Chr.12: 48925527-48925527 on Build GRCh38
rs929012544 Chr.12: 48925580-48925580 on Build GRCh38
rs777093017 Chr.12: 48925341-48925341 on Build GRCh38
rs371849381 Chr.12: 48925419-48925419 on Build GRCh38
rs918930828 Chr.12: 48925375-48925375 on Build GRCh38
rs762210425 Chr.12: 48925289-48925290 on Build GRCh38
rs537800981 Chr.12: 48925654-48925654 on Build GRCh38
rs985625431 Chr.12: 48925517-48925517 on Build GRCh38
rs928929024 Chr.12: 48925404-48925404 on Build GRCh38
rs1010772714 Chr.12: 48925538-48925538 on Build GRCh38
rs540792153 Chr.12: 48925216-48925216 on Build GRCh38
rs765253540 Chr.12: 48925255-48925255 on Build GRCh38
rs200072412 Chr.12: 48925417-48925417 on Build GRCh38
rs751022211 Chr.12: 48925433-48925433 on Build GRCh38
rs745622342 Chr.12: 48925376-48925381 on Build GRCh38
rs1053236077 Chr.12: 48925650-48925650 on Build GRCh38
rs145495143 Chr.12: 48925372-48925372 on Build GRCh38
rs374838023 Chr.12: 48925410-48925410 on Build GRCh38
rs140839248 Chr.12: 48925620-48925620 on Build GRCh38
rs74591075 Chr.12: 48925231-48925231 on Build GRCh38
rs748618203 Chr.12: 48925336-48925336 on Build GRCh38
rs746534787 Chr.12: 48925346-48925346 on Build GRCh38
rs148830127 Chr.12: 48925411-48925411 on Build GRCh38

View Full Product Details