Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2228904-2229444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKI
Entrez Gene ID
6497
Gene Name
SKI proto-oncogene
Gene Aliases
SGS, SKV
UniGene
Hs.656507
Gene Chromosome Location
Chr.1: 2228695-2310213 on Build GRCh38
Transcripts
NM_003036.3 XM_005244775.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4494721 c.438C>T p.L146L Chr.1: 2229204-2229204 on Build GRCh38
6374174 c.379C>A p.L127I Chr.1: 2229145-2229145 on Build GRCh38
334250 c.302G>T p.C101F Chr.1: 2229068-2229068 on Build GRCh38
7390007 c.676G>C p.G226R Chr.1: 2229442-2229442 on Build GRCh38
3770240 c.419A>T p.N140I Chr.1: 2229185-2229185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886039149 Chr.1: 2229077-2229077 on Build GRCh38
rs149898447 Chr.1: 2229222-2229222 on Build GRCh38
rs867726673 Chr.1: 2229229-2229229 on Build GRCh38
rs758717415 Chr.1: 2228998-2228998 on Build GRCh38
rs201144990 Chr.1: 2229090-2229090 on Build GRCh38
rs766957830 Chr.1: 2229309-2229309 on Build GRCh38
rs564817940 Chr.1: 2229084-2229084 on Build GRCh38
rs1027820020 Chr.1: 2228978-2228978 on Build GRCh38
rs761071712 Chr.1: 2229294-2229294 on Build GRCh38
rs764061670 Chr.1: 2228943-2228943 on Build GRCh38
rs757121499 Chr.1: 2229351-2229351 on Build GRCh38
rs761138564 Chr.1: 2229388-2229388 on Build GRCh38
rs368195821 Chr.1: 2229078-2229078 on Build GRCh38
rs752031916 Chr.1: 2229102-2229102 on Build GRCh38
rs542970011 Chr.1: 2229396-2229396 on Build GRCh38
rs994969301 Chr.1: 2228975-2228975 on Build GRCh38
rs905113274 Chr.1: 2228942-2228947 on Build GRCh38
rs201590284 Chr.1: 2229013-2229013 on Build GRCh38
rs528051602 Chr.1: 2229033-2229033 on Build GRCh38
rs559020511 Chr.1: 2229230-2229230 on Build GRCh38
rs542659326 Chr.1: 2229008-2229008 on Build GRCh38
rs746882046 Chr.1: 2229347-2229347 on Build GRCh38
rs762117658 Chr.1: 2229277-2229277 on Build GRCh38
rs139179843 Chr.1: 2229406-2229406 on Build GRCh38
rs771747350 Chr.1: 2229288-2229288 on Build GRCh38
rs546957439 Chr.1: 2229213-2229213 on Build GRCh38
rs764373577 Chr.1: 2229096-2229096 on Build GRCh38
rs781231059 Chr.1: 2229352-2229352 on Build GRCh38
rs863223722 Chr.1: 2229305-2229305 on Build GRCh38
rs115746142 Chr.1: 2229060-2229060 on Build GRCh38
rs772192813 Chr.1: 2229069-2229069 on Build GRCh38
rs573863671 Chr.1: 2229334-2229334 on Build GRCh38
rs753070477 Chr.1: 2228993-2228993 on Build GRCh38
rs770970008 Chr.1: 2229247-2229247 on Build GRCh38
rs376575233 Chr.1: 2229047-2229047 on Build GRCh38
rs768733290 Chr.1: 2229264-2229264 on Build GRCh38
rs769506718 Chr.1: 2228929-2228929 on Build GRCh38
rs538703800 Chr.1: 2229216-2229216 on Build GRCh38
rs753251990 Chr.1: 2229319-2229319 on Build GRCh38
rs988601938 Chr.1: 2229048-2229048 on Build GRCh38
rs775713153 Chr.1: 2229172-2229172 on Build GRCh38
rs377092419 Chr.1: 2229328-2229328 on Build GRCh38
rs757751828 Chr.1: 2229107-2229107 on Build GRCh38
rs776829569 Chr.1: 2229075-2229075 on Build GRCh38
rs979256214 Chr.1: 2229086-2229086 on Build GRCh38
rs977288994 Chr.1: 2229371-2229371 on Build GRCh38
rs750951866 Chr.1: 2229195-2229195 on Build GRCh38
rs869312901 Chr.1: 2229115-2229115 on Build GRCh38
rs867327122 Chr.1: 2228928-2228928 on Build GRCh38
rs765605691 Chr.1: 2228992-2228992 on Build GRCh38
rs776226534 Chr.1: 2229033-2229035 on Build GRCh38
rs373291824 Chr.1: 2229255-2229255 on Build GRCh38
rs748004059 Chr.1: 2229159-2229159 on Build GRCh38
rs775345694 Chr.1: 2228942-2228942 on Build GRCh38
rs746731483 Chr.1: 2229006-2229008 on Build GRCh38
rs762205602 Chr.1: 2229188-2229188 on Build GRCh38
rs371061756 Chr.1: 2229204-2229204 on Build GRCh38
rs398122889 Chr.1: 2229049-2229057 on Build GRCh38
rs758199656 Chr.1: 2229338-2229338 on Build GRCh38
rs952491836 Chr.1: 2229246-2229246 on Build GRCh38
rs777704639 Chr.1: 2229339-2229339 on Build GRCh38
rs758928152 Chr.1: 2229364-2229366 on Build GRCh38
rs387907303 Chr.1: 2229113-2229113 on Build GRCh38
rs749071609 Chr.1: 2229044-2229044 on Build GRCh38
rs398122914 Chr.1: 2229046-2229057 on Build GRCh38
rs968009270 Chr.1: 2229401-2229401 on Build GRCh38
rs968117294 Chr.1: 2229411-2229411 on Build GRCh38
rs869025525 Chr.1: 2229118-2229118 on Build GRCh38
rs763083721 Chr.1: 2229177-2229177 on Build GRCh38
rs772336192 Chr.1: 2229377-2229377 on Build GRCh38
rs778608658 Chr.1: 2229021-2229021 on Build GRCh38
rs937379586 Chr.1: 2229165-2229165 on Build GRCh38
rs778291636 Chr.1: 2229007-2229007 on Build GRCh38
rs369090536 Chr.1: 2229144-2229144 on Build GRCh38
rs878854599 Chr.1: 2229253-2229253 on Build GRCh38
rs1026586604 Chr.1: 2229231-2229231 on Build GRCh38
rs767973122 Chr.1: 2229189-2229189 on Build GRCh38
rs768360897 Chr.1: 2229051-2229051 on Build GRCh38
rs182513685 Chr.1: 2229282-2229282 on Build GRCh38
rs774588048 Chr.1: 2229270-2229270 on Build GRCh38
rs150306072 Chr.1: 2229162-2229162 on Build GRCh38
rs1013962872 Chr.1: 2229186-2229186 on Build GRCh38
rs375024753 Chr.1: 2229126-2229126 on Build GRCh38
rs763316588 Chr.1: 2229093-2229093 on Build GRCh38
rs756778048 Chr.1: 2228982-2228982 on Build GRCh38
rs569206617 Chr.1: 2229237-2229237 on Build GRCh38
rs774502280 Chr.1: 2229357-2229357 on Build GRCh38
rs765800099 Chr.1: 2229207-2229207 on Build GRCh38
rs777792553 Chr.1: 2229234-2229234 on Build GRCh38
rs774187595 Chr.1: 2228949-2228949 on Build GRCh38
rs754036418 Chr.1: 2228956-2228956 on Build GRCh38
rs144874401 Chr.1: 2229183-2229183 on Build GRCh38
rs768420010 Chr.1: 2229150-2229150 on Build GRCh38
rs764842901 Chr.1: 2229408-2229408 on Build GRCh38
rs897964442 Chr.1: 2228958-2228958 on Build GRCh38
rs748376935 Chr.1: 2229373-2229373 on Build GRCh38
rs752321098 Chr.1: 2229418-2229418 on Build GRCh38
rs770423509 Chr.1: 2229005-2229006 on Build GRCh38
rs28384811 Chr.1: 2228951-2228951 on Build GRCh38
rs769774481 Chr.1: 2229171-2229171 on Build GRCh38
rs749291332 Chr.1: 2229356-2229356 on Build GRCh38
rs863223723 Chr.1: 2229112-2229112 on Build GRCh38
rs745444860 Chr.1: 2229029-2229029 on Build GRCh38
rs773540675 Chr.1: 2229385-2229385 on Build GRCh38
rs766569890 Chr.1: 2228954-2228954 on Build GRCh38
rs753444459 Chr.1: 2229208-2229208 on Build GRCh38
rs773310002 Chr.1: 2229168-2229168 on Build GRCh38
rs886039166 Chr.1: 2229017-2229018 on Build GRCh38
rs863223724 Chr.1: 2229265-2229266 on Build GRCh38
rs749438586 Chr.1: 2229261-2229261 on Build GRCh38
rs771298442 Chr.1: 2229391-2229391 on Build GRCh38
rs566906261 Chr.1: 2229219-2229219 on Build GRCh38
rs1021005276 Chr.1: 2229405-2229405 on Build GRCh38
rs780679449 Chr.1: 2229024-2229024 on Build GRCh38
rs41310345 Chr.1: 2229375-2229375 on Build GRCh38
rs777216862 Chr.1: 2229403-2229403 on Build GRCh38
rs557157137 Chr.1: 2229324-2229324 on Build GRCh38
rs567637491 Chr.1: 2229127-2229127 on Build GRCh38
rs113737327 Chr.1: 2229054-2229054 on Build GRCh38
rs781703488 Chr.1: 2229117-2229117 on Build GRCh38
rs371497152 Chr.1: 2229147-2229147 on Build GRCh38
rs752411031 Chr.1: 2229333-2229333 on Build GRCh38
rs374862468 Chr.1: 2228990-2228990 on Build GRCh38

View Full Product Details