Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6025668-6026194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNAB2
Entrez Gene ID
8514
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 2
Gene Aliases
AKR6A5, HKvbeta2, HKvbeta2.1, HKvbeta2.2, KCNA2B, KV-BETA-2
UniGene
Hs.440497
Gene Chromosome Location
Chr.1: 5992298-6101193 on Build GRCh38
Transcripts
NM_001199861.1 NM_003636.3 NM_172130.2 XM_005263514.2 XM_011542321.2 XM_017002618.1 XM_017002619.1 XM_017002621.1

SNP Detail

SNP ID Chromosome Location
rs959641182 Chr.1: 6025896-6025896 on Build GRCh38
rs952903413 Chr.1: 6025850-6025850 on Build GRCh38
rs139985118 Chr.1: 6025991-6025991 on Build GRCh38
rs539283718 Chr.1: 6025915-6025915 on Build GRCh38
rs901889404 Chr.1: 6025824-6025824 on Build GRCh38
rs1015836602 Chr.1: 6025864-6025864 on Build GRCh38
rs990910527 Chr.1: 6025903-6025903 on Build GRCh38
rs944601731 Chr.1: 6025959-6025959 on Build GRCh38
rs753894626 Chr.1: 6026088-6026088 on Build GRCh38
rs972518201 Chr.1: 6025916-6025916 on Build GRCh38
rs927928598 Chr.1: 6025889-6025889 on Build GRCh38
rs901918630 Chr.1: 6026075-6026075 on Build GRCh38
rs1040468406 Chr.1: 6026069-6026069 on Build GRCh38
rs982377057 Chr.1: 6025873-6025873 on Build GRCh38
rs900133228 Chr.1: 6025976-6025976 on Build GRCh38
rs946622911 Chr.1: 6025993-6025993 on Build GRCh38
rs1034761489 Chr.1: 6025926-6025926 on Build GRCh38
rs1042230938 Chr.1: 6025939-6025939 on Build GRCh38
rs867753305 Chr.1: 6026101-6026101 on Build GRCh38
rs1015590903 Chr.1: 6026166-6026166 on Build GRCh38
rs76852674 Chr.1: 6025840-6025840 on Build GRCh38
rs961556705 Chr.1: 6026049-6026049 on Build GRCh38
rs759238668 Chr.1: 6025816-6025816 on Build GRCh38
rs761990363 Chr.1: 6025760-6025760 on Build GRCh38
rs1035712164 Chr.1: 6026107-6026107 on Build GRCh38
rs76110241 Chr.1: 6025987-6025987 on Build GRCh38
rs1042280790 Chr.1: 6025696-6025696 on Build GRCh38
rs1040354629 Chr.1: 6025780-6025780 on Build GRCh38
rs946550757 Chr.1: 6025938-6025938 on Build GRCh38
rs961654218 Chr.1: 6025869-6025869 on Build GRCh38
rs887547275 Chr.1: 6025988-6025988 on Build GRCh38
rs959396248 Chr.1: 6026155-6026155 on Build GRCh38
rs80313586 Chr.1: 6026011-6026011 on Build GRCh38
rs567948528 Chr.1: 6026173-6026173 on Build GRCh38
rs1029224807 Chr.1: 6025838-6025838 on Build GRCh38
rs960387752 Chr.1: 6025939-6025939 on Build GRCh38
rs764184004 Chr.1: 6025996-6025996 on Build GRCh38
rs796632216 Chr.1: 6025837-6025853 on Build GRCh38
rs771842401 Chr.1: 6025827-6025827 on Build GRCh38
rs540207714 Chr.1: 6025966-6025966 on Build GRCh38
rs142230888 Chr.1: 6026004-6026004 on Build GRCh38
rs532313666 Chr.1: 6025829-6025838 on Build GRCh38
rs888012966 Chr.1: 6025842-6025842 on Build GRCh38
rs527980784 Chr.1: 6026096-6026096 on Build GRCh38
rs1018536117 Chr.1: 6025773-6025773 on Build GRCh38
rs1027825946 Chr.1: 6025844-6025844 on Build GRCh38
rs555644570 Chr.1: 6025981-6025981 on Build GRCh38
rs557700928 Chr.1: 6025942-6025942 on Build GRCh38
rs571174656 Chr.1: 6025904-6025904 on Build GRCh38
rs990464502 Chr.1: 6025965-6025965 on Build GRCh38
rs150605623 Chr.1: 6026172-6026172 on Build GRCh38
rs114010715 Chr.1: 6026082-6026082 on Build GRCh38
rs1005712377 Chr.1: 6025989-6025989 on Build GRCh38
rs946563223 Chr.1: 6025734-6025734 on Build GRCh38
rs923789075 Chr.1: 6025949-6025949 on Build GRCh38
rs774268055 Chr.1: 6025730-6025730 on Build GRCh38
rs544640676 Chr.1: 6025983-6025983 on Build GRCh38
rs753203937 Chr.1: 6026000-6026000 on Build GRCh38
rs961423040 Chr.1: 6025899-6025899 on Build GRCh38
rs573905870 Chr.1: 6025982-6025982 on Build GRCh38
rs902460588 Chr.1: 6025753-6025753 on Build GRCh38
rs984735730 Chr.1: 6025857-6025857 on Build GRCh38
rs915348841 Chr.1: 6025923-6025923 on Build GRCh38
rs1008504121 Chr.1: 6025761-6025761 on Build GRCh38
rs573069096 Chr.1: 6025958-6025958 on Build GRCh38
rs888034688 Chr.1: 6026090-6026090 on Build GRCh38
rs935162554 Chr.1: 6026044-6026044 on Build GRCh38

View Full Product Details