Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52587540-52588032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT72
Entrez Gene ID
140807
Gene Name
keratin 72
Gene Aliases
K6IRS2, K6irs, KRT6, KRT6IRS2
UniGene
Hs.662013
Gene Chromosome Location
Chr.12: 52585589-52602900 on Build GRCh38
Transcripts
NM_001146225.1 NM_001146226.1 NM_080747.2 XM_017018833.1 XM_017018834.1 XM_017018835.1 XM_017018836.1 XM_017018837.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4042881 c.1282C>T p.R428C Chr.12: 52587659-52587659 on Build GRCh38
7312874 c.1167G>A p.L389L Chr.12: 52587774-52587774 on Build GRCh38
234890 c.1293G>A p.L431L Chr.12: 52587648-52587648 on Build GRCh38
3812326 c.1276A>C p.T426P Chr.12: 52587665-52587665 on Build GRCh38
2010910 c.1232A>T p.Q411L Chr.12: 52587709-52587709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774780531 Chr.12: 52587700-52587700 on Build GRCh38
rs371280790 Chr.12: 52587805-52587805 on Build GRCh38
rs781275788 Chr.12: 52587659-52587659 on Build GRCh38
rs958924864 Chr.12: 52587678-52587678 on Build GRCh38
rs748635916 Chr.12: 52587784-52587784 on Build GRCh38
rs761447085 Chr.12: 52587606-52587607 on Build GRCh38
rs908863004 Chr.12: 52587956-52587956 on Build GRCh38
rs761584332 Chr.12: 52587634-52587634 on Build GRCh38
rs148898485 Chr.12: 52587677-52587677 on Build GRCh38
rs201218547 Chr.12: 52587734-52587734 on Build GRCh38
rs753871950 Chr.12: 52587646-52587646 on Build GRCh38
rs771059020 Chr.12: 52587582-52587582 on Build GRCh38
rs760315693 Chr.12: 52587799-52587799 on Build GRCh38
rs754938575 Chr.12: 52587648-52587648 on Build GRCh38
rs150710240 Chr.12: 52587812-52587812 on Build GRCh38
rs145882334 Chr.12: 52587758-52587758 on Build GRCh38
rs778921794 Chr.12: 52587655-52587655 on Build GRCh38
rs372915015 Chr.12: 52587882-52587882 on Build GRCh38
rs931608893 Chr.12: 52587760-52587760 on Build GRCh38
rs914619911 Chr.12: 52587705-52587705 on Build GRCh38
rs773510817 Chr.12: 52587824-52587824 on Build GRCh38
rs199938241 Chr.12: 52587586-52587586 on Build GRCh38
rs941800427 Chr.12: 52587967-52587967 on Build GRCh38
rs779025679 Chr.12: 52587816-52587816 on Build GRCh38
rs187575531 Chr.12: 52587621-52587621 on Build GRCh38
rs371927675 Chr.12: 52587605-52587605 on Build GRCh38
rs991579459 Chr.12: 52587704-52587704 on Build GRCh38
rs755068681 Chr.12: 52587814-52587814 on Build GRCh38
rs747394885 Chr.12: 52587825-52587825 on Build GRCh38
rs982965654 Chr.12: 52587872-52587872 on Build GRCh38
rs138110764 Chr.12: 52587727-52587727 on Build GRCh38
rs750021860 Chr.12: 52587641-52587641 on Build GRCh38
rs202134032 Chr.12: 52587702-52587702 on Build GRCh38
rs767089241 Chr.12: 52587638-52587638 on Build GRCh38
rs747278068 Chr.12: 52587581-52587581 on Build GRCh38
rs774866254 Chr.12: 52587773-52587773 on Build GRCh38
rs200608954 Chr.12: 52587768-52587768 on Build GRCh38
rs369224848 Chr.12: 52587823-52587823 on Build GRCh38
rs1003341263 Chr.12: 52587810-52587810 on Build GRCh38
rs7310156 Chr.12: 52587892-52587892 on Build GRCh38
rs985795530 Chr.12: 52587912-52587912 on Build GRCh38
rs764480814 Chr.12: 52587620-52587620 on Build GRCh38
rs200656498 Chr.12: 52587821-52587821 on Build GRCh38
rs372905818 Chr.12: 52587846-52587846 on Build GRCh38
rs771445358 Chr.12: 52587561-52587561 on Build GRCh38
rs532195834 Chr.12: 52587834-52587834 on Build GRCh38
rs767878464 Chr.12: 52587709-52587709 on Build GRCh38
rs750817986 Chr.12: 52587908-52587908 on Build GRCh38
rs769598194 Chr.12: 52587619-52587622 on Build GRCh38
rs746063905 Chr.12: 52587660-52587660 on Build GRCh38
rs376165041 Chr.12: 52587840-52587840 on Build GRCh38
rs775526937 Chr.12: 52587668-52587668 on Build GRCh38
rs184630537 Chr.12: 52587811-52587811 on Build GRCh38
rs141162784 Chr.12: 52587806-52587806 on Build GRCh38
rs773591423 Chr.12: 52587797-52587797 on Build GRCh38
rs775355202 Chr.12: 52587785-52587785 on Build GRCh38
rs750560666 Chr.12: 52587854-52587854 on Build GRCh38
rs956755555 Chr.12: 52587701-52587701 on Build GRCh38
rs747482687 Chr.12: 52587900-52587900 on Build GRCh38
rs750063172 Chr.12: 52587710-52587710 on Build GRCh38
rs199978375 Chr.12: 52587801-52587801 on Build GRCh38
rs777383912 Chr.12: 52587615-52587615 on Build GRCh38
rs11170183 Chr.12: 52587658-52587658 on Build GRCh38
rs764801794 Chr.12: 52587802-52587802 on Build GRCh38
rs532914124 Chr.12: 52587944-52587944 on Build GRCh38
rs541621527 Chr.12: 52587770-52587770 on Build GRCh38
rs61740862 Chr.12: 52587771-52587771 on Build GRCh38
rs146990804 Chr.12: 52587697-52587697 on Build GRCh38
rs751386463 Chr.12: 52587845-52587845 on Build GRCh38
rs766231870 Chr.12: 52587856-52587856 on Build GRCh38
rs374981564 Chr.12: 52587901-52587901 on Build GRCh38
rs769788780 Chr.12: 52587666-52587666 on Build GRCh38
rs753396101 Chr.12: 52587722-52587722 on Build GRCh38
rs774725571 Chr.12: 52587686-52587686 on Build GRCh38
rs562844592 Chr.12: 52587781-52587782 on Build GRCh38
rs773326351 Chr.12: 52587771-52587779 on Build GRCh38
rs973586178 Chr.12: 52587933-52587933 on Build GRCh38
rs7310138 Chr.12: 52587843-52587843 on Build GRCh38
rs374793445 Chr.12: 52587883-52587883 on Build GRCh38
rs758159396 Chr.12: 52587740-52587740 on Build GRCh38
rs562426021 Chr.12: 52587741-52587741 on Build GRCh38
rs755607003 Chr.12: 52587642-52587642 on Build GRCh38
rs887956957 Chr.12: 52587565-52587565 on Build GRCh38
rs140024110 Chr.12: 52587835-52587835 on Build GRCh38
rs760349724 Chr.12: 52587718-52587718 on Build GRCh38
rs200151867 Chr.12: 52587649-52587649 on Build GRCh38
rs61740861 Chr.12: 52587728-52587728 on Build GRCh38
rs776011079 Chr.12: 52587613-52587613 on Build GRCh38
rs368945209 Chr.12: 52587887-52587887 on Build GRCh38
rs962349195 Chr.12: 52587630-52587630 on Build GRCh38
rs370491283 Chr.12: 52587772-52587772 on Build GRCh38
rs777577511 Chr.12: 52587754-52587754 on Build GRCh38
rs778133919 Chr.12: 52587944-52587944 on Build GRCh38
rs372773585 Chr.12: 52587827-52587827 on Build GRCh38
rs756323393 Chr.12: 52587759-52587759 on Build GRCh38
rs1029927342 Chr.12: 52587850-52587850 on Build GRCh38
rs145640018 Chr.12: 52587669-52587669 on Build GRCh38
rs751353624 Chr.12: 52587807-52587807 on Build GRCh38
rs867251979 Chr.12: 52587623-52587623 on Build GRCh38
rs758355540 Chr.12: 52587893-52587893 on Build GRCh38
rs958736818 Chr.12: 52587822-52587822 on Build GRCh38
rs1040671594 Chr.12: 52587750-52587750 on Build GRCh38
rs769845687 Chr.12: 52587606-52587606 on Build GRCh38
rs534822174 Chr.12: 52587583-52587583 on Build GRCh38
rs776455684 Chr.12: 52587828-52587828 on Build GRCh38
rs906453378 Chr.12: 52587808-52587808 on Build GRCh38
rs368648240 Chr.12: 52587881-52587881 on Build GRCh38
rs539599708 Chr.12: 52587706-52587706 on Build GRCh38
rs766030329 Chr.12: 52587719-52587719 on Build GRCh38
rs780163323 Chr.12: 52587764-52587764 on Build GRCh38
rs1044679947 Chr.12: 52587971-52587971 on Build GRCh38
rs368767777 Chr.12: 52587800-52587800 on Build GRCh38
rs113410377 Chr.12: 52587859-52587859 on Build GRCh38
rs778351311 Chr.12: 52587898-52587898 on Build GRCh38
rs531913888 Chr.12: 52587941-52587941 on Build GRCh38
rs763403292 Chr.12: 52587617-52587617 on Build GRCh38
rs1037382114 Chr.12: 52587791-52587791 on Build GRCh38

View Full Product Details